Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98823072-98823578 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
UniGene
Hs.469398
Gene Chromosome Location
Chr.2: 98793846-98936221 on Build GRCh38
Transcripts
NM_207362.2 XM_005263923.3 XM_005263924.3 XM_005263926.3 XM_011511093.1 XM_011511094.1 XM_011511095.2 XM_011511096.1 XM_017003971.1 XM_017003972.1 XM_017003973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6378439 c.898C>T p.P300S Chr.2: 98823375-98823375 on Build GRCh38
5733486 c.739G>A p.A247T Chr.2: 98823534-98823534 on Build GRCh38
1718244 c.1081_1082CC>TT p.P361F Chr.2: 98823191-98823192 on Build GRCh38
4096560 c.1196C>T p.A399V Chr.2: 98823077-98823077 on Build GRCh38
229925 c.982C>T p.P328S Chr.2: 98823291-98823291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13014854 Chr.2: 98823327-98823327 on Build GRCh38
rs764688697 Chr.2: 98823110-98823110 on Build GRCh38
rs750845880 Chr.2: 98823196-98823196 on Build GRCh38
rs747206439 Chr.2: 98823475-98823475 on Build GRCh38
rs753438324 Chr.2: 98823103-98823103 on Build GRCh38
rs891555064 Chr.2: 98823191-98823191 on Build GRCh38
rs752693470 Chr.2: 98823386-98823386 on Build GRCh38
rs751200694 Chr.2: 98823408-98823408 on Build GRCh38
rs755478502 Chr.2: 98823232-98823232 on Build GRCh38
rs752367709 Chr.2: 98823122-98823122 on Build GRCh38
rs748390102 Chr.2: 98823149-98823149 on Build GRCh38
rs774518111 Chr.2: 98823146-98823146 on Build GRCh38
rs749829186 Chr.2: 98823214-98823214 on Build GRCh38
rs374696900 Chr.2: 98823492-98823492 on Build GRCh38
rs369975254 Chr.2: 98823535-98823535 on Build GRCh38
rs555701910 Chr.2: 98823257-98823257 on Build GRCh38
rs550777088 Chr.2: 98823161-98823161 on Build GRCh38
rs12987333 Chr.2: 98823223-98823223 on Build GRCh38
rs975613469 Chr.2: 98823092-98823092 on Build GRCh38
rs984888078 Chr.2: 98823364-98823364 on Build GRCh38
rs866521579 Chr.2: 98823368-98823368 on Build GRCh38
rs756400465 Chr.2: 98823198-98823198 on Build GRCh38
rs768881020 Chr.2: 98823144-98823144 on Build GRCh38
rs755858070 Chr.2: 98823475-98823477 on Build GRCh38
rs938347056 Chr.2: 98823177-98823177 on Build GRCh38
rs753411182 Chr.2: 98823549-98823549 on Build GRCh38
rs773278161 Chr.2: 98823279-98823279 on Build GRCh38
rs13014659 Chr.2: 98823224-98823224 on Build GRCh38
rs762906593 Chr.2: 98823520-98823520 on Build GRCh38
rs781028672 Chr.2: 98823488-98823488 on Build GRCh38
rs780423414 Chr.2: 98823209-98823209 on Build GRCh38
rs773846672 Chr.2: 98823544-98823544 on Build GRCh38
rs775039721 Chr.2: 98823380-98823380 on Build GRCh38
rs1028353193 Chr.2: 98823102-98823102 on Build GRCh38
rs149622421 Chr.2: 98823140-98823140 on Build GRCh38
rs892527485 Chr.2: 98823313-98823313 on Build GRCh38
rs201668722 Chr.2: 98823434-98823434 on Build GRCh38
rs1052455732 Chr.2: 98823273-98823273 on Build GRCh38
rs1046596559 Chr.2: 98823381-98823381 on Build GRCh38
rs745644740 Chr.2: 98823489-98823489 on Build GRCh38
rs749417186 Chr.2: 98823142-98823142 on Build GRCh38
rs1028988964 Chr.2: 98823288-98823288 on Build GRCh38
rs1011241920 Chr.2: 98823252-98823252 on Build GRCh38
rs759470403 Chr.2: 98823357-98823357 on Build GRCh38
rs368429942 Chr.2: 98823134-98823134 on Build GRCh38
rs779205825 Chr.2: 98823233-98823233 on Build GRCh38
rs925200590 Chr.2: 98823240-98823240 on Build GRCh38
rs758442092 Chr.2: 98823437-98823437 on Build GRCh38
rs3731660 Chr.2: 98823329-98823329 on Build GRCh38
rs537525119 Chr.2: 98823151-98823151 on Build GRCh38
rs767494728 Chr.2: 98823142-98823142 on Build GRCh38
rs955537794 Chr.2: 98823127-98823127 on Build GRCh38
rs551912252 Chr.2: 98823139-98823139 on Build GRCh38
rs761473727 Chr.2: 98823415-98823415 on Build GRCh38
rs754455638 Chr.2: 98823109-98823109 on Build GRCh38
rs1008651896 Chr.2: 98823307-98823307 on Build GRCh38
rs534531677 Chr.2: 98823397-98823397 on Build GRCh38
rs763992010 Chr.2: 98823547-98823552 on Build GRCh38
rs767555812 Chr.2: 98823344-98823344 on Build GRCh38
rs912420725 Chr.2: 98823195-98823195 on Build GRCh38
rs747217856 Chr.2: 98823300-98823300 on Build GRCh38
rs766381405 Chr.2: 98823162-98823162 on Build GRCh38
rs762347153 Chr.2: 98823385-98823385 on Build GRCh38
rs779544460 Chr.2: 98823428-98823428 on Build GRCh38
rs372620989 Chr.2: 98823420-98823420 on Build GRCh38
rs775373754 Chr.2: 98823517-98823517 on Build GRCh38
rs777863533 Chr.2: 98823443-98823443 on Build GRCh38
rs772183469 Chr.2: 98823150-98823150 on Build GRCh38
rs752865979 Chr.2: 98823433-98823433 on Build GRCh38
rs752134948 Chr.2: 98823174-98823174 on Build GRCh38
rs997130982 Chr.2: 98823269-98823269 on Build GRCh38
rs557905657 Chr.2: 98823477-98823477 on Build GRCh38
rs905422583 Chr.2: 98823133-98823133 on Build GRCh38
rs759418878 Chr.2: 98823171-98823171 on Build GRCh38
rs776583129 Chr.2: 98823168-98823168 on Build GRCh38
rs1015554225 Chr.2: 98823326-98823326 on Build GRCh38
rs955435112 Chr.2: 98823297-98823297 on Build GRCh38
rs536639314 Chr.2: 98823187-98823187 on Build GRCh38
rs766857080 Chr.2: 98823418-98823418 on Build GRCh38
rs765151815 Chr.2: 98823363-98823363 on Build GRCh38
rs369121202 Chr.2: 98823416-98823416 on Build GRCh38
rs899430218 Chr.2: 98823552-98823552 on Build GRCh38
rs567697961 Chr.2: 98823172-98823172 on Build GRCh38
rs746210600 Chr.2: 98823135-98823135 on Build GRCh38
rs904002251 Chr.2: 98823258-98823258 on Build GRCh38
rs758137296 Chr.2: 98823180-98823180 on Build GRCh38
rs942611100 Chr.2: 98823205-98823205 on Build GRCh38
rs772390480 Chr.2: 98823262-98823262 on Build GRCh38
rs375696418 Chr.2: 98823129-98823129 on Build GRCh38

View Full Product Details