Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127050043-127050539 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN1
Entrez Gene ID
274
Gene Name
bridging integrator 1
Gene Aliases
AMPH2, AMPHL, SH3P9
UniGene
Hs.193163
Gene Chromosome Location
Chr.2: 127048023-127107400 on Build GRCh38
Transcripts
NM_001320632.1 NM_001320633.1 NM_001320634.1 NM_001320640.1 NM_001320641.1 NM_001320642.1 NM_004305.3 NM_139343.2 NM_139344.2 NM_139345.2 NM_139346.2 NM_139347.2 NM_139348.2 NM_139349.2 NM_139350.2 NM_139351.2 XM_005263642.3 XM_005263643.3 XM_005263645.2 XM_006712425.3 XM_006712426.3 XM_006712427.2 XM_006712428.3 XM_006712429.3 XM_006712431.3 XM_006712432.3 XM_006712433.3 XM_006712434.3 XM_011510975.2 XM_017003819.1 XM_017003820.1 XM_017003821.1 XM_017003822.1 XM_017003823.1 XM_017003824.1 XM_017003825.1 XM_017003826.1 XM_017003827.1 XM_017003828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184714 c.1573G>T p.V525L Chr.2: 127050522-127050522 on Build GRCh38
4665717 c.1596G>A p.T532T Chr.2: 127050499-127050499 on Build GRCh38
1184712 c.1609G>T p.D537Y Chr.2: 127050486-127050486 on Build GRCh38
7357512 c.1625A>G p.K542R Chr.2: 127050470-127050470 on Build GRCh38
3364259 c.1334T>G p.L445R Chr.2: 127050473-127050473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144458131 Chr.2: 127050508-127050508 on Build GRCh38
rs937489152 Chr.2: 127050263-127050263 on Build GRCh38
rs554132723 Chr.2: 127050267-127050267 on Build GRCh38
rs537592617 Chr.2: 127050243-127050243 on Build GRCh38
rs940961963 Chr.2: 127050210-127050210 on Build GRCh38
rs766215210 Chr.2: 127050414-127050414 on Build GRCh38
rs899165063 Chr.2: 127050112-127050112 on Build GRCh38
rs751049356 Chr.2: 127050104-127050104 on Build GRCh38
rs72846605 Chr.2: 127050192-127050192 on Build GRCh38
rs970643198 Chr.2: 127050408-127050408 on Build GRCh38
rs920648756 Chr.2: 127050296-127050296 on Build GRCh38
rs890453979 Chr.2: 127050309-127050309 on Build GRCh38
rs762965764 Chr.2: 127050402-127050402 on Build GRCh38
rs757208826 Chr.2: 127050185-127050185 on Build GRCh38
rs777274076 Chr.2: 127050100-127050100 on Build GRCh38
rs757814365 Chr.2: 127050481-127050481 on Build GRCh38
rs993473210 Chr.2: 127050118-127050118 on Build GRCh38
rs746208232 Chr.2: 127050487-127050487 on Build GRCh38
rs1047025998 Chr.2: 127050131-127050131 on Build GRCh38
rs567071677 Chr.2: 127050134-127050134 on Build GRCh38
rs772786604 Chr.2: 127050507-127050507 on Build GRCh38
rs796972322 Chr.2: 127050316-127050316 on Build GRCh38
rs184036842 Chr.2: 127050306-127050306 on Build GRCh38
rs747661696 Chr.2: 127050464-127050464 on Build GRCh38
rs886043878 Chr.2: 127050518-127050518 on Build GRCh38
rs1033325734 Chr.2: 127050303-127050303 on Build GRCh38
rs1008127372 Chr.2: 127050147-127050147 on Build GRCh38
rs964218503 Chr.2: 127050142-127050142 on Build GRCh38
rs547345695 Chr.2: 127050107-127050107 on Build GRCh38
rs768318573 Chr.2: 127050392-127050392 on Build GRCh38
rs769628248 Chr.2: 127050298-127050298 on Build GRCh38
rs747889344 Chr.2: 127050496-127050496 on Build GRCh38
rs996607853 Chr.2: 127050327-127050327 on Build GRCh38
rs904307183 Chr.2: 127050347-127050347 on Build GRCh38
rs770217470 Chr.2: 127050451-127050458 on Build GRCh38
rs764293546 Chr.2: 127050474-127050474 on Build GRCh38
rs767826347 Chr.2: 127050435-127050435 on Build GRCh38
rs774321875 Chr.2: 127050425-127050425 on Build GRCh38
rs1013935534 Chr.2: 127050266-127050266 on Build GRCh38
rs963878171 Chr.2: 127050176-127050176 on Build GRCh38
rs577080553 Chr.2: 127050349-127050349 on Build GRCh38
rs943961294 Chr.2: 127050313-127050313 on Build GRCh38
rs937564049 Chr.2: 127050463-127050463 on Build GRCh38
rs960604641 Chr.2: 127050265-127050265 on Build GRCh38
rs114740593 Chr.2: 127050229-127050229 on Build GRCh38
rs899683814 Chr.2: 127050321-127050321 on Build GRCh38
rs748563618 Chr.2: 127050385-127050386 on Build GRCh38
rs899369350 Chr.2: 127050231-127050231 on Build GRCh38
rs1019900818 Chr.2: 127050150-127050150 on Build GRCh38
rs143258043 Chr.2: 127050466-127050466 on Build GRCh38
rs377599745 Chr.2: 127050394-127050394 on Build GRCh38
rs780738375 Chr.2: 127050407-127050407 on Build GRCh38
rs7568161 Chr.2: 127050365-127050365 on Build GRCh38
rs968369869 Chr.2: 127050214-127050214 on Build GRCh38
rs377047659 Chr.2: 127050093-127050093 on Build GRCh38
rs948874070 Chr.2: 127050270-127050270 on Build GRCh38
rs1031330111 Chr.2: 127050352-127050352 on Build GRCh38
rs773732601 Chr.2: 127050488-127050488 on Build GRCh38
rs776310891 Chr.2: 127050393-127050393 on Build GRCh38
rs138047593 Chr.2: 127050470-127050470 on Build GRCh38
rs777945600 Chr.2: 127050454-127050456 on Build GRCh38
rs374227714 Chr.2: 127050401-127050401 on Build GRCh38
rs763496411 Chr.2: 127050520-127050520 on Build GRCh38
rs887052761 Chr.2: 127050142-127050142 on Build GRCh38
rs916070568 Chr.2: 127050269-127050269 on Build GRCh38
rs12466852 Chr.2: 127050278-127050278 on Build GRCh38
rs759786484 Chr.2: 127050432-127050432 on Build GRCh38
rs558892790 Chr.2: 127050194-127050194 on Build GRCh38
rs112318500 Chr.2: 127050500-127050500 on Build GRCh38
rs879189124 Chr.2: 127050238-127050238 on Build GRCh38
rs558650714 Chr.2: 127050184-127050184 on Build GRCh38
rs543266648 Chr.2: 127050366-127050366 on Build GRCh38
rs142523172 Chr.2: 127050484-127050484 on Build GRCh38
rs967338346 Chr.2: 127050078-127050078 on Build GRCh38
rs1029481097 Chr.2: 127050318-127050318 on Build GRCh38
rs561244491 Chr.2: 127050475-127050475 on Build GRCh38
rs146325986 Chr.2: 127050315-127050315 on Build GRCh38
rs771017384 Chr.2: 127050517-127050517 on Build GRCh38
rs1021402367 Chr.2: 127050328-127050328 on Build GRCh38
rs974848306 Chr.2: 127050143-127050143 on Build GRCh38
rs370637665 Chr.2: 127050399-127050399 on Build GRCh38
rs780816118 Chr.2: 127050494-127050494 on Build GRCh38

View Full Product Details