Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127050443-127050940 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN1
Entrez Gene ID
274
Gene Name
bridging integrator 1
Gene Aliases
AMPH2, AMPHL, SH3P9
UniGene
Hs.193163
Gene Chromosome Location
Chr.2: 127048023-127107400 on Build GRCh38
Transcripts
NM_001320632.1 NM_001320633.1 NM_001320634.1 NM_001320640.1 NM_001320641.1 NM_001320642.1 NM_004305.3 NM_139343.2 NM_139344.2 NM_139345.2 NM_139346.2 NM_139347.2 NM_139348.2 NM_139349.2 NM_139350.2 NM_139351.2 XM_005263642.3 XM_005263643.3 XM_005263645.2 XM_006712425.3 XM_006712426.3 XM_006712427.2 XM_006712428.3 XM_006712429.3 XM_006712431.3 XM_006712432.3 XM_006712433.3 XM_006712434.3 XM_011510975.2 XM_017003819.1 XM_017003820.1 XM_017003821.1 XM_017003822.1 XM_017003823.1 XM_017003824.1 XM_017003825.1 XM_017003826.1 XM_017003827.1 XM_017003828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1184714 c.1573G>T p.V525L Chr.2: 127050522-127050522 on Build GRCh38
4393912 c.1261C>T p.L421L Chr.2: 127050825-127050825 on Build GRCh38
4665717 c.1596G>A p.T532T Chr.2: 127050499-127050499 on Build GRCh38
1184712 c.1609G>T p.D537Y Chr.2: 127050486-127050486 on Build GRCh38
5860051 c.1462-1G>A p.? Chr.2: 127050913-127050913 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144458131 Chr.2: 127050508-127050508 on Build GRCh38
rs759738849 Chr.2: 127050593-127050593 on Build GRCh38
rs371571307 Chr.2: 127050858-127050858 on Build GRCh38
rs759941022 Chr.2: 127050910-127050910 on Build GRCh38
rs768399754 Chr.2: 127050864-127050864 on Build GRCh38
rs746208232 Chr.2: 127050487-127050487 on Build GRCh38
rs761929168 Chr.2: 127050547-127050547 on Build GRCh38
rs772786604 Chr.2: 127050507-127050507 on Build GRCh38
rs747661696 Chr.2: 127050464-127050464 on Build GRCh38
rs553500292 Chr.2: 127050643-127050643 on Build GRCh38
rs760955602 Chr.2: 127050536-127050536 on Build GRCh38
rs367735527 Chr.2: 127050542-127050542 on Build GRCh38
rs756816560 Chr.2: 127050840-127050840 on Build GRCh38
rs770606419 Chr.2: 127050763-127050763 on Build GRCh38
rs561095507 Chr.2: 127050615-127050615 on Build GRCh38
rs764293546 Chr.2: 127050474-127050474 on Build GRCh38
rs202120244 Chr.2: 127050770-127050770 on Build GRCh38
rs556315073 Chr.2: 127050713-127050713 on Build GRCh38
rs867904674 Chr.2: 127050628-127050628 on Build GRCh38
rs766615886 Chr.2: 127050833-127050833 on Build GRCh38
rs780407368 Chr.2: 127050557-127050557 on Build GRCh38
rs573164881 Chr.2: 127050535-127050535 on Build GRCh38
rs777452575 Chr.2: 127050837-127050837 on Build GRCh38
rs772059686 Chr.2: 127050534-127050534 on Build GRCh38
rs377119796 Chr.2: 127050758-127050758 on Build GRCh38
rs755351031 Chr.2: 127050836-127050836 on Build GRCh38
rs1053291620 Chr.2: 127050792-127050792 on Build GRCh38
rs771128647 Chr.2: 127050774-127050789 on Build GRCh38
rs766890661 Chr.2: 127050908-127050908 on Build GRCh38
rs973823531 Chr.2: 127050590-127050590 on Build GRCh38
rs765354438 Chr.2: 127050816-127050816 on Build GRCh38
rs138047593 Chr.2: 127050470-127050470 on Build GRCh38
rs763496411 Chr.2: 127050520-127050520 on Build GRCh38
rs774764729 Chr.2: 127050909-127050909 on Build GRCh38
rs563283906 Chr.2: 127050575-127050576 on Build GRCh38
rs771936924 Chr.2: 127050768-127050768 on Build GRCh38
rs774704128 Chr.2: 127050790-127050791 on Build GRCh38
rs753612525 Chr.2: 127050838-127050838 on Build GRCh38
rs924368753 Chr.2: 127050674-127050674 on Build GRCh38
rs763458588 Chr.2: 127050829-127050829 on Build GRCh38
rs375583449 Chr.2: 127050859-127050859 on Build GRCh38
rs771017384 Chr.2: 127050517-127050517 on Build GRCh38
rs758746626 Chr.2: 127050556-127050556 on Build GRCh38
rs12466912 Chr.2: 127050540-127050540 on Build GRCh38
rs1009023363 Chr.2: 127050552-127050552 on Build GRCh38
rs148422103 Chr.2: 127050834-127050834 on Build GRCh38
rs750869889 Chr.2: 127050826-127050826 on Build GRCh38
rs12466914 Chr.2: 127050551-127050551 on Build GRCh38
rs983554366 Chr.2: 127050697-127050697 on Build GRCh38
rs1024500063 Chr.2: 127050902-127050902 on Build GRCh38
rs757814365 Chr.2: 127050481-127050481 on Build GRCh38
rs995090564 Chr.2: 127050879-127050879 on Build GRCh38
rs7558000 Chr.2: 127050585-127050585 on Build GRCh38
rs773313892 Chr.2: 127050895-127050895 on Build GRCh38
rs767828834 Chr.2: 127050911-127050911 on Build GRCh38
rs78068615 Chr.2: 127050797-127050797 on Build GRCh38
rs375440790 Chr.2: 127050541-127050541 on Build GRCh38
rs533512617 Chr.2: 127050618-127050618 on Build GRCh38
rs745860741 Chr.2: 127050767-127050767 on Build GRCh38
rs886043878 Chr.2: 127050518-127050518 on Build GRCh38
rs768900274 Chr.2: 127050782-127050782 on Build GRCh38
rs779756862 Chr.2: 127050901-127050901 on Build GRCh38
rs188763849 Chr.2: 127050698-127050698 on Build GRCh38
rs747889344 Chr.2: 127050496-127050496 on Build GRCh38
rs368238742 Chr.2: 127050882-127050882 on Build GRCh38
rs745758782 Chr.2: 127050849-127050849 on Build GRCh38
rs2071270 Chr.2: 127050650-127050650 on Build GRCh38
rs748813574 Chr.2: 127050761-127050761 on Build GRCh38
rs755824771 Chr.2: 127050563-127050563 on Build GRCh38
rs759376501 Chr.2: 127050533-127050533 on Build GRCh38
rs937564049 Chr.2: 127050463-127050463 on Build GRCh38
rs966514993 Chr.2: 127050723-127050723 on Build GRCh38
rs776757332 Chr.2: 127050787-127050787 on Build GRCh38
rs761873801 Chr.2: 127050791-127050791 on Build GRCh38
rs778580743 Chr.2: 127050844-127050844 on Build GRCh38
rs147182000 Chr.2: 127050856-127050856 on Build GRCh38
rs143258043 Chr.2: 127050466-127050466 on Build GRCh38
rs777103179 Chr.2: 127050567-127050567 on Build GRCh38
rs774481177 Chr.2: 127050531-127050531 on Build GRCh38
rs760758221 Chr.2: 127050776-127050776 on Build GRCh38
rs200384643 Chr.2: 127050915-127050915 on Build GRCh38
rs773732601 Chr.2: 127050488-127050488 on Build GRCh38
rs752227751 Chr.2: 127050562-127050562 on Build GRCh38
rs144459969 Chr.2: 127050894-127050894 on Build GRCh38
rs1027832381 Chr.2: 127050653-127050653 on Build GRCh38
rs7558001 Chr.2: 127050594-127050594 on Build GRCh38
rs112318500 Chr.2: 127050500-127050500 on Build GRCh38
rs569388445 Chr.2: 127050584-127050584 on Build GRCh38
rs142523172 Chr.2: 127050484-127050484 on Build GRCh38
rs765698397 Chr.2: 127050550-127050550 on Build GRCh38
rs778989524 Chr.2: 127050766-127050766 on Build GRCh38
rs895489070 Chr.2: 127050597-127050597 on Build GRCh38
rs561244491 Chr.2: 127050475-127050475 on Build GRCh38
rs974796988 Chr.2: 127050733-127050733 on Build GRCh38
rs772155930 Chr.2: 127050850-127050850 on Build GRCh38
rs950822160 Chr.2: 127050672-127050672 on Build GRCh38
rs780816118 Chr.2: 127050494-127050494 on Build GRCh38

View Full Product Details