Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127053335-127053842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN1
Entrez Gene ID
274
Gene Name
bridging integrator 1
Gene Aliases
AMPH2, AMPHL, SH3P9
UniGene
Hs.193163
Gene Chromosome Location
Chr.2: 127048023-127107400 on Build GRCh38
Transcripts
NM_001320632.1 NM_001320633.1 NM_001320634.1 NM_001320640.1 NM_001320641.1 NM_001320642.1 NM_004305.3 NM_139343.2 NM_139344.2 NM_139345.2 NM_139346.2 NM_139347.2 NM_139348.2 NM_139349.2 NM_139350.2 NM_139351.2 XM_005263642.3 XM_005263643.3 XM_005263645.2 XM_006712425.3 XM_006712426.3 XM_006712427.2 XM_006712428.3 XM_006712429.3 XM_006712431.3 XM_006712432.3 XM_006712433.3 XM_006712434.3 XM_011510975.2 XM_017003819.1 XM_017003820.1 XM_017003821.1 XM_017003822.1 XM_017003823.1 XM_017003824.1 XM_017003825.1 XM_017003826.1 XM_017003827.1 XM_017003828.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398157 c.1241C>G p.S414* Chr.2: 127053444-127053444 on Build GRCh38
5527361 c.1260G>A p.W420* Chr.2: 127053425-127053425 on Build GRCh38
3566063 c.1252G>A p.D418N Chr.2: 127053433-127053433 on Build GRCh38
7260126 c.1240-6G>A p.? Chr.2: 127053451-127053451 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs572824852 Chr.2: 127053734-127053734 on Build GRCh38
rs767455338 Chr.2: 127053487-127053487 on Build GRCh38
rs774661155 Chr.2: 127053475-127053475 on Build GRCh38
rs761602967 Chr.2: 127053498-127053498 on Build GRCh38
rs759536531 Chr.2: 127053558-127053558 on Build GRCh38
rs559070746 Chr.2: 127053653-127053653 on Build GRCh38
rs775174601 Chr.2: 127053371-127053371 on Build GRCh38
rs373614051 Chr.2: 127053618-127053618 on Build GRCh38
rs767984867 Chr.2: 127053462-127053462 on Build GRCh38
rs1022605491 Chr.2: 127053692-127053692 on Build GRCh38
rs1038024858 Chr.2: 127053796-127053796 on Build GRCh38
rs752845587 Chr.2: 127053492-127053492 on Build GRCh38
rs756676565 Chr.2: 127053440-127053440 on Build GRCh38
rs929140162 Chr.2: 127053725-127053725 on Build GRCh38
rs960451815 Chr.2: 127053737-127053737 on Build GRCh38
rs746676469 Chr.2: 127053452-127053452 on Build GRCh38
rs910187551 Chr.2: 127053656-127053656 on Build GRCh38
rs111501179 Chr.2: 127053473-127053473 on Build GRCh38
rs1025707510 Chr.2: 127053545-127053545 on Build GRCh38
rs978353784 Chr.2: 127053710-127053710 on Build GRCh38
rs751900148 Chr.2: 127053410-127053410 on Build GRCh38
rs889056204 Chr.2: 127053504-127053504 on Build GRCh38
rs1021096614 Chr.2: 127053808-127053808 on Build GRCh38
rs1043384869 Chr.2: 127053501-127053501 on Build GRCh38
rs773160345 Chr.2: 127053389-127053389 on Build GRCh38
rs758683621 Chr.2: 127053747-127053747 on Build GRCh38
rs759813870 Chr.2: 127053483-127053483 on Build GRCh38
rs190020084 Chr.2: 127053788-127053788 on Build GRCh38
rs916267343 Chr.2: 127053758-127053758 on Build GRCh38
rs967768131 Chr.2: 127053619-127053619 on Build GRCh38
rs761815010 Chr.2: 127053383-127053383 on Build GRCh38
rs113666720 Chr.2: 127053790-127053790 on Build GRCh38
rs548956444 Chr.2: 127053451-127053451 on Build GRCh38
rs370492816 Chr.2: 127053534-127053534 on Build GRCh38
rs1005553069 Chr.2: 127053745-127053745 on Build GRCh38
rs987906095 Chr.2: 127053459-127053459 on Build GRCh38
rs569766405 Chr.2: 127053405-127053405 on Build GRCh38
rs939064907 Chr.2: 127053540-127053540 on Build GRCh38
rs373567000 Chr.2: 127053450-127053450 on Build GRCh38
rs777951748 Chr.2: 127053445-127053445 on Build GRCh38
rs753764530 Chr.2: 127053548-127053548 on Build GRCh38
rs538904399 Chr.2: 127053583-127053583 on Build GRCh38
rs768268793 Chr.2: 127053373-127053373 on Build GRCh38
rs760718492 Chr.2: 127053495-127053495 on Build GRCh38
rs894779103 Chr.2: 127053565-127053565 on Build GRCh38
rs747982957 Chr.2: 127053464-127053464 on Build GRCh38
rs776373718 Chr.2: 127053375-127053375 on Build GRCh38
rs1029810359 Chr.2: 127053615-127053615 on Build GRCh38
rs767652810 Chr.2: 127053415-127053415 on Build GRCh38
rs966640943 Chr.2: 127053697-127053697 on Build GRCh38
rs994771118 Chr.2: 127053608-127053608 on Build GRCh38
rs901667377 Chr.2: 127053541-127053541 on Build GRCh38
rs989433439 Chr.2: 127053684-127053684 on Build GRCh38
rs766353114 Chr.2: 127053372-127053372 on Build GRCh38
rs145705260 Chr.2: 127053616-127053616 on Build GRCh38
rs377476606 Chr.2: 127053465-127053465 on Build GRCh38
rs1039002660 Chr.2: 127053577-127053577 on Build GRCh38
rs923119281 Chr.2: 127053707-127053709 on Build GRCh38
rs762840542 Chr.2: 127053472-127053472 on Build GRCh38
rs774660666 Chr.2: 127053739-127053739 on Build GRCh38
rs964634754 Chr.2: 127053463-127053463 on Build GRCh38
rs941045274 Chr.2: 127053574-127053574 on Build GRCh38
rs114383514 Chr.2: 127053569-127053569 on Build GRCh38
rs961218507 Chr.2: 127053798-127053798 on Build GRCh38
rs1006923701 Chr.2: 127053621-127053621 on Build GRCh38
rs753079936 Chr.2: 127053439-127053439 on Build GRCh38
rs998713902 Chr.2: 127053544-127053544 on Build GRCh38
rs764898585 Chr.2: 127053607-127053607 on Build GRCh38
rs905199854 Chr.2: 127053781-127053781 on Build GRCh38
rs138274544 Chr.2: 127053805-127053805 on Build GRCh38
rs887210139 Chr.2: 127053739-127053739 on Build GRCh38
rs764896643 Chr.2: 127053384-127053384 on Build GRCh38
rs955442228 Chr.2: 127053442-127053442 on Build GRCh38
rs76414720 Chr.2: 127053466-127053466 on Build GRCh38
rs766271775 Chr.2: 127053535-127053535 on Build GRCh38
rs78967885 Chr.2: 127053411-127053411 on Build GRCh38
rs1018241014 Chr.2: 127053649-127053649 on Build GRCh38
rs542788152 Chr.2: 127053632-127053632 on Build GRCh38
rs927780831 Chr.2: 127053731-127053731 on Build GRCh38
rs558399490 Chr.2: 127053500-127053500 on Build GRCh38
rs1054829179 Chr.2: 127053552-127053552 on Build GRCh38
rs957330061 Chr.2: 127053669-127053669 on Build GRCh38
rs376482460 Chr.2: 127053391-127053391 on Build GRCh38
rs979097261 Chr.2: 127053648-127053648 on Build GRCh38

View Full Product Details