Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130139250-130139751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC74B
Entrez Gene ID
91409
Gene Name
coiled-coil domain containing 74B
Gene Aliases
-
UniGene
Hs.29383
Gene Chromosome Location
Chr.2: 130139283-130145134 on Build GRCh38
Transcripts
NM_001258307.1 NM_207310.2 XM_005263840.2 XM_005263842.2 XM_006712833.2 XM_006712834.3 XM_006712835.2 XM_006712837.2 XM_011512142.2 NR_033903.1

Gene Symbol
MED15P9
Entrez Gene ID
285103
Gene Name
mediator complex subunit 15 pseudogene 9
Gene Aliases
CCDC74B-AS1, MED15P2
UniGene
Hs.570106
Gene Chromosome Location
Chr.2: 130129623-130139413 on Build GRCh38
Transcripts
NM_001258307.1 NM_207310.2 XM_005263840.2 XM_005263842.2 XM_006712833.2 XM_006712834.3 XM_006712835.2 XM_006712837.2 XM_011512142.2 NR_033903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1247548 c.1066C>A p.P356T Chr.2: 130139632-130139632 on Build GRCh38
7360001 c.1130G>C p.R377P Chr.2: 130139568-130139568 on Build GRCh38
3757621 c.1359A>G p.A453A Chr.2: 130139645-130139645 on Build GRCh38
4582649 c.1325G>A p.S442N Chr.2: 130139679-130139679 on Build GRCh38
4998055 c.1037G>A p.R346H Chr.2: 130139661-130139661 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779450387 Chr.2: 130139669-130139669 on Build GRCh38
rs28751929 Chr.2: 130139482-130139482 on Build GRCh38
rs199976091 Chr.2: 130139635-130139635 on Build GRCh38
rs375087118 Chr.2: 130139476-130139476 on Build GRCh38
rs540783590 Chr.2: 130139463-130139463 on Build GRCh38
rs12622823 Chr.2: 130139465-130139465 on Build GRCh38
rs745606246 Chr.2: 130139525-130139525 on Build GRCh38
rs1025875323 Chr.2: 130139345-130139345 on Build GRCh38
rs763710118 Chr.2: 130139577-130139577 on Build GRCh38
rs986949965 Chr.2: 130139405-130139405 on Build GRCh38
rs760407589 Chr.2: 130139632-130139632 on Build GRCh38
rs955738511 Chr.2: 130139348-130139348 on Build GRCh38
rs146190210 Chr.2: 130139568-130139568 on Build GRCh38
rs181554754 Chr.2: 130139490-130139490 on Build GRCh38
rs148241925 Chr.2: 130139613-130139613 on Build GRCh38
rs530528147 Chr.2: 130139699-130139699 on Build GRCh38
rs144853884 Chr.2: 130139718-130139718 on Build GRCh38
rs769205055 Chr.2: 130139698-130139698 on Build GRCh38
rs757202496 Chr.2: 130139513-130139513 on Build GRCh38
rs13006246 Chr.2: 130139645-130139645 on Build GRCh38
rs766982044 Chr.2: 130139539-130139540 on Build GRCh38
rs749770499 Chr.2: 130139666-130139666 on Build GRCh38
rs764770813 Chr.2: 130139638-130139638 on Build GRCh38
rs573590058 Chr.2: 130139344-130139344 on Build GRCh38
rs766243365 Chr.2: 130139603-130139603 on Build GRCh38
rs145277627 Chr.2: 130139631-130139631 on Build GRCh38
rs1001975360 Chr.2: 130139280-130139280 on Build GRCh38
rs397833887 Chr.2: 130139445-130139445 on Build GRCh38
rs368544670 Chr.2: 130139695-130139695 on Build GRCh38
rs767871488 Chr.2: 130139724-130139724 on Build GRCh38
rs774894233 Chr.2: 130139613-130139613 on Build GRCh38
rs753882095 Chr.2: 130139509-130139509 on Build GRCh38
rs563201571 Chr.2: 130139697-130139697 on Build GRCh38
rs758665665 Chr.2: 130139534-130139534 on Build GRCh38
rs772629812 Chr.2: 130139475-130139475 on Build GRCh38
rs1047052375 Chr.2: 130139688-130139688 on Build GRCh38
rs544811183 Chr.2: 130139667-130139667 on Build GRCh38
rs368361030 Chr.2: 130139596-130139596 on Build GRCh38
rs528357118 Chr.2: 130139730-130139730 on Build GRCh38
rs937391309 Chr.2: 130139507-130139507 on Build GRCh38
rs2599951 Chr.2: 130139728-130139728 on Build GRCh38
rs867346139 Chr.2: 130139312-130139312 on Build GRCh38
rs776460116 Chr.2: 130139679-130139679 on Build GRCh38
rs146986944 Chr.2: 130139665-130139665 on Build GRCh38
rs558456261 Chr.2: 130139321-130139321 on Build GRCh38
rs528371250 Chr.2: 130139580-130139580 on Build GRCh38
rs199737933 Chr.2: 130139655-130139655 on Build GRCh38
rs758163811 Chr.2: 130139590-130139590 on Build GRCh38
rs370710410 Chr.2: 130139706-130139706 on Build GRCh38
rs568199837 Chr.2: 130139599-130139599 on Build GRCh38
rs2599952 Chr.2: 130139731-130139731 on Build GRCh38
rs374773706 Chr.2: 130139549-130139549 on Build GRCh38
rs1018482693 Chr.2: 130139442-130139442 on Build GRCh38
rs751795227 Chr.2: 130139611-130139611 on Build GRCh38
rs752887091 Chr.2: 130139662-130139662 on Build GRCh38
rs540610545 Chr.2: 130139657-130139657 on Build GRCh38
rs778772323 Chr.2: 130139518-130139518 on Build GRCh38
rs144580593 Chr.2: 130139618-130139618 on Build GRCh38
rs569391388 Chr.2: 130139630-130139630 on Build GRCh38
rs140718616 Chr.2: 130139647-130139647 on Build GRCh38
rs760388920 Chr.2: 130139576-130139576 on Build GRCh38
rs188560003 Chr.2: 130139578-130139578 on Build GRCh38
rs917802308 Chr.2: 130139487-130139487 on Build GRCh38
rs755215770 Chr.2: 130139658-130139658 on Build GRCh38
rs138499909 Chr.2: 130139663-130139663 on Build GRCh38
rs145251569 Chr.2: 130139565-130139565 on Build GRCh38
rs748662969 Chr.2: 130139564-130139564 on Build GRCh38
rs184239466 Chr.2: 130139501-130139501 on Build GRCh38
rs750350697 Chr.2: 130139582-130139582 on Build GRCh38
rs372587202 Chr.2: 130139634-130139634 on Build GRCh38
rs775071350 Chr.2: 130139569-130139569 on Build GRCh38
rs547188613 Chr.2: 130139717-130139717 on Build GRCh38
rs3177472 Chr.2: 130139661-130139661 on Build GRCh38
rs370509041 Chr.2: 130139548-130139548 on Build GRCh38
rs759937180 Chr.2: 130139693-130139693 on Build GRCh38
rs898774125 Chr.2: 130139337-130139337 on Build GRCh38
rs143728442 Chr.2: 130139678-130139678 on Build GRCh38
rs768382538 Chr.2: 130139633-130139633 on Build GRCh38
rs200926787 Chr.2: 130139547-130139547 on Build GRCh38
rs781284784 Chr.2: 130139614-130139614 on Build GRCh38
rs766259377 Chr.2: 130139700-130139700 on Build GRCh38
rs139278006 Chr.2: 130139629-130139629 on Build GRCh38
rs546767071 Chr.2: 130139581-130139581 on Build GRCh38
rs1046687 Chr.2: 130139506-130139506 on Build GRCh38
rs1044898750 Chr.2: 130139502-130139502 on Build GRCh38

View Full Product Details