Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130139713-130140208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC74B
Entrez Gene ID
91409
Gene Name
coiled-coil domain containing 74B
Gene Aliases
-
UniGene
Hs.29383
Gene Chromosome Location
Chr.2: 130139283-130145134 on Build GRCh38
Transcripts
NM_001258307.1 NM_207310.2 XM_005263840.2 XM_005263842.2 XM_006712833.2 XM_006712834.3 XM_006712835.2 XM_006712837.2 XM_011512142.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3797957 c.965C>T p.T322M Chr.2: 130139933-130139933 on Build GRCh38
5479966 c.881A>T p.Q294L Chr.2: 130140092-130140092 on Build GRCh38
3734102 c.1232T>C p.V411A Chr.2: 130140047-130140047 on Build GRCh38
382703 c.1007+1G>T p.? Chr.2: 130139890-130139890 on Build GRCh38
5528277 c.979G>A p.V327I Chr.2: 130139919-130139919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370468132 Chr.2: 130140068-130140068 on Build GRCh38
rs768299950 Chr.2: 130139971-130139971 on Build GRCh38
rs1002238795 Chr.2: 130139985-130139985 on Build GRCh38
rs757988551 Chr.2: 130140128-130140128 on Build GRCh38
rs753981925 Chr.2: 130139900-130139900 on Build GRCh38
rs775286324 Chr.2: 130139864-130139864 on Build GRCh38
rs368450970 Chr.2: 130139744-130139744 on Build GRCh38
rs750046724 Chr.2: 130140127-130140127 on Build GRCh38
rs762110491 Chr.2: 130139977-130139977 on Build GRCh38
rs2599953 Chr.2: 130139817-130139817 on Build GRCh38
rs145421181 Chr.2: 130140182-130140182 on Build GRCh38
rs776975404 Chr.2: 130139972-130139972 on Build GRCh38
rs757449095 Chr.2: 130139905-130139905 on Build GRCh38
rs2599954 Chr.2: 130140092-130140092 on Build GRCh38
rs549366225 Chr.2: 130140080-130140080 on Build GRCh38
rs74727782 Chr.2: 130139800-130139800 on Build GRCh38
rs555755677 Chr.2: 130139801-130139801 on Build GRCh38
rs369545972 Chr.2: 130139885-130139885 on Build GRCh38
rs759903276 Chr.2: 130140095-130140095 on Build GRCh38
rs762159818 Chr.2: 130140069-130140069 on Build GRCh38
rs771955923 Chr.2: 130139950-130139950 on Build GRCh38
rs139725529 Chr.2: 130140087-130140087 on Build GRCh38
rs766963833 Chr.2: 130140004-130140004 on Build GRCh38
rs368129654 Chr.2: 130140120-130140120 on Build GRCh38
rs10184921 Chr.2: 130140123-130140123 on Build GRCh38
rs566991375 Chr.2: 130140063-130140063 on Build GRCh38
rs143752511 Chr.2: 130140044-130140044 on Build GRCh38
rs551931772 Chr.2: 130140050-130140050 on Build GRCh38
rs755279496 Chr.2: 130139932-130139932 on Build GRCh38
rs377326897 Chr.2: 130140006-130140006 on Build GRCh38
rs546608145 Chr.2: 130139740-130139740 on Build GRCh38
rs749509237 Chr.2: 130140172-130140172 on Build GRCh38
rs781065179 Chr.2: 130140157-130140157 on Build GRCh38
rs374511863 Chr.2: 130139965-130139965 on Build GRCh38
rs763580990 Chr.2: 130139990-130139990 on Build GRCh38
rs752800102 Chr.2: 130139734-130139735 on Build GRCh38
rs773549349 Chr.2: 130140082-130140082 on Build GRCh38
rs529047326 Chr.2: 130139743-130139743 on Build GRCh38
rs970776830 Chr.2: 130139765-130139765 on Build GRCh38
rs760838172 Chr.2: 130139891-130139891 on Build GRCh38
rs755438041 Chr.2: 130140022-130140022 on Build GRCh38
rs114106389 Chr.2: 130139793-130139793 on Build GRCh38
rs373709374 Chr.2: 130139892-130139892 on Build GRCh38
rs561795482 Chr.2: 130139929-130139929 on Build GRCh38
rs150631926 Chr.2: 130140043-130140043 on Build GRCh38
rs758911313 Chr.2: 130139917-130139917 on Build GRCh38
rs768408277 Chr.2: 130140149-130140149 on Build GRCh38
rs375465316 Chr.2: 130139946-130139946 on Build GRCh38
rs769963621 Chr.2: 130140009-130140009 on Build GRCh38
rs182090597 Chr.2: 130140065-130140065 on Build GRCh38
rs556340360 Chr.2: 130139816-130139816 on Build GRCh38
rs532862631 Chr.2: 130139755-130139755 on Build GRCh38
rs764328099 Chr.2: 130139893-130139893 on Build GRCh38
rs956447813 Chr.2: 130139748-130139748 on Build GRCh38
rs2599952 Chr.2: 130139731-130139731 on Build GRCh38
rs376263599 Chr.2: 130139845-130139845 on Build GRCh38
rs780599881 Chr.2: 130139924-130139924 on Build GRCh38
rs767931161 Chr.2: 130140117-130140117 on Build GRCh38
rs2258632 Chr.2: 130139986-130139986 on Build GRCh38
rs2259332 Chr.2: 130140047-130140047 on Build GRCh38
rs375043900 Chr.2: 130140176-130140176 on Build GRCh38
rs540240396 Chr.2: 130139909-130139909 on Build GRCh38
rs545531426 Chr.2: 130139844-130139844 on Build GRCh38
rs936906562 Chr.2: 130140140-130140140 on Build GRCh38
rs781398863 Chr.2: 130140030-130140030 on Build GRCh38
rs185141509 Chr.2: 130139809-130139809 on Build GRCh38
rs771057699 Chr.2: 130140173-130140173 on Build GRCh38
rs765400933 Chr.2: 130139906-130139906 on Build GRCh38
rs921546731 Chr.2: 130140130-130140130 on Build GRCh38
rs900090192 Chr.2: 130139874-130139874 on Build GRCh38
rs372856673 Chr.2: 130139737-130139737 on Build GRCh38
rs539985577 Chr.2: 130139771-130139771 on Build GRCh38
rs767312334 Chr.2: 130139855-130139855 on Build GRCh38
rs572724786 Chr.2: 130139850-130139850 on Build GRCh38
rs973264570 Chr.2: 130140023-130140023 on Build GRCh38
rs368943554 Chr.2: 130139732-130139732 on Build GRCh38
rs770488358 Chr.2: 130139940-130139940 on Build GRCh38
rs574988095 Chr.2: 130139829-130139829 on Build GRCh38
rs748008068 Chr.2: 130140166-130140166 on Build GRCh38
rs963212504 Chr.2: 130139794-130139794 on Build GRCh38
rs180885011 Chr.2: 130139753-130139753 on Build GRCh38
rs769684730 Chr.2: 130140168-130140168 on Build GRCh38
rs750493358 Chr.2: 130139987-130139987 on Build GRCh38
rs200403503 Chr.2: 130140134-130140134 on Build GRCh38
rs372108145 Chr.2: 130139933-130139933 on Build GRCh38
rs773794192 Chr.2: 130139942-130139942 on Build GRCh38
rs2599955 Chr.2: 130140118-130140118 on Build GRCh38
rs574290373 Chr.2: 130139808-130139808 on Build GRCh38
rs780076323 Chr.2: 130140062-130140062 on Build GRCh38
rs773635371 Chr.2: 130140171-130140171 on Build GRCh38
rs747763506 Chr.2: 130139738-130139738 on Build GRCh38
rs568367830 Chr.2: 130139741-130139741 on Build GRCh38
rs551790252 Chr.2: 130139786-130139786 on Build GRCh38
rs746976764 Chr.2: 130140135-130140135 on Build GRCh38
rs371159646 Chr.2: 130139936-130139936 on Build GRCh38

View Full Product Details