Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 130141833-130142327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC74B
Entrez Gene ID
91409
Gene Name
coiled-coil domain containing 74B
Gene Aliases
-
UniGene
Hs.29383
Gene Chromosome Location
Chr.2: 130139283-130145134 on Build GRCh38
Transcripts
NM_001258307.1 NM_207310.2 XM_005263840.2 XM_005263842.2 XM_006712833.2 XM_006712834.3 XM_006712835.2 XM_006712837.2 XM_011512142.2 XM_011512147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4999715 c.482G>A p.R161H Chr.2: 130142195-130142195 on Build GRCh38
1006459 c.472G>A p.D158N Chr.2: 130142205-130142205 on Build GRCh38
3766514 c.446A>G p.H149R Chr.2: 130142231-130142231 on Build GRCh38
4670015 c.805C>A p.L269I Chr.2: 130142178-130142178 on Build GRCh38
3766513 c.765G>A p.A255A Chr.2: 130142218-130142218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003539368 Chr.2: 130142052-130142052 on Build GRCh38
rs1035721645 Chr.2: 130142054-130142054 on Build GRCh38
rs376592368 Chr.2: 130142087-130142087 on Build GRCh38
rs768537618 Chr.2: 130142209-130142209 on Build GRCh38
rs748786994 Chr.2: 130142197-130142198 on Build GRCh38
rs754876378 Chr.2: 130142268-130142268 on Build GRCh38
rs2599969 Chr.2: 130142045-130142045 on Build GRCh38
rs771593923 Chr.2: 130142286-130142286 on Build GRCh38
rs777058872 Chr.2: 130142214-130142214 on Build GRCh38
rs745776114 Chr.2: 130142170-130142170 on Build GRCh38
rs115740913 Chr.2: 130142041-130142041 on Build GRCh38
rs762022559 Chr.2: 130142188-130142188 on Build GRCh38
rs773783471 Chr.2: 130142193-130142193 on Build GRCh38
rs1054198632 Chr.2: 130141934-130141934 on Build GRCh38
rs147577700 Chr.2: 130142195-130142195 on Build GRCh38
rs536402804 Chr.2: 130141852-130141852 on Build GRCh38
rs369980509 Chr.2: 130142276-130142276 on Build GRCh38
rs765402726 Chr.2: 130142190-130142190 on Build GRCh38
rs777291005 Chr.2: 130142150-130142150 on Build GRCh38
rs188925101 Chr.2: 130142069-130142069 on Build GRCh38
rs3833550 Chr.2: 130141957-130141958 on Build GRCh38
rs781068310 Chr.2: 130142269-130142269 on Build GRCh38
rs374183865 Chr.2: 130142115-130142115 on Build GRCh38
rs752399930 Chr.2: 130142135-130142135 on Build GRCh38
rs147529259 Chr.2: 130142297-130142297 on Build GRCh38
rs2599972 Chr.2: 130142231-130142231 on Build GRCh38
rs774253510 Chr.2: 130142091-130142091 on Build GRCh38
rs760985170 Chr.2: 130142180-130142180 on Build GRCh38
rs764270343 Chr.2: 130142114-130142114 on Build GRCh38
rs1021001142 Chr.2: 130142022-130142022 on Build GRCh38
rs184349150 Chr.2: 130142171-130142171 on Build GRCh38
rs563910835 Chr.2: 130142119-130142119 on Build GRCh38
rs138145722 Chr.2: 130142208-130142208 on Build GRCh38
rs549861352 Chr.2: 130141970-130141970 on Build GRCh38
rs773126193 Chr.2: 130142216-130142216 on Build GRCh38
rs769486824 Chr.2: 130142099-130142100 on Build GRCh38
rs959594856 Chr.2: 130142058-130142058 on Build GRCh38
rs749850911 Chr.2: 130142246-130142246 on Build GRCh38
rs138883979 Chr.2: 130142225-130142225 on Build GRCh38
rs766798286 Chr.2: 130142197-130142197 on Build GRCh38
rs763780838 Chr.2: 130142202-130142202 on Build GRCh38
rs767858668 Chr.2: 130142226-130142226 on Build GRCh38
rs758857890 Chr.2: 130142120-130142120 on Build GRCh38
rs372365368 Chr.2: 130142107-130142107 on Build GRCh38
rs376959994 Chr.2: 130142219-130142219 on Build GRCh38
rs57389073 Chr.2: 130141951-130141952 on Build GRCh38
rs369555594 Chr.2: 130142220-130142220 on Build GRCh38
rs777209633 Chr.2: 130142152-130142155 on Build GRCh38
rs748414618 Chr.2: 130142271-130142271 on Build GRCh38
rs148885074 Chr.2: 130142206-130142206 on Build GRCh38
rs770093188 Chr.2: 130142274-130142274 on Build GRCh38
rs749371703 Chr.2: 130142288-130142288 on Build GRCh38
rs748850887 Chr.2: 130142152-130142152 on Build GRCh38
rs764829669 Chr.2: 130142238-130142238 on Build GRCh38
rs775836110 Chr.2: 130142103-130142103 on Build GRCh38
rs377208822 Chr.2: 130142256-130142256 on Build GRCh38
rs576799767 Chr.2: 130142084-130142084 on Build GRCh38
rs900408238 Chr.2: 130141858-130141858 on Build GRCh38
rs893918797 Chr.2: 130141956-130141957 on Build GRCh38
rs372517815 Chr.2: 130142255-130142255 on Build GRCh38
rs771756391 Chr.2: 130142101-130142101 on Build GRCh38
rs753158880 Chr.2: 130142035-130142035 on Build GRCh38
rs776970886 Chr.2: 130142185-130142185 on Build GRCh38
rs2599968 Chr.2: 130141892-130141892 on Build GRCh38
rs367750467 Chr.2: 130142116-130142116 on Build GRCh38
rs141892357 Chr.2: 130142218-130142218 on Build GRCh38
rs2599970 Chr.2: 130142046-130142046 on Build GRCh38
rs756833897 Chr.2: 130142235-130142235 on Build GRCh38
rs533204838 Chr.2: 130141951-130141951 on Build GRCh38
rs149705123 Chr.2: 130142196-130142196 on Build GRCh38
rs554516844 Chr.2: 130141860-130141860 on Build GRCh38
rs771043266 Chr.2: 130142290-130142290 on Build GRCh38
rs371953371 Chr.2: 130142205-130142205 on Build GRCh38
rs761473067 Chr.2: 130142307-130142307 on Build GRCh38
rs983608342 Chr.2: 130142137-130142137 on Build GRCh38
rs549928835 Chr.2: 130142200-130142200 on Build GRCh38
rs770400267 Chr.2: 130142159-130142159 on Build GRCh38
rs780226584 Chr.2: 130142130-130142130 on Build GRCh38
rs759418083 Chr.2: 130142100-130142100 on Build GRCh38
rs748324768 Chr.2: 130142215-130142215 on Build GRCh38
rs770811239 Chr.2: 130142089-130142089 on Build GRCh38
rs565963814 Chr.2: 130142296-130142296 on Build GRCh38
rs755682285 Chr.2: 130142138-130142138 on Build GRCh38
rs775788405 Chr.2: 130142300-130142300 on Build GRCh38
rs765892490 Chr.2: 130142118-130142118 on Build GRCh38
rs931854542 Chr.2: 130141896-130141896 on Build GRCh38
rs778891774 Chr.2: 130142166-130142166 on Build GRCh38
rs768950206 Chr.2: 130142183-130142183 on Build GRCh38
rs921874970 Chr.2: 130142232-130142232 on Build GRCh38
rs192240110 Chr.2: 130142088-130142088 on Build GRCh38
rs1040697006 Chr.2: 130141853-130141853 on Build GRCh38
rs766602052 Chr.2: 130141950-130141951 on Build GRCh38

View Full Product Details