Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131048976-131049485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168B
Entrez Gene ID
130074
Gene Name
family with sequence similarity 168 member B
Gene Aliases
MANI
UniGene
Hs.534679
Gene Chromosome Location
Chr.2: 131047876-131093468 on Build GRCh38
Transcripts
NM_001009993.3 NM_001321743.1 NM_001321744.1 NM_001321745.1 NM_001321746.1 NM_001321747.1 NM_001321748.1 XM_011510594.2 XM_017003327.1 XM_017003328.1

SNP Detail

SNP ID Chromosome Location
rs1026426006 Chr.2: 131049410-131049410 on Build GRCh38
rs893373520 Chr.2: 131049187-131049187 on Build GRCh38
rs866482255 Chr.2: 131049451-131049451 on Build GRCh38
rs575588296 Chr.2: 131049183-131049183 on Build GRCh38
rs893362178 Chr.2: 131049196-131049196 on Build GRCh38
rs1016034336 Chr.2: 131049421-131049421 on Build GRCh38
rs757377303 Chr.2: 131049210-131049210 on Build GRCh38
rs984624871 Chr.2: 131049274-131049274 on Build GRCh38
rs934769400 Chr.2: 131049348-131049348 on Build GRCh38
rs1000499864 Chr.2: 131049186-131049186 on Build GRCh38
rs535234274 Chr.2: 131049103-131049103 on Build GRCh38
rs981510468 Chr.2: 131049055-131049055 on Build GRCh38
rs950564575 Chr.2: 131049273-131049273 on Build GRCh38
rs115350279 Chr.2: 131049332-131049332 on Build GRCh38
rs370598814 Chr.2: 131049081-131049081 on Build GRCh38
rs1034426461 Chr.2: 131049271-131049271 on Build GRCh38
rs184101230 Chr.2: 131049259-131049259 on Build GRCh38
rs1020492933 Chr.2: 131049246-131049246 on Build GRCh38
rs1013091149 Chr.2: 131049357-131049357 on Build GRCh38
rs548355143 Chr.2: 131049147-131049147 on Build GRCh38
rs909069620 Chr.2: 131049279-131049279 on Build GRCh38
rs575231803 Chr.2: 131049137-131049137 on Build GRCh38
rs765582770 Chr.2: 131049120-131049120 on Build GRCh38
rs754850599 Chr.2: 131049364-131049364 on Build GRCh38
rs985065995 Chr.2: 131049033-131049033 on Build GRCh38
rs138951348 Chr.2: 131049004-131049004 on Build GRCh38
rs964476718 Chr.2: 131049428-131049428 on Build GRCh38
rs570773520 Chr.2: 131049390-131049390 on Build GRCh38
rs1013145691 Chr.2: 131049190-131049190 on Build GRCh38
rs971441957 Chr.2: 131049010-131049010 on Build GRCh38
rs571055110 Chr.2: 131049057-131049057 on Build GRCh38
rs527662064 Chr.2: 131049205-131049205 on Build GRCh38
rs767578496 Chr.2: 131049282-131049282 on Build GRCh38
rs912436594 Chr.2: 131049361-131049363 on Build GRCh38
rs747932643 Chr.2: 131049436-131049436 on Build GRCh38
rs1051085270 Chr.2: 131049327-131049327 on Build GRCh38
rs74465372 Chr.2: 131049240-131049240 on Build GRCh38
rs890946604 Chr.2: 131049149-131049149 on Build GRCh38
rs909131605 Chr.2: 131049063-131049063 on Build GRCh38
rs974608773 Chr.2: 131049293-131049293 on Build GRCh38
rs760119356 Chr.2: 131049091-131049091 on Build GRCh38
rs929817540 Chr.2: 131049139-131049139 on Build GRCh38
rs191979065 Chr.2: 131049179-131049179 on Build GRCh38
rs971011084 Chr.2: 131049215-131049215 on Build GRCh38
rs547746200 Chr.2: 131049353-131049353 on Build GRCh38
rs899545098 Chr.2: 131049184-131049184 on Build GRCh38
rs775868785 Chr.2: 131049122-131049122 on Build GRCh38
rs907485605 Chr.2: 131049382-131049382 on Build GRCh38
rs559197255 Chr.2: 131049415-131049415 on Build GRCh38
rs535799771 Chr.2: 131049170-131049170 on Build GRCh38
rs375086797 Chr.2: 131049416-131049416 on Build GRCh38
rs933959741 Chr.2: 131049313-131049313 on Build GRCh38
rs747822176 Chr.2: 131049140-131049140 on Build GRCh38
rs923193696 Chr.2: 131049310-131049310 on Build GRCh38
rs753831164 Chr.2: 131049355-131049355 on Build GRCh38
rs1039667560 Chr.2: 131049099-131049099 on Build GRCh38
rs923183817 Chr.2: 131049442-131049442 on Build GRCh38
rs899817126 Chr.2: 131049136-131049136 on Build GRCh38
rs141239119 Chr.2: 131049108-131049108 on Build GRCh38
rs929118690 Chr.2: 131049026-131049026 on Build GRCh38

View Full Product Details