Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131050106-131050617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168B
Entrez Gene ID
130074
Gene Name
family with sequence similarity 168 member B
Gene Aliases
MANI
UniGene
Hs.534679
Gene Chromosome Location
Chr.2: 131047876-131093468 on Build GRCh38
Transcripts
NM_001009993.3 NM_001321743.1 NM_001321744.1 NM_001321745.1 NM_001321746.1 NM_001321747.1 NM_001321748.1 XM_011510594.2 XM_017003327.1 XM_017003328.1

SNP Detail

SNP ID Chromosome Location
rs996760015 Chr.2: 131050153-131050153 on Build GRCh38
rs563439689 Chr.2: 131050386-131050386 on Build GRCh38
rs754902297 Chr.2: 131050573-131050573 on Build GRCh38
rs970060659 Chr.2: 131050252-131050252 on Build GRCh38
rs927653174 Chr.2: 131050387-131050387 on Build GRCh38
rs1051424689 Chr.2: 131050366-131050366 on Build GRCh38
rs112954822 Chr.2: 131050145-131050145 on Build GRCh38
rs553506808 Chr.2: 131050550-131050551 on Build GRCh38
rs574334301 Chr.2: 131050169-131050169 on Build GRCh38
rs555432024 Chr.2: 131050130-131050131 on Build GRCh38
rs908006275 Chr.2: 131050538-131050538 on Build GRCh38
rs931515120 Chr.2: 131050410-131050410 on Build GRCh38
rs938816477 Chr.2: 131050275-131050275 on Build GRCh38
rs890236685 Chr.2: 131050377-131050377 on Build GRCh38
rs1021666205 Chr.2: 131050160-131050160 on Build GRCh38
rs557297585 Chr.2: 131050504-131050505 on Build GRCh38
rs571737274 Chr.2: 131050138-131050138 on Build GRCh38
rs369614058 Chr.2: 131050136-131050137 on Build GRCh38
rs893703216 Chr.2: 131050403-131050403 on Build GRCh38
rs908118579 Chr.2: 131050307-131050307 on Build GRCh38
rs942326532 Chr.2: 131050343-131050343 on Build GRCh38
rs542105164 Chr.2: 131050265-131050265 on Build GRCh38
rs963896640 Chr.2: 131050549-131050549 on Build GRCh38
rs928708385 Chr.2: 131050268-131050268 on Build GRCh38
rs1028243736 Chr.2: 131050582-131050582 on Build GRCh38
rs767596901 Chr.2: 131050136-131050136 on Build GRCh38
rs1035770114 Chr.2: 131050483-131050483 on Build GRCh38
rs1037930382 Chr.2: 131050344-131050344 on Build GRCh38
rs528178613 Chr.2: 131050462-131050462 on Build GRCh38
rs74379666 Chr.2: 131050409-131050409 on Build GRCh38
rs893691503 Chr.2: 131050583-131050583 on Build GRCh38
rs960190569 Chr.2: 131050501-131050501 on Build GRCh38
rs984007583 Chr.2: 131050505-131050505 on Build GRCh38
rs966478152 Chr.2: 131050367-131050367 on Build GRCh38
rs534303816 Chr.2: 131050240-131050240 on Build GRCh38
rs371222744 Chr.2: 131050490-131050490 on Build GRCh38
rs991761418 Chr.2: 131050395-131050395 on Build GRCh38
rs931651642 Chr.2: 131050351-131050351 on Build GRCh38
rs1049014275 Chr.2: 131050423-131050423 on Build GRCh38
rs973586733 Chr.2: 131050565-131050565 on Build GRCh38
rs189176824 Chr.2: 131050459-131050459 on Build GRCh38
rs913663638 Chr.2: 131050354-131050354 on Build GRCh38
rs750433795 Chr.2: 131050284-131050284 on Build GRCh38
rs997220068 Chr.2: 131050579-131050579 on Build GRCh38
rs990536581 Chr.2: 131050137-131050137 on Build GRCh38
rs559108976 Chr.2: 131050141-131050141 on Build GRCh38
rs1023576623 Chr.2: 131050440-131050440 on Build GRCh38
rs999760990 Chr.2: 131050379-131050379 on Build GRCh38
rs139066381 Chr.2: 131050392-131050392 on Build GRCh38
rs759461760 Chr.2: 131050393-131050393 on Build GRCh38
rs922194862 Chr.2: 131050574-131050574 on Build GRCh38
rs931640108 Chr.2: 131050591-131050591 on Build GRCh38
rs900152339 Chr.2: 131050348-131050348 on Build GRCh38
rs368499053 Chr.2: 131050457-131050457 on Build GRCh38
rs548839743 Chr.2: 131050568-131050568 on Build GRCh38
rs957905456 Chr.2: 131050291-131050291 on Build GRCh38
rs111578594 Chr.2: 131050537-131050537 on Build GRCh38

View Full Product Details