Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 131050515-131051025 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM168B
Entrez Gene ID
130074
Gene Name
family with sequence similarity 168 member B
Gene Aliases
MANI
UniGene
Hs.534679
Gene Chromosome Location
Chr.2: 131047876-131093468 on Build GRCh38
Transcripts
NM_001009993.3 NM_001321743.1 NM_001321744.1 NM_001321745.1 NM_001321746.1 NM_001321747.1 NM_001321748.1 XM_011510594.2 XM_017003327.1 XM_017003328.1

SNP Detail

SNP ID Chromosome Location
rs946525007 Chr.2: 131050871-131050871 on Build GRCh38
rs542334982 Chr.2: 131050905-131050905 on Build GRCh38
rs963543740 Chr.2: 131050785-131050785 on Build GRCh38
rs754902297 Chr.2: 131050573-131050573 on Build GRCh38
rs139831749 Chr.2: 131050994-131050994 on Build GRCh38
rs752552764 Chr.2: 131050630-131050630 on Build GRCh38
rs911535316 Chr.2: 131050827-131050827 on Build GRCh38
rs994944583 Chr.2: 131050991-131050991 on Build GRCh38
rs553506808 Chr.2: 131050550-131050551 on Build GRCh38
rs1042329944 Chr.2: 131050984-131050984 on Build GRCh38
rs908766482 Chr.2: 131050884-131050884 on Build GRCh38
rs577142616 Chr.2: 131050802-131050802 on Build GRCh38
rs534119308 Chr.2: 131050995-131050995 on Build GRCh38
rs984373366 Chr.2: 131050876-131050876 on Build GRCh38
rs940185933 Chr.2: 131050773-131050773 on Build GRCh38
rs899283117 Chr.2: 131050977-131050977 on Build GRCh38
rs10201624 Chr.2: 131050896-131050896 on Build GRCh38
rs758449101 Chr.2: 131050992-131050992 on Build GRCh38
rs1035718184 Chr.2: 131050768-131050768 on Build GRCh38
rs953653314 Chr.2: 131050822-131050822 on Build GRCh38
rs865992884 Chr.2: 131050816-131050816 on Build GRCh38
rs765169584 Chr.2: 131050598-131050598 on Build GRCh38
rs1052628517 Chr.2: 131050644-131050644 on Build GRCh38
rs960326231 Chr.2: 131050770-131050770 on Build GRCh38
rs535672870 Chr.2: 131050800-131050800 on Build GRCh38
rs935123135 Chr.2: 131050612-131050612 on Build GRCh38
rs11691178 Chr.2: 131050804-131050804 on Build GRCh38
rs1001534069 Chr.2: 131050752-131050752 on Build GRCh38
rs376151537 Chr.2: 131050877-131050877 on Build GRCh38
rs932687083 Chr.2: 131050792-131050792 on Build GRCh38
rs114987054 Chr.2: 131050870-131050870 on Build GRCh38
rs117566269 Chr.2: 131050775-131050775 on Build GRCh38
rs963896640 Chr.2: 131050549-131050549 on Build GRCh38
rs771737221 Chr.2: 131050653-131050658 on Build GRCh38
rs902406045 Chr.2: 131050988-131050988 on Build GRCh38
rs867809259 Chr.2: 131050718-131050718 on Build GRCh38
rs576554111 Chr.2: 131050889-131050889 on Build GRCh38
rs1028243736 Chr.2: 131050582-131050582 on Build GRCh38
rs534021405 Chr.2: 131050960-131050960 on Build GRCh38
rs990574960 Chr.2: 131050868-131050868 on Build GRCh38
rs566563556 Chr.2: 131050951-131050951 on Build GRCh38
rs370138517 Chr.2: 131050666-131050666 on Build GRCh38
rs1003491345 Chr.2: 131050919-131050919 on Build GRCh38
rs893691503 Chr.2: 131050583-131050583 on Build GRCh38
rs145937400 Chr.2: 131050826-131050826 on Build GRCh38
rs758249845 Chr.2: 131050771-131050771 on Build GRCh38
rs905144527 Chr.2: 131050751-131050751 on Build GRCh38
rs868194039 Chr.2: 131050874-131050874 on Build GRCh38
rs893619957 Chr.2: 131050656-131050656 on Build GRCh38
rs911610522 Chr.2: 131050605-131050605 on Build GRCh38
rs1034784390 Chr.2: 131050859-131050859 on Build GRCh38
rs973586733 Chr.2: 131050565-131050565 on Build GRCh38
rs746720323 Chr.2: 131050812-131050812 on Build GRCh38
rs959027352 Chr.2: 131050721-131050721 on Build GRCh38
rs149916701 Chr.2: 131050721-131050721 on Build GRCh38
rs997220068 Chr.2: 131050579-131050579 on Build GRCh38
rs1020822096 Chr.2: 131050635-131050635 on Build GRCh38
rs1034731953 Chr.2: 131050719-131050719 on Build GRCh38
rs149142730 Chr.2: 131050983-131050984 on Build GRCh38
rs563110032 Chr.2: 131050858-131050858 on Build GRCh38
rs1052153805 Chr.2: 131050867-131050867 on Build GRCh38
rs922194862 Chr.2: 131050574-131050574 on Build GRCh38
rs931640108 Chr.2: 131050591-131050591 on Build GRCh38
rs554823947 Chr.2: 131050764-131050764 on Build GRCh38
rs376390702 Chr.2: 131050689-131050690 on Build GRCh38
rs765475669 Chr.2: 131050928-131050928 on Build GRCh38
rs548839743 Chr.2: 131050568-131050568 on Build GRCh38
rs574528653 Chr.2: 131050993-131050993 on Build GRCh38
rs1056802213 Chr.2: 131050924-131050924 on Build GRCh38
rs114441663 Chr.2: 131050976-131050976 on Build GRCh38
rs546043262 Chr.2: 131050851-131050851 on Build GRCh38

View Full Product Details