Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 138014524-138015066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNMT
Entrez Gene ID
3176
Gene Name
histamine N-methyltransferase
Gene Aliases
HMT, HNMT-S1, HNMT-S2, MRT51
UniGene
Hs.42151
Gene Chromosome Location
Chr.2: 137964068-138016364 on Build GRCh38
Transcripts
NM_006895.2 XM_011511064.2 XM_017003948.1

SNP Detail

SNP ID Chromosome Location
rs976555703 Chr.2: 138014883-138014883 on Build GRCh38
rs374217730 Chr.2: 138014594-138014594 on Build GRCh38
rs528350543 Chr.2: 138015000-138015000 on Build GRCh38
rs996338466 Chr.2: 138014686-138014686 on Build GRCh38
rs151225151 Chr.2: 138014566-138014566 on Build GRCh38
rs893837699 Chr.2: 138014658-138014659 on Build GRCh38
rs768440182 Chr.2: 138014675-138014675 on Build GRCh38
rs1034474095 Chr.2: 138014774-138014774 on Build GRCh38
rs911174194 Chr.2: 138014846-138014846 on Build GRCh38
rs1012896617 Chr.2: 138014662-138014662 on Build GRCh38
rs563148016 Chr.2: 138014768-138014768 on Build GRCh38
rs944378229 Chr.2: 138014977-138014977 on Build GRCh38
rs967965612 Chr.2: 138014922-138014922 on Build GRCh38
rs959188286 Chr.2: 138014783-138014783 on Build GRCh38
rs990699480 Chr.2: 138014872-138014872 on Build GRCh38
rs900638270 Chr.2: 138014652-138014652 on Build GRCh38
rs915123172 Chr.2: 138014893-138014893 on Build GRCh38
rs1028314250 Chr.2: 138014704-138014704 on Build GRCh38
rs545798347 Chr.2: 138014853-138014853 on Build GRCh38
rs981487462 Chr.2: 138014770-138014770 on Build GRCh38
rs902203412 Chr.2: 138014574-138014574 on Build GRCh38
rs1053900656 Chr.2: 138014959-138014959 on Build GRCh38
rs888063784 Chr.2: 138014705-138014705 on Build GRCh38
rs1040096228 Chr.2: 138015003-138015003 on Build GRCh38
rs561932563 Chr.2: 138014573-138014573 on Build GRCh38
rs1014898250 Chr.2: 138014761-138014761 on Build GRCh38
rs972363096 Chr.2: 138014767-138014767 on Build GRCh38
rs575376161 Chr.2: 138014640-138014640 on Build GRCh38
rs985777185 Chr.2: 138014820-138014820 on Build GRCh38
rs532150011 Chr.2: 138014838-138014838 on Build GRCh38
rs924001086 Chr.2: 138014920-138014920 on Build GRCh38
rs1035400706 Chr.2: 138014706-138014706 on Build GRCh38
rs775516525 Chr.2: 138014755-138014755 on Build GRCh38
rs1053720546 Chr.2: 138014602-138014602 on Build GRCh38
rs553476503 Chr.2: 138015036-138015036 on Build GRCh38
rs139176343 Chr.2: 138014966-138014966 on Build GRCh38
rs16840132 Chr.2: 138014765-138014765 on Build GRCh38
rs1040105534 Chr.2: 138014609-138014609 on Build GRCh38
rs376078510 Chr.2: 138015032-138015032 on Build GRCh38
rs978425847 Chr.2: 138014947-138014947 on Build GRCh38
rs879257724 Chr.2: 138014663-138014663 on Build GRCh38
rs952434408 Chr.2: 138014806-138014806 on Build GRCh38

View Full Product Details