Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162361520-162362041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
UniGene
Hs.377894
Gene Chromosome Location
Chr.2: 162318768-162371605 on Build GRCh38
Transcripts
NM_012198.3 XM_005246446.2 XM_006712398.3 XM_006712400.3 XM_011510926.2 XM_011510928.2 XM_017003767.1

SNP Detail

SNP ID Chromosome Location
rs944358218 Chr.2: 162361663-162361663 on Build GRCh38
rs762198602 Chr.2: 162361933-162361933 on Build GRCh38
rs982130528 Chr.2: 162361601-162361601 on Build GRCh38
rs1050107841 Chr.2: 162361627-162361627 on Build GRCh38
rs559715861 Chr.2: 162361619-162361619 on Build GRCh38
rs572818921 Chr.2: 162361902-162361902 on Build GRCh38
rs894268877 Chr.2: 162361837-162361837 on Build GRCh38
rs1005563714 Chr.2: 162361556-162361556 on Build GRCh38
rs1023736955 Chr.2: 162361547-162361547 on Build GRCh38
rs970691842 Chr.2: 162361557-162361557 on Build GRCh38
rs937021857 Chr.2: 162361647-162361647 on Build GRCh38
rs929142111 Chr.2: 162361897-162361897 on Build GRCh38
rs569066055 Chr.2: 162361845-162361845 on Build GRCh38
rs953647921 Chr.2: 162361937-162361937 on Build GRCh38
rs34199765 Chr.2: 162361983-162361983 on Build GRCh38
rs867168740 Chr.2: 162361568-162361568 on Build GRCh38
rs144571022 Chr.2: 162361870-162361870 on Build GRCh38
rs529614167 Chr.2: 162361993-162361993 on Build GRCh38
rs770414768 Chr.2: 162361594-162361595 on Build GRCh38
rs746640268 Chr.2: 162361544-162361544 on Build GRCh38
rs962642372 Chr.2: 162361617-162361617 on Build GRCh38
rs1015376674 Chr.2: 162361914-162361914 on Build GRCh38
rs373641642 Chr.2: 162362018-162362018 on Build GRCh38
rs1032782822 Chr.2: 162361673-162361673 on Build GRCh38
rs547170019 Chr.2: 162361836-162361836 on Build GRCh38
rs906642732 Chr.2: 162361854-162361854 on Build GRCh38
rs749609714 Chr.2: 162361848-162361848 on Build GRCh38
rs186858153 Chr.2: 162361982-162361982 on Build GRCh38
rs769450228 Chr.2: 162361725-162361726 on Build GRCh38
rs920720162 Chr.2: 162361620-162361620 on Build GRCh38
rs372479542 Chr.2: 162361855-162361855 on Build GRCh38
rs899837053 Chr.2: 162361955-162361955 on Build GRCh38
rs752956232 Chr.2: 162361658-162361658 on Build GRCh38
rs908111291 Chr.2: 162361612-162361612 on Build GRCh38
rs933961318 Chr.2: 162361961-162361961 on Build GRCh38
rs903095004 Chr.2: 162361706-162361706 on Build GRCh38
rs1033574596 Chr.2: 162362000-162362000 on Build GRCh38
rs774603485 Chr.2: 162361797-162361797 on Build GRCh38
rs113030817 Chr.2: 162361965-162361965 on Build GRCh38
rs944495673 Chr.2: 162361996-162361996 on Build GRCh38
rs539730793 Chr.2: 162361849-162361849 on Build GRCh38
rs1054383559 Chr.2: 162361808-162361808 on Build GRCh38
rs892923937 Chr.2: 162362010-162362010 on Build GRCh38
rs977055010 Chr.2: 162362012-162362012 on Build GRCh38
rs991191926 Chr.2: 162361732-162361732 on Build GRCh38
rs879103340 Chr.2: 162361736-162361736 on Build GRCh38
rs1027919751 Chr.2: 162361936-162361936 on Build GRCh38
rs554926191 Chr.2: 162361947-162361947 on Build GRCh38
rs1041396257 Chr.2: 162361697-162361697 on Build GRCh38
rs1012773538 Chr.2: 162361839-162361839 on Build GRCh38
rs528721734 Chr.2: 162361621-162361621 on Build GRCh38
rs557242911 Chr.2: 162361703-162361703 on Build GRCh38
rs998943163 Chr.2: 162361672-162361672 on Build GRCh38
rs182912309 Chr.2: 162361932-162361932 on Build GRCh38

View Full Product Details