Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174380685-174381182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIR1
Entrez Gene ID
9541
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
CIR
UniGene
Hs.632531
Gene Chromosome Location
Chr.2: 174348150-174395715 on Build GRCh38
Transcripts
NM_004882.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1009733 c.253-4T>G p.? Chr.2: 174380719-174380719 on Build GRCh38
7155425 c.265G>A p.E89K Chr.2: 174380703-174380703 on Build GRCh38
3570106 c.252+2T>C p.? Chr.2: 174380802-174380802 on Build GRCh38
4582816 c.277G>A p.E93K Chr.2: 174380691-174380691 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs548066149 Chr.2: 174381078-174381078 on Build GRCh38
rs200562118 Chr.2: 174380719-174380719 on Build GRCh38
rs201729042 Chr.2: 174380902-174380902 on Build GRCh38
rs753888108 Chr.2: 174380795-174380795 on Build GRCh38
rs188363097 Chr.2: 174380980-174380980 on Build GRCh38
rs753381459 Chr.2: 174380727-174380727 on Build GRCh38
rs373143920 Chr.2: 174380743-174380743 on Build GRCh38
rs749893172 Chr.2: 174380999-174380999 on Build GRCh38
rs992505831 Chr.2: 174381105-174381105 on Build GRCh38
rs749357380 Chr.2: 174380782-174380782 on Build GRCh38
rs376701335 Chr.2: 174380954-174380957 on Build GRCh38
rs763810407 Chr.2: 174380824-174380824 on Build GRCh38
rs1050434125 Chr.2: 174380955-174380955 on Build GRCh38
rs560843690 Chr.2: 174380892-174380892 on Build GRCh38
rs550923261 Chr.2: 174381016-174381016 on Build GRCh38
rs772628379 Chr.2: 174380794-174380794 on Build GRCh38
rs563514111 Chr.2: 174380949-174380951 on Build GRCh38
rs1053895076 Chr.2: 174380894-174380894 on Build GRCh38
rs200952103 Chr.2: 174380862-174380862 on Build GRCh38
rs373421470 Chr.2: 174381045-174381045 on Build GRCh38
rs936860037 Chr.2: 174381138-174381138 on Build GRCh38
rs779758795 Chr.2: 174380755-174380755 on Build GRCh38
rs757262101 Chr.2: 174380729-174380729 on Build GRCh38
rs758006078 Chr.2: 174381133-174381133 on Build GRCh38
rs778749470 Chr.2: 174380738-174380738 on Build GRCh38
rs769980974 Chr.2: 174380771-174380771 on Build GRCh38
rs542179858 Chr.2: 174380884-174380884 on Build GRCh38
rs761712781 Chr.2: 174380887-174380887 on Build GRCh38
rs765894566 Chr.2: 174380807-174380808 on Build GRCh38
rs557207666 Chr.2: 174380822-174380822 on Build GRCh38
rs990889988 Chr.2: 174380764-174380764 on Build GRCh38
rs376227677 Chr.2: 174380788-174380788 on Build GRCh38
rs560992947 Chr.2: 174381046-174381046 on Build GRCh38
rs774695042 Chr.2: 174381051-174381051 on Build GRCh38
rs994406756 Chr.2: 174381028-174381028 on Build GRCh38
rs891045167 Chr.2: 174380944-174380944 on Build GRCh38
rs952589595 Chr.2: 174380774-174380774 on Build GRCh38
rs748436436 Chr.2: 174380768-174380768 on Build GRCh38
rs764846860 Chr.2: 174380910-174380913 on Build GRCh38
rs574926191 Chr.2: 174380825-174380825 on Build GRCh38
rs183779134 Chr.2: 174380940-174380940 on Build GRCh38
rs764748351 Chr.2: 174380834-174380834 on Build GRCh38
rs138237113 Chr.2: 174380900-174380902 on Build GRCh38
rs918210157 Chr.2: 174380770-174380770 on Build GRCh38
rs370241477 Chr.2: 174380723-174380723 on Build GRCh38
rs372088051 Chr.2: 174380955-174380958 on Build GRCh38
rs540066509 Chr.2: 174380963-174380963 on Build GRCh38
rs1008884939 Chr.2: 174381000-174381000 on Build GRCh38
rs1026730813 Chr.2: 174381040-174381040 on Build GRCh38
rs1015455107 Chr.2: 174381009-174381009 on Build GRCh38
rs754372739 Chr.2: 174380736-174380736 on Build GRCh38
rs34190829 Chr.2: 174381139-174381140 on Build GRCh38
rs755821475 Chr.2: 174380722-174380722 on Build GRCh38
rs760016641 Chr.2: 174380793-174380793 on Build GRCh38
rs36115469 Chr.2: 174380902-174380904 on Build GRCh38
rs765262620 Chr.2: 174380899-174380900 on Build GRCh38
rs946247956 Chr.2: 174380874-174380874 on Build GRCh38
rs535159948 Chr.2: 174381144-174381144 on Build GRCh38
rs959108011 Chr.2: 174381053-174381053 on Build GRCh38
rs193292635 Chr.2: 174381059-174381059 on Build GRCh38
rs531837092 Chr.2: 174380952-174380955 on Build GRCh38
rs549311121 Chr.2: 174381101-174381101 on Build GRCh38
rs770990428 Chr.2: 174380786-174380786 on Build GRCh38
rs796088263 Chr.2: 174380904-174380904 on Build GRCh38
rs567662787 Chr.2: 174381102-174381102 on Build GRCh38
rs142894686 Chr.2: 174380986-174380986 on Build GRCh38
rs1025036434 Chr.2: 174381067-174381067 on Build GRCh38
rs190171771 Chr.2: 174380805-174380805 on Build GRCh38
rs897025199 Chr.2: 174380985-174380985 on Build GRCh38
rs758143266 Chr.2: 174380747-174380747 on Build GRCh38
rs750332925 Chr.2: 174380853-174380853 on Build GRCh38

View Full Product Details