Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 174395418-174395907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIR1
Entrez Gene ID
9541
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
CIR
UniGene
Hs.632531
Gene Chromosome Location
Chr.2: 174348150-174395715 on Build GRCh38
Transcripts
NM_004882.3 NM_001193528.1 NM_024583.4 XM_005246853.2 XM_005246854.4 XM_005246855.2 XM_005246856.2 XM_005246857.2 XM_011511839.2 XM_017004909.1 XM_017004910.1

Gene Symbol
SCRN3
Entrez Gene ID
79634
Gene Name
secernin 3
Gene Aliases
SES3
UniGene
Hs.470679
Gene Chromosome Location
Chr.2: 174395729-174430071 on Build GRCh38
Transcripts
NM_004882.3 NM_001193528.1 NM_024583.4 XM_005246853.2 XM_005246854.4 XM_005246855.2 XM_005246856.2 XM_005246857.2 XM_011511839.2 XM_017004909.1 XM_017004910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4479171 c.22C>T p.P8S Chr.2: 174395778-174395778 on Build GRCh38
274552 c.69A>C p.K23N Chr.2: 174395554-174395554 on Build GRCh38
3570107 c.33G>A p.K11K Chr.2: 174395590-174395590 on Build GRCh38
5353937 c.72delA p.V25fs*18 Chr.2: 174395551-174395551 on Build GRCh38
5004046 c.61+10G>A p.? Chr.2: 174395827-174395827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs719161 Chr.2: 174395627-174395627 on Build GRCh38
rs754924140 Chr.2: 174395698-174395698 on Build GRCh38
rs79388135 Chr.2: 174395733-174395733 on Build GRCh38
rs367756576 Chr.2: 174395664-174395664 on Build GRCh38
rs955981819 Chr.2: 174395879-174395879 on Build GRCh38
rs112137665 Chr.2: 174395773-174395773 on Build GRCh38
rs564734463 Chr.2: 174395632-174395632 on Build GRCh38
rs750160899 Chr.2: 174395830-174395830 on Build GRCh38
rs3739149 Chr.2: 174395496-174395496 on Build GRCh38
rs535450442 Chr.2: 174395500-174395501 on Build GRCh38
rs182671080 Chr.2: 174395715-174395715 on Build GRCh38
rs756562167 Chr.2: 174395709-174395709 on Build GRCh38
rs889119072 Chr.2: 174395701-174395701 on Build GRCh38
rs1027179720 Chr.2: 174395774-174395774 on Build GRCh38
rs754970904 Chr.2: 174395537-174395537 on Build GRCh38
rs371090849 Chr.2: 174395486-174395486 on Build GRCh38
rs111342617 Chr.2: 174395684-174395684 on Build GRCh38
rs536221654 Chr.2: 174395876-174395876 on Build GRCh38
rs924550408 Chr.2: 174395859-174395859 on Build GRCh38
rs200177533 Chr.2: 174395507-174395507 on Build GRCh38
rs768457283 Chr.2: 174395728-174395728 on Build GRCh38
rs769652150 Chr.2: 174395744-174395744 on Build GRCh38
rs200881306 Chr.2: 174395749-174395749 on Build GRCh38
rs532739938 Chr.2: 174395729-174395729 on Build GRCh38
rs752693865 Chr.2: 174395786-174395786 on Build GRCh38
rs866654461 Chr.2: 174395607-174395607 on Build GRCh38
rs568832269 Chr.2: 174395469-174395469 on Build GRCh38
rs1041915375 Chr.2: 174395658-174395658 on Build GRCh38
rs140675205 Chr.2: 174395646-174395646 on Build GRCh38
rs781732770 Chr.2: 174395541-174395541 on Build GRCh38
rs370797697 Chr.2: 174395611-174395611 on Build GRCh38
rs765522551 Chr.2: 174395829-174395829 on Build GRCh38
rs757580199 Chr.2: 174395716-174395716 on Build GRCh38
rs542725549 Chr.2: 174395713-174395713 on Build GRCh38
rs777068455 Chr.2: 174395685-174395685 on Build GRCh38
rs142122274 Chr.2: 174395574-174395574 on Build GRCh38
rs571799847 Chr.2: 174395441-174395441 on Build GRCh38
rs371398064 Chr.2: 174395650-174395650 on Build GRCh38
rs766678626 Chr.2: 174395661-174395661 on Build GRCh38
rs756504474 Chr.2: 174395572-174395572 on Build GRCh38
rs758689823 Chr.2: 174395528-174395528 on Build GRCh38
rs1025475525 Chr.2: 174395817-174395817 on Build GRCh38
rs368265963 Chr.2: 174395515-174395515 on Build GRCh38
rs958370860 Chr.2: 174395884-174395884 on Build GRCh38
rs748994097 Chr.2: 174395587-174395587 on Build GRCh38
rs751669563 Chr.2: 174395536-174395536 on Build GRCh38
rs767689720 Chr.2: 174395763-174395763 on Build GRCh38
rs538637334 Chr.2: 174395444-174395444 on Build GRCh38
rs984038997 Chr.2: 174395871-174395871 on Build GRCh38
rs773585701 Chr.2: 174395877-174395877 on Build GRCh38
rs775748167 Chr.2: 174395554-174395554 on Build GRCh38
rs779664150 Chr.2: 174395855-174395855 on Build GRCh38
rs753313612 Chr.2: 174395669-174395669 on Build GRCh38
rs775515187 Chr.2: 174395628-174395628 on Build GRCh38
rs753086532 Chr.2: 174395556-174395556 on Build GRCh38
rs761281708 Chr.2: 174395789-174395789 on Build GRCh38
rs568794275 Chr.2: 174395820-174395820 on Build GRCh38
rs771845957 Chr.2: 174395800-174395807 on Build GRCh38
rs759515379 Chr.2: 174395500-174395500 on Build GRCh38
rs746472271 Chr.2: 174395858-174395858 on Build GRCh38
rs528662753 Chr.2: 174395719-174395719 on Build GRCh38
rs765671888 Chr.2: 174395810-174395810 on Build GRCh38
rs764613618 Chr.2: 174395795-174395795 on Build GRCh38
rs544488763 Chr.2: 174395662-174395662 on Build GRCh38
rs754296434 Chr.2: 174395813-174395813 on Build GRCh38
rs775253511 Chr.2: 174395514-174395514 on Build GRCh38
rs768563138 Chr.2: 174395644-174395644 on Build GRCh38
rs778431893 Chr.2: 174395603-174395603 on Build GRCh38
rs916974846 Chr.2: 174395648-174395648 on Build GRCh38
rs1025509354 Chr.2: 174395802-174395802 on Build GRCh38
rs1054195650 Chr.2: 174395679-174395679 on Build GRCh38
rs759673854 Chr.2: 174395758-174395758 on Build GRCh38
rs766625368 Chr.2: 174395529-174395529 on Build GRCh38
rs1007638618 Chr.2: 174395446-174395446 on Build GRCh38
rs546749376 Chr.2: 174395721-174395721 on Build GRCh38
rs762382756 Chr.2: 174395760-174395760 on Build GRCh38
rs970753540 Chr.2: 174395838-174395838 on Build GRCh38
rs565466377 Chr.2: 174395722-174395722 on Build GRCh38
rs375964359 Chr.2: 174395663-174395663 on Build GRCh38
rs978461986 Chr.2: 174395850-174395850 on Build GRCh38
rs757973669 Chr.2: 174395832-174395832 on Build GRCh38
rs750679444 Chr.2: 174395526-174395526 on Build GRCh38
rs747757531 Chr.2: 174395834-174395834 on Build GRCh38
rs770545829 Chr.2: 174395590-174395590 on Build GRCh38
rs550345011 Chr.2: 174395455-174395455 on Build GRCh38
rs746947455 Chr.2: 174395459-174395459 on Build GRCh38
rs995691406 Chr.2: 174395766-174395766 on Build GRCh38

View Full Product Details