Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 176119207-176119750 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXD10
Entrez Gene ID
3236
Gene Name
homeobox D10
Gene Aliases
HOX4, HOX4D, HOX4E, Hox-4.4
UniGene
Hs.123070
Gene Chromosome Location
Chr.2: 176116764-176119942 on Build GRCh38
Transcripts
NM_002148.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4878518 c.1003G>A p.A335T Chr.2: 176119211-176119211 on Build GRCh38
717649 c.1005C>A p.A335A Chr.2: 176119213-176119213 on Build GRCh38
4086938 c.1020T>G p.S340S Chr.2: 176119228-176119228 on Build GRCh38
3570196 c.1015T>C p.F339L Chr.2: 176119223-176119223 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756628680 Chr.2: 176119231-176119241 on Build GRCh38
rs886055160 Chr.2: 176119462-176119465 on Build GRCh38
rs931706986 Chr.2: 176119287-176119287 on Build GRCh38
rs925547565 Chr.2: 176119639-176119639 on Build GRCh38
rs576578841 Chr.2: 176119375-176119375 on Build GRCh38
rs1050607335 Chr.2: 176119610-176119610 on Build GRCh38
rs532592274 Chr.2: 176119699-176119699 on Build GRCh38
rs61070787 Chr.2: 176119286-176119286 on Build GRCh38
rs180928138 Chr.2: 176119637-176119637 on Build GRCh38
rs777750832 Chr.2: 176119440-176119440 on Build GRCh38
rs748072688 Chr.2: 176119282-176119282 on Build GRCh38
rs760677821 Chr.2: 176119253-176119253 on Build GRCh38
rs189196087 Chr.2: 176119582-176119582 on Build GRCh38
rs67520913 Chr.2: 176119436-176119437 on Build GRCh38
rs1047822073 Chr.2: 176119408-176119408 on Build GRCh38
rs768070742 Chr.2: 176119280-176119280 on Build GRCh38
rs1006190198 Chr.2: 176119461-176119466 on Build GRCh38
rs143734503 Chr.2: 176119453-176119466 on Build GRCh38
rs1038058305 Chr.2: 176119463-176119466 on Build GRCh38
rs1027385857 Chr.2: 176119430-176119430 on Build GRCh38
rs150556295 Chr.2: 176119510-176119510 on Build GRCh38
rs1039183428 Chr.2: 176119342-176119345 on Build GRCh38
rs923376552 Chr.2: 176119351-176119351 on Build GRCh38
rs952848509 Chr.2: 176119433-176119433 on Build GRCh38
rs71409068 Chr.2: 176119435-176119436 on Build GRCh38
rs962360665 Chr.2: 176119420-176119420 on Build GRCh38
rs1034406618 Chr.2: 176119462-176119462 on Build GRCh38
rs374054273 Chr.2: 176119337-176119337 on Build GRCh38
rs757743019 Chr.2: 176119267-176119267 on Build GRCh38
rs536574279 Chr.2: 176119291-176119291 on Build GRCh38
rs1035474856 Chr.2: 176119515-176119515 on Build GRCh38
rs964975488 Chr.2: 176119680-176119680 on Build GRCh38
rs1009944095 Chr.2: 176119563-176119564 on Build GRCh38
rs558709855 Chr.2: 176119307-176119307 on Build GRCh38
rs1050159244 Chr.2: 176119293-176119293 on Build GRCh38
rs1008365837 Chr.2: 176119314-176119314 on Build GRCh38
rs767843035 Chr.2: 176119242-176119242 on Build GRCh38
rs750772082 Chr.2: 176119273-176119273 on Build GRCh38
rs866981654 Chr.2: 176119698-176119698 on Build GRCh38
rs931637898 Chr.2: 176119556-176119556 on Build GRCh38
rs35586190 Chr.2: 176119643-176119644 on Build GRCh38
rs886055161 Chr.2: 176119466-176119466 on Build GRCh38
rs867263804 Chr.2: 176119464-176119464 on Build GRCh38
rs933402948 Chr.2: 176119378-176119378 on Build GRCh38
rs754325981 Chr.2: 176119259-176119259 on Build GRCh38
rs772869682 Chr.2: 176119540-176119540 on Build GRCh38
rs375226686 Chr.2: 176119257-176119257 on Build GRCh38
rs570165195 Chr.2: 176119252-176119252 on Build GRCh38
rs748118788 Chr.2: 176119499-176119499 on Build GRCh38
rs867143553 Chr.2: 176119439-176119439 on Build GRCh38
rs886055159 Chr.2: 176119464-176119465 on Build GRCh38
rs968823143 Chr.2: 176119618-176119618 on Build GRCh38
rs769828708 Chr.2: 176119283-176119283 on Build GRCh38
rs113041382 Chr.2: 176119698-176119699 on Build GRCh38
rs995488163 Chr.2: 176119443-176119443 on Build GRCh38
rs779290864 Chr.2: 176119268-176119268 on Build GRCh38
rs1043109481 Chr.2: 176119285-176119285 on Build GRCh38
rs769503768 Chr.2: 176119231-176119231 on Build GRCh38
rs6147035 Chr.2: 176119465-176119466 on Build GRCh38
rs886444573 Chr.2: 176119437-176119438 on Build GRCh38
rs1002040804 Chr.2: 176119435-176119435 on Build GRCh38
rs905778529 Chr.2: 176119490-176119491 on Build GRCh38
rs757341101 Chr.2: 176119363-176119363 on Build GRCh38
rs768524606 Chr.2: 176119676-176119676 on Build GRCh38
rs13403839 Chr.2: 176119726-176119726 on Build GRCh38
rs897407762 Chr.2: 176119402-176119402 on Build GRCh38
rs868079494 Chr.2: 176119437-176119437 on Build GRCh38
rs759890279 Chr.2: 176119241-176119241 on Build GRCh38
rs116807022 Chr.2: 176119233-176119233 on Build GRCh38
rs1019956641 Chr.2: 176119564-176119567 on Build GRCh38
rs76652386 Chr.2: 176119466-176119466 on Build GRCh38
rs758113477 Chr.2: 176119277-176119277 on Build GRCh38
rs540598489 Chr.2: 176119635-176119635 on Build GRCh38
rs565659273 Chr.2: 176119509-176119509 on Build GRCh38
rs114746583 Chr.2: 176119278-176119278 on Build GRCh38
rs779201977 Chr.2: 176119501-176119501 on Build GRCh38
rs34995639 Chr.2: 176119684-176119685 on Build GRCh38
rs550795371 Chr.2: 176119709-176119709 on Build GRCh38
rs113753022 Chr.2: 176119450-176119450 on Build GRCh38
rs200449718 Chr.2: 176119455-176119466 on Build GRCh38
rs944226022 Chr.2: 176119691-176119691 on Build GRCh38
rs994202274 Chr.2: 176119411-176119411 on Build GRCh38
rs756537008 Chr.2: 176119230-176119231 on Build GRCh38

View Full Product Details