Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 183095251-183095778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP19
Entrez Gene ID
142679
Gene Name
dual specificity phosphatase 19
Gene Aliases
DUSP17, LMWDSP3, SKRP1, TS-DSP1
UniGene
Hs.132237
Gene Chromosome Location
Chr.2: 183078559-183100008 on Build GRCh38
Transcripts
NM_001142314.1 NM_001321519.1 NM_080876.3 NR_135688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
74463 c.532T>C p.S178P Chr.2: 183095536-183095536 on Build GRCh38
3575017 c.427-1G>A p.? Chr.2: 183095430-183095430 on Build GRCh38
5960703 c.650C>T p.S217L Chr.2: 183095654-183095654 on Build GRCh38
3695142 c.620A>G p.N207S Chr.2: 183095624-183095624 on Build GRCh38
476634 c.577T>C p.F193L Chr.2: 183095581-183095581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772878671 Chr.2: 183095411-183095412 on Build GRCh38
rs145225950 Chr.2: 183095519-183095519 on Build GRCh38
rs757529411 Chr.2: 183095699-183095699 on Build GRCh38
rs753832511 Chr.2: 183095432-183095432 on Build GRCh38
rs2675070 Chr.2: 183095305-183095305 on Build GRCh38
rs752233807 Chr.2: 183095629-183095629 on Build GRCh38
rs113637602 Chr.2: 183095482-183095482 on Build GRCh38
rs369540552 Chr.2: 183095415-183095416 on Build GRCh38
rs773891858 Chr.2: 183095416-183095416 on Build GRCh38
rs758873162 Chr.2: 183095566-183095566 on Build GRCh38
rs186460064 Chr.2: 183095419-183095419 on Build GRCh38
rs780501461 Chr.2: 183095670-183095670 on Build GRCh38
rs144839699 Chr.2: 183095459-183095459 on Build GRCh38
rs373293671 Chr.2: 183095469-183095469 on Build GRCh38
rs945445298 Chr.2: 183095742-183095742 on Build GRCh38
rs769827455 Chr.2: 183095586-183095586 on Build GRCh38
rs754152797 Chr.2: 183095642-183095644 on Build GRCh38
rs202043149 Chr.2: 183095431-183095431 on Build GRCh38
rs776778271 Chr.2: 183095634-183095634 on Build GRCh38
rs768070376 Chr.2: 183095416-183095417 on Build GRCh38
rs1003374103 Chr.2: 183095537-183095537 on Build GRCh38
rs747272881 Chr.2: 183095485-183095485 on Build GRCh38
rs143295197 Chr.2: 183095624-183095624 on Build GRCh38
rs1053934097 Chr.2: 183095702-183095702 on Build GRCh38
rs768846446 Chr.2: 183095489-183095489 on Build GRCh38
rs770962122 Chr.2: 183095544-183095544 on Build GRCh38
rs758929033 Chr.2: 183095650-183095650 on Build GRCh38
rs748381584 Chr.2: 183095689-183095689 on Build GRCh38
rs746429428 Chr.2: 183095565-183095565 on Build GRCh38
rs764513770 Chr.2: 183095606-183095607 on Build GRCh38
rs765228730 Chr.2: 183095493-183095493 on Build GRCh38
rs771489024 Chr.2: 183095476-183095476 on Build GRCh38
rs16823987 Chr.2: 183095652-183095652 on Build GRCh38
rs564715672 Chr.2: 183095415-183095415 on Build GRCh38
rs972819968 Chr.2: 183095618-183095618 on Build GRCh38
rs768899774 Chr.2: 183095577-183095577 on Build GRCh38
rs546721279 Chr.2: 183095589-183095589 on Build GRCh38
rs191055792 Chr.2: 183095463-183095463 on Build GRCh38
rs773333552 Chr.2: 183095587-183095587 on Build GRCh38
rs761762447 Chr.2: 183095490-183095490 on Build GRCh38
rs369033410 Chr.2: 183095464-183095464 on Build GRCh38
rs915322991 Chr.2: 183095738-183095738 on Build GRCh38
rs534821816 Chr.2: 183095628-183095628 on Build GRCh38
rs774307614 Chr.2: 183095597-183095597 on Build GRCh38
rs755787807 Chr.2: 183095631-183095631 on Build GRCh38
rs758067276 Chr.2: 183095437-183095437 on Build GRCh38
rs754724910 Chr.2: 183095515-183095515 on Build GRCh38
rs976885985 Chr.2: 183095333-183095337 on Build GRCh38
rs377469252 Chr.2: 183095680-183095680 on Build GRCh38
rs1056952397 Chr.2: 183095447-183095447 on Build GRCh38
rs749870641 Chr.2: 183095384-183095384 on Build GRCh38
rs111726180 Chr.2: 183095427-183095428 on Build GRCh38
rs746248531 Chr.2: 183095481-183095481 on Build GRCh38
rs770023394 Chr.2: 183095700-183095700 on Build GRCh38
rs78011252 Chr.2: 183095427-183095427 on Build GRCh38
rs199585473 Chr.2: 183095728-183095728 on Build GRCh38
rs779487619 Chr.2: 183095528-183095528 on Build GRCh38
rs573841927 Chr.2: 183095688-183095688 on Build GRCh38
rs771308590 Chr.2: 183095388-183095388 on Build GRCh38
rs779869888 Chr.2: 183095438-183095438 on Build GRCh38
rs751311577 Chr.2: 183095455-183095455 on Build GRCh38
rs762557070 Chr.2: 183095415-183095415 on Build GRCh38
rs954256479 Chr.2: 183095731-183095731 on Build GRCh38
rs748932935 Chr.2: 183095351-183095351 on Build GRCh38
rs78404316 Chr.2: 183095363-183095363 on Build GRCh38
rs770484426 Chr.2: 183095740-183095740 on Build GRCh38
rs201526202 Chr.2: 183095567-183095567 on Build GRCh38
rs756565339 Chr.2: 183095418-183095419 on Build GRCh38
rs57818913 Chr.2: 183095426-183095427 on Build GRCh38
rs767227404 Chr.2: 183095517-183095517 on Build GRCh38
rs766340119 Chr.2: 183095512-183095512 on Build GRCh38
rs760874498 Chr.2: 183095429-183095429 on Build GRCh38
rs750266893 Chr.2: 183095436-183095436 on Build GRCh38
rs961126713 Chr.2: 183095612-183095612 on Build GRCh38
rs763021481 Chr.2: 183095591-183095591 on Build GRCh38
rs776653198 Chr.2: 183095580-183095580 on Build GRCh38
rs542212638 Chr.2: 183095303-183095303 on Build GRCh38
rs373510412 Chr.2: 183095677-183095677 on Build GRCh38
rs142968986 Chr.2: 183095518-183095518 on Build GRCh38
rs765039924 Chr.2: 183095433-183095433 on Build GRCh38
rs372197241 Chr.2: 183095404-183095404 on Build GRCh38
rs774826016 Chr.2: 183095411-183095411 on Build GRCh38
rs528229079 Chr.2: 183095347-183095347 on Build GRCh38
rs542197769 Chr.2: 183095508-183095508 on Build GRCh38
rs369154833 Chr.2: 183095585-183095585 on Build GRCh38
rs559925578 Chr.2: 183095356-183095356 on Build GRCh38
rs557829011 Chr.2: 183095648-183095648 on Build GRCh38
rs772968986 Chr.2: 183095504-183095504 on Build GRCh38
rs35941903 Chr.2: 183095749-183095749 on Build GRCh38
rs748226150 Chr.2: 183095513-183095513 on Build GRCh38
rs775622751 Chr.2: 183095428-183095428 on Build GRCh38

View Full Product Details