Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 187345132-187345665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
UniGene
Hs.470882
Gene Chromosome Location
Chr.2: 187341963-187448307 on Build GRCh38
Transcripts
NM_001271751.1 NM_005795.5 XM_005246231.3 XM_005246232.3 XM_005246234.3 XM_017003151.1

SNP Detail

SNP ID Chromosome Location
rs543117140 Chr.2: 187345254-187345254 on Build GRCh38
rs989571560 Chr.2: 187345526-187345526 on Build GRCh38
rs796793500 Chr.2: 187345252-187345252 on Build GRCh38
rs756020343 Chr.2: 187345392-187345392 on Build GRCh38
rs916666229 Chr.2: 187345534-187345534 on Build GRCh38
rs935166231 Chr.2: 187345640-187345640 on Build GRCh38
rs772339230 Chr.2: 187345508-187345508 on Build GRCh38
rs1033457668 Chr.2: 187345618-187345618 on Build GRCh38
rs539217011 Chr.2: 187345266-187345266 on Build GRCh38
rs185098690 Chr.2: 187345479-187345479 on Build GRCh38
rs140455559 Chr.2: 187345211-187345214 on Build GRCh38
rs940285407 Chr.2: 187345338-187345338 on Build GRCh38
rs918198549 Chr.2: 187345322-187345322 on Build GRCh38
rs751596116 Chr.2: 187345369-187345369 on Build GRCh38
rs1019210175 Chr.2: 187345422-187345422 on Build GRCh38
rs747138354 Chr.2: 187345593-187345593 on Build GRCh38
rs1046352674 Chr.2: 187345330-187345330 on Build GRCh38
rs992806641 Chr.2: 187345280-187345280 on Build GRCh38
rs1026744182 Chr.2: 187345380-187345380 on Build GRCh38
rs1020290636 Chr.2: 187345183-187345183 on Build GRCh38
rs777804093 Chr.2: 187345566-187345566 on Build GRCh38
rs193150283 Chr.2: 187345371-187345371 on Build GRCh38
rs113345773 Chr.2: 187345601-187345601 on Build GRCh38
rs696092 Chr.2: 187345487-187345487 on Build GRCh38
rs948114135 Chr.2: 187345582-187345582 on Build GRCh38
rs562793352 Chr.2: 187345529-187345529 on Build GRCh38
rs967754946 Chr.2: 187345212-187345212 on Build GRCh38
rs938582409 Chr.2: 187345352-187345352 on Build GRCh38
rs966044748 Chr.2: 187345444-187345444 on Build GRCh38
rs533027963 Chr.2: 187345530-187345530 on Build GRCh38
rs553873959 Chr.2: 187345359-187345359 on Build GRCh38
rs979177425 Chr.2: 187345238-187345238 on Build GRCh38
rs145758812 Chr.2: 187345478-187345478 on Build GRCh38
rs965636546 Chr.2: 187345205-187345205 on Build GRCh38
rs578138774 Chr.2: 187345173-187345173 on Build GRCh38
rs369659548 Chr.2: 187345394-187345394 on Build GRCh38
rs1020643442 Chr.2: 187345180-187345180 on Build GRCh38
rs1036964460 Chr.2: 187345339-187345339 on Build GRCh38
rs1019251568 Chr.2: 187345411-187345411 on Build GRCh38
rs898510792 Chr.2: 187345348-187345348 on Build GRCh38
rs766039156 Chr.2: 187345230-187345230 on Build GRCh38
rs542631132 Chr.2: 187345409-187345409 on Build GRCh38

View Full Product Details