Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 187346241-187346743 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALCRL
Entrez Gene ID
10203
Gene Name
calcitonin receptor like receptor
Gene Aliases
CGRPR, CRLR
UniGene
Hs.470882
Gene Chromosome Location
Chr.2: 187341963-187448307 on Build GRCh38
Transcripts
NM_001271751.1 NM_005795.5 XM_005246231.3 XM_005246232.3 XM_005246234.3 XM_017003151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1660959 c.1324A>T p.N442Y Chr.2: 187346246-187346246 on Build GRCh38
6156322 c.1312A>G p.S438G Chr.2: 187346258-187346258 on Build GRCh38
3575273 c.1328G>A p.G443E Chr.2: 187346242-187346242 on Build GRCh38
5590040 c.1327G>A p.G443R Chr.2: 187346243-187346243 on Build GRCh38
3575274 c.1315G>A p.E439K Chr.2: 187346255-187346255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777499186 Chr.2: 187346433-187346433 on Build GRCh38
rs184264594 Chr.2: 187346431-187346431 on Build GRCh38
rs371222442 Chr.2: 187346284-187346284 on Build GRCh38
rs751731212 Chr.2: 187346402-187346402 on Build GRCh38
rs905406059 Chr.2: 187346520-187346520 on Build GRCh38
rs3771095 Chr.2: 187346385-187346385 on Build GRCh38
rs547591919 Chr.2: 187346500-187346500 on Build GRCh38
rs777141533 Chr.2: 187346330-187346330 on Build GRCh38
rs1020297416 Chr.2: 187346712-187346712 on Build GRCh38
rs936534641 Chr.2: 187346531-187346531 on Build GRCh38
rs150400535 Chr.2: 187346391-187346391 on Build GRCh38
rs965387732 Chr.2: 187346692-187346692 on Build GRCh38
rs1027068673 Chr.2: 187346595-187346595 on Build GRCh38
rs976625006 Chr.2: 187346654-187346654 on Build GRCh38
rs750793098 Chr.2: 187346389-187346389 on Build GRCh38
rs376143930 Chr.2: 187346698-187346698 on Build GRCh38
rs773778926 Chr.2: 187346320-187346320 on Build GRCh38
rs922393729 Chr.2: 187346335-187346335 on Build GRCh38
rs201275836 Chr.2: 187346328-187346328 on Build GRCh38
rs766602483 Chr.2: 187346392-187346392 on Build GRCh38
rs146718835 Chr.2: 187346313-187346313 on Build GRCh38
rs761424831 Chr.2: 187346314-187346314 on Build GRCh38
rs532641771 Chr.2: 187346481-187346481 on Build GRCh38
rs840599 Chr.2: 187346569-187346569 on Build GRCh38
rs10931283 Chr.2: 187346716-187346716 on Build GRCh38
rs752699369 Chr.2: 187346409-187346409 on Build GRCh38
rs201789608 Chr.2: 187346321-187346321 on Build GRCh38
rs188797129 Chr.2: 187346533-187346533 on Build GRCh38
rs755140887 Chr.2: 187346407-187346407 on Build GRCh38
rs1025180256 Chr.2: 187346341-187346341 on Build GRCh38
rs780066182 Chr.2: 187346572-187346572 on Build GRCh38
rs147063602 Chr.2: 187346618-187346618 on Build GRCh38
rs755016433 Chr.2: 187346283-187346283 on Build GRCh38
rs976848948 Chr.2: 187346419-187346419 on Build GRCh38
rs1018970707 Chr.2: 187346621-187346621 on Build GRCh38
rs748847754 Chr.2: 187346590-187346590 on Build GRCh38
rs749099077 Chr.2: 187346444-187346444 on Build GRCh38
rs577981659 Chr.2: 187346663-187346664 on Build GRCh38
rs977133774 Chr.2: 187346693-187346693 on Build GRCh38
rs577862332 Chr.2: 187346679-187346679 on Build GRCh38
rs779489226 Chr.2: 187346420-187346423 on Build GRCh38
rs549732183 Chr.2: 187346394-187346394 on Build GRCh38
rs565897064 Chr.2: 187346505-187346505 on Build GRCh38
rs748059138 Chr.2: 187346309-187346309 on Build GRCh38
rs756053175 Chr.2: 187346413-187346413 on Build GRCh38
rs913329548 Chr.2: 187346469-187346469 on Build GRCh38
rs770616041 Chr.2: 187346318-187346318 on Build GRCh38
rs961556216 Chr.2: 187346345-187346345 on Build GRCh38
rs113360035 Chr.2: 187346577-187346577 on Build GRCh38
rs947004142 Chr.2: 187346498-187346499 on Build GRCh38
rs986094049 Chr.2: 187346607-187346607 on Build GRCh38
rs528274609 Chr.2: 187346617-187346617 on Build GRCh38
rs994320895 Chr.2: 187346354-187346354 on Build GRCh38
rs76176669 Chr.2: 187346602-187346602 on Build GRCh38
rs762248625 Chr.2: 187346353-187346353 on Build GRCh38
rs750440495 Chr.2: 187346504-187346504 on Build GRCh38
rs554691096 Chr.2: 187346568-187346568 on Build GRCh38
rs946131255 Chr.2: 187346483-187346483 on Build GRCh38
rs894804726 Chr.2: 187346538-187346538 on Build GRCh38
rs767656620 Chr.2: 187346408-187346408 on Build GRCh38
rs748900537 Chr.2: 187346317-187346317 on Build GRCh38
rs922414438 Chr.2: 187346685-187346688 on Build GRCh38
rs112628833 Chr.2: 187346564-187346564 on Build GRCh38
rs977891445 Chr.2: 187346491-187346491 on Build GRCh38
rs148965095 Chr.2: 187346278-187346278 on Build GRCh38

View Full Product Details