Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189032512-189033011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A2
Entrez Gene ID
1290
Gene Name
collagen type V alpha 2 chain
Gene Aliases
EDSC
UniGene
Hs.445827
Gene Chromosome Location
Chr.2: 189031915-189225301 on Build GRCh38
Transcripts
NM_000393.3 XM_011510573.2

SNP Detail

SNP ID Chromosome Location
rs1053008419 Chr.2: 189032831-189032831 on Build GRCh38
rs950351253 Chr.2: 189032683-189032683 on Build GRCh38
rs141087839 Chr.2: 189032888-189032888 on Build GRCh38
rs541070195 Chr.2: 189032552-189032552 on Build GRCh38
rs11186 Chr.2: 189032668-189032668 on Build GRCh38
rs886055349 Chr.2: 189032610-189032610 on Build GRCh38
rs921426289 Chr.2: 189032773-189032773 on Build GRCh38
rs1051271990 Chr.2: 189032796-189032796 on Build GRCh38
rs905194313 Chr.2: 189032840-189032840 on Build GRCh38
rs1035632532 Chr.2: 189032672-189032672 on Build GRCh38
rs147584410 Chr.2: 189032824-189032824 on Build GRCh38
rs562447249 Chr.2: 189032564-189032564 on Build GRCh38
rs969848547 Chr.2: 189032642-189032642 on Build GRCh38
rs185683558 Chr.2: 189032967-189032967 on Build GRCh38
rs559091923 Chr.2: 189032636-189032636 on Build GRCh38
rs893184481 Chr.2: 189032652-189032652 on Build GRCh38
rs774898320 Chr.2: 189032837-189032837 on Build GRCh38
rs942406420 Chr.2: 189032692-189032692 on Build GRCh38
rs554868853 Chr.2: 189032858-189032858 on Build GRCh38
rs753977034 Chr.2: 189032693-189032693 on Build GRCh38
rs1035102883 Chr.2: 189032859-189032859 on Build GRCh38
rs781647776 Chr.2: 189032832-189032832 on Build GRCh38
rs141248891 Chr.2: 189032708-189032708 on Build GRCh38
rs1140641 Chr.2: 189032587-189032587 on Build GRCh38
rs1022269978 Chr.2: 189032823-189032823 on Build GRCh38
rs537693025 Chr.2: 189032906-189032906 on Build GRCh38
rs1051324547 Chr.2: 189032561-189032561 on Build GRCh38
rs983086937 Chr.2: 189032689-189032689 on Build GRCh38
rs969950625 Chr.2: 189032669-189032669 on Build GRCh38
rs538758154 Chr.2: 189032764-189032764 on Build GRCh38
rs934930233 Chr.2: 189032813-189032813 on Build GRCh38
rs547728483 Chr.2: 189032772-189032772 on Build GRCh38
rs114330868 Chr.2: 189032691-189032691 on Build GRCh38
rs933800135 Chr.2: 189032755-189032755 on Build GRCh38
rs1044524346 Chr.2: 189032919-189032919 on Build GRCh38
rs886055348 Chr.2: 189032605-189032605 on Build GRCh38
rs562299073 Chr.2: 189032646-189032646 on Build GRCh38
rs891126206 Chr.2: 189032633-189032633 on Build GRCh38
rs13914 Chr.2: 189032905-189032905 on Build GRCh38
rs752412412 Chr.2: 189032793-189032793 on Build GRCh38
rs932865181 Chr.2: 189032549-189032549 on Build GRCh38
rs1140642 Chr.2: 189032585-189032585 on Build GRCh38
rs1031644207 Chr.2: 189032649-189032649 on Build GRCh38
rs73981486 Chr.2: 189032653-189032653 on Build GRCh38
rs146947961 Chr.2: 189032739-189032739 on Build GRCh38
rs930919048 Chr.2: 189032681-189032681 on Build GRCh38
rs1010894671 Chr.2: 189032570-189032570 on Build GRCh38
rs375499247 Chr.2: 189032750-189032750 on Build GRCh38
rs12886 Chr.2: 189032670-189032670 on Build GRCh38
rs565751511 Chr.2: 189032778-189032778 on Build GRCh38
rs752812362 Chr.2: 189032620-189032621 on Build GRCh38
rs1035559027 Chr.2: 189032656-189032657 on Build GRCh38
rs757944809 Chr.2: 189032806-189032806 on Build GRCh38
rs905672968 Chr.2: 189032921-189032921 on Build GRCh38

View Full Product Details