Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 190060313-190060816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf88
Entrez Gene ID
84281
Gene Name
chromosome 2 open reading frame 88
Gene Aliases
smAKAP
UniGene
Hs.389311
Gene Chromosome Location
Chr.2: 189879479-190203484 on Build GRCh38
Transcripts
XM_005246905.1 XM_011511982.1 XM_011511986.2 NM_005259.2

Gene Symbol
MSTN
Entrez Gene ID
2660
Gene Name
myostatin
Gene Aliases
GDF8, MSLHP
UniGene
Hs.41565
Gene Chromosome Location
Chr.2: 190055700-190062729 on Build GRCh38
Transcripts
XM_005246905.1 XM_011511982.1 XM_011511986.2 NM_005259.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4089689 c.461C>T p.A154V Chr.2: 190060348-190060348 on Build GRCh38
3896232 c.425T>G p.F142C Chr.2: 190060384-190060384 on Build GRCh38
1014089 c.441A>G p.Q147Q Chr.2: 190060368-190060368 on Build GRCh38
4089688 c.487G>A p.V163I Chr.2: 190060322-190060322 on Build GRCh38
4985044 c.458A>G p.K153R Chr.2: 190060351-190060351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568057853 Chr.2: 190060451-190060451 on Build GRCh38
rs559214765 Chr.2: 190060525-190060525 on Build GRCh38
rs147813198 Chr.2: 190060434-190060434 on Build GRCh38
rs151205793 Chr.2: 190060718-190060718 on Build GRCh38
rs369029405 Chr.2: 190060636-190060636 on Build GRCh38
rs35493945 Chr.2: 190060343-190060343 on Build GRCh38
rs866258301 Chr.2: 190060635-190060635 on Build GRCh38
rs990854889 Chr.2: 190060670-190060670 on Build GRCh38
rs568890201 Chr.2: 190060520-190060520 on Build GRCh38
rs916623496 Chr.2: 190060673-190060673 on Build GRCh38
rs760227704 Chr.2: 190060566-190060566 on Build GRCh38
rs779712124 Chr.2: 190060395-190060395 on Build GRCh38
rs535148115 Chr.2: 190060473-190060473 on Build GRCh38
rs974542606 Chr.2: 190060721-190060721 on Build GRCh38
rs759857200 Chr.2: 190060448-190060448 on Build GRCh38
rs1018402701 Chr.2: 190060712-190060712 on Build GRCh38
rs920121332 Chr.2: 190060763-190060763 on Build GRCh38
rs771836980 Chr.2: 190060522-190060522 on Build GRCh38
rs1008797390 Chr.2: 190060527-190060527 on Build GRCh38
rs772180319 Chr.2: 190060403-190060403 on Build GRCh38
rs528391865 Chr.2: 190060378-190060378 on Build GRCh38
rs59937732 Chr.2: 190060587-190060587 on Build GRCh38
rs756825446 Chr.2: 190060714-190060714 on Build GRCh38
rs754186571 Chr.2: 190060341-190060341 on Build GRCh38
rs1054272691 Chr.2: 190060624-190060624 on Build GRCh38
rs758969290 Chr.2: 190060356-190060356 on Build GRCh38
rs749046927 Chr.2: 190060396-190060396 on Build GRCh38
rs10184778 Chr.2: 190060565-190060565 on Build GRCh38
rs780648804 Chr.2: 190060366-190060366 on Build GRCh38
rs1019801892 Chr.2: 190060533-190060533 on Build GRCh38
rs760865305 Chr.2: 190060417-190060417 on Build GRCh38
rs957984166 Chr.2: 190060656-190060656 on Build GRCh38
rs955359766 Chr.2: 190060639-190060639 on Build GRCh38
rs773406441 Chr.2: 190060413-190060413 on Build GRCh38
rs748033945 Chr.2: 190060402-190060402 on Build GRCh38
rs755715730 Chr.2: 190060386-190060386 on Build GRCh38
rs1008228210 Chr.2: 190060707-190060707 on Build GRCh38
rs765715434 Chr.2: 190060349-190060349 on Build GRCh38
rs142195885 Chr.2: 190060423-190060423 on Build GRCh38
rs898422665 Chr.2: 190060737-190060739 on Build GRCh38
rs775809829 Chr.2: 190060631-190060631 on Build GRCh38
rs539527155 Chr.2: 190060524-190060524 on Build GRCh38
rs553428678 Chr.2: 190060633-190060633 on Build GRCh38
rs1805086 Chr.2: 190060351-190060351 on Build GRCh38
rs549290019 Chr.2: 190060379-190060379 on Build GRCh38
rs2293284 Chr.2: 190060519-190060519 on Build GRCh38
rs768350810 Chr.2: 190060398-190060398 on Build GRCh38
rs561870363 Chr.2: 190060482-190060482 on Build GRCh38
rs765513543 Chr.2: 190060471-190060471 on Build GRCh38
rs764302205 Chr.2: 190060746-190060746 on Build GRCh38
rs758626586 Chr.2: 190060404-190060404 on Build GRCh38
rs949717014 Chr.2: 190060545-190060545 on Build GRCh38
rs928527580 Chr.2: 190060715-190060715 on Build GRCh38
rs751919666 Chr.2: 190060755-190060755 on Build GRCh38
rs547418756 Chr.2: 190060720-190060720 on Build GRCh38
rs764105684 Chr.2: 190060780-190060780 on Build GRCh38
rs774263948 Chr.2: 190060401-190060401 on Build GRCh38
rs920187101 Chr.2: 190060489-190060489 on Build GRCh38
rs949526292 Chr.2: 190060699-190060699 on Build GRCh38

View Full Product Details