Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 195737942-195738407 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAH7
Entrez Gene ID
56171
Gene Name
dynein axonemal heavy chain 7
Gene Aliases
-
UniGene
Hs.97403
Gene Chromosome Location
Chr.2: 195737604-196075188 on Build GRCh38
Transcripts
NM_018897.2 XM_011511487.2 XM_011511488.2 XM_011511489.2 XM_011511490.2 XM_011511497.2 XM_017004504.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3575744 c.11982G>A p.T3994T Chr.2: 195738014-195738014 on Build GRCh38
3575745 c.11948G>A p.R3983Q Chr.2: 195738048-195738048 on Build GRCh38
3575742 c.12041G>A p.R4014Q Chr.2: 195737955-195737955 on Build GRCh38
3575743 c.12025G>A p.E4009K Chr.2: 195737971-195737971 on Build GRCh38
718746 c.12026A>C p.E4009A Chr.2: 195737970-195737970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780577995 Chr.2: 195738057-195738058 on Build GRCh38
rs543734443 Chr.2: 195738316-195738316 on Build GRCh38
rs80112322 Chr.2: 195738062-195738062 on Build GRCh38
rs930327420 Chr.2: 195738319-195738319 on Build GRCh38
rs770890721 Chr.2: 195738081-195738082 on Build GRCh38
rs374587789 Chr.2: 195738040-195738040 on Build GRCh38
rs774430677 Chr.2: 195738144-195738144 on Build GRCh38
rs529780833 Chr.2: 195738265-195738265 on Build GRCh38
rs549858847 Chr.2: 195738284-195738284 on Build GRCh38
rs542727184 Chr.2: 195738053-195738053 on Build GRCh38
rs781492764 Chr.2: 195738014-195738014 on Build GRCh38
rs924031682 Chr.2: 195738263-195738265 on Build GRCh38
rs781176906 Chr.2: 195738114-195738114 on Build GRCh38
rs539134873 Chr.2: 195738128-195738128 on Build GRCh38
rs1049296150 Chr.2: 195738102-195738102 on Build GRCh38
rs562000709 Chr.2: 195738363-195738366 on Build GRCh38
rs1047772618 Chr.2: 195738322-195738322 on Build GRCh38
rs747777418 Chr.2: 195738028-195738028 on Build GRCh38
rs772950823 Chr.2: 195738042-195738042 on Build GRCh38
rs114621989 Chr.2: 195738049-195738049 on Build GRCh38
rs749068708 Chr.2: 195738077-195738078 on Build GRCh38
rs761060517 Chr.2: 195738175-195738175 on Build GRCh38
rs746430865 Chr.2: 195738015-195738015 on Build GRCh38
rs201569053 Chr.2: 195738078-195738078 on Build GRCh38
rs77570341 Chr.2: 195738315-195738315 on Build GRCh38
rs879079914 Chr.2: 195738309-195738309 on Build GRCh38
rs750898539 Chr.2: 195738068-195738068 on Build GRCh38
rs200924607 Chr.2: 195737995-195737995 on Build GRCh38
rs776717085 Chr.2: 195737965-195737965 on Build GRCh38
rs762880664 Chr.2: 195738130-195738130 on Build GRCh38
rs751818199 Chr.2: 195738065-195738065 on Build GRCh38
rs199830841 Chr.2: 195738105-195738105 on Build GRCh38
rs538618797 Chr.2: 195738290-195738290 on Build GRCh38
rs373653544 Chr.2: 195738067-195738067 on Build GRCh38
rs62203618 Chr.2: 195738048-195738048 on Build GRCh38
rs757642841 Chr.2: 195738001-195738001 on Build GRCh38
rs762361155 Chr.2: 195738029-195738029 on Build GRCh38
rs759634085 Chr.2: 195737973-195737973 on Build GRCh38
rs758824171 Chr.2: 195737970-195737981 on Build GRCh38
rs764152876 Chr.2: 195738063-195738063 on Build GRCh38
rs778435892 Chr.2: 195738027-195738027 on Build GRCh38
rs373707263 Chr.2: 195738296-195738296 on Build GRCh38
rs767846597 Chr.2: 195738148-195738148 on Build GRCh38
rs750737304 Chr.2: 195738170-195738170 on Build GRCh38
rs747618775 Chr.2: 195738094-195738094 on Build GRCh38
rs771578144 Chr.2: 195738031-195738031 on Build GRCh38
rs780505382 Chr.2: 195737984-195737990 on Build GRCh38
rs547987923 Chr.2: 195738129-195738129 on Build GRCh38
rs895889861 Chr.2: 195738335-195738335 on Build GRCh38
rs769327704 Chr.2: 195738059-195738060 on Build GRCh38
rs751828555 Chr.2: 195737997-195737997 on Build GRCh38
rs746736939 Chr.2: 195738100-195738100 on Build GRCh38
rs754480100 Chr.2: 195738018-195738018 on Build GRCh38
rs775657398 Chr.2: 195738059-195738059 on Build GRCh38
rs557345342 Chr.2: 195738234-195738234 on Build GRCh38
rs910758038 Chr.2: 195738200-195738200 on Build GRCh38
rs759568786 Chr.2: 195738153-195738154 on Build GRCh38
rs956870075 Chr.2: 195738285-195738289 on Build GRCh38
rs752089844 Chr.2: 195737987-195737987 on Build GRCh38
rs758739049 Chr.2: 195737992-195737992 on Build GRCh38
rs200947523 Chr.2: 195738087-195738087 on Build GRCh38
rs752967172 Chr.2: 195737990-195737990 on Build GRCh38
rs1014370420 Chr.2: 195738369-195738369 on Build GRCh38
rs943608809 Chr.2: 195738291-195738291 on Build GRCh38
rs955699383 Chr.2: 195738088-195738088 on Build GRCh38
rs1051902951 Chr.2: 195737985-195737985 on Build GRCh38
rs1012245208 Chr.2: 195738208-195738208 on Build GRCh38
rs751811911 Chr.2: 195738183-195738183 on Build GRCh38
rs1037920333 Chr.2: 195738314-195738314 on Build GRCh38
rs754431451 Chr.2: 195738043-195738044 on Build GRCh38
rs774393217 Chr.2: 195738096-195738097 on Build GRCh38
rs968158119 Chr.2: 195738009-195738009 on Build GRCh38
rs765475024 Chr.2: 195737989-195737989 on Build GRCh38
rs777236267 Chr.2: 195738099-195738099 on Build GRCh38
rs912822965 Chr.2: 195738058-195738058 on Build GRCh38
rs868282237 Chr.2: 195738053-195738054 on Build GRCh38
rs534464201 Chr.2: 195738260-195738260 on Build GRCh38
rs746717841 Chr.2: 195738043-195738043 on Build GRCh38
rs757046314 Chr.2: 195737987-195737987 on Build GRCh38
rs993911694 Chr.2: 195738024-195738024 on Build GRCh38
rs116269254 Chr.2: 195738125-195738125 on Build GRCh38
rs979477129 Chr.2: 195738262-195738263 on Build GRCh38
rs777538565 Chr.2: 195738075-195738076 on Build GRCh38
rs987397046 Chr.2: 195738286-195738286 on Build GRCh38
rs968042946 Chr.2: 195738238-195738238 on Build GRCh38
rs768837015 Chr.2: 195738131-195738131 on Build GRCh38
rs1023679710 Chr.2: 195738223-195738223 on Build GRCh38
rs757932879 Chr.2: 195738096-195738096 on Build GRCh38
rs777419950 Chr.2: 195738164-195738165 on Build GRCh38
rs569695983 Chr.2: 195738285-195738285 on Build GRCh38

View Full Product Details