Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196199556-196200053 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

SNP Detail

SNP ID Chromosome Location
rs768193496 Chr.2: 196199646-196199646 on Build GRCh38
rs918376498 Chr.2: 196199914-196199914 on Build GRCh38
rs986376329 Chr.2: 196199830-196199830 on Build GRCh38
rs762507251 Chr.2: 196199854-196199855 on Build GRCh38
rs889485790 Chr.2: 196199649-196199649 on Build GRCh38
rs981545928 Chr.2: 196199941-196199941 on Build GRCh38
rs967451299 Chr.2: 196199766-196199766 on Build GRCh38
rs909353142 Chr.2: 196199610-196199610 on Build GRCh38
rs187282776 Chr.2: 196199663-196199663 on Build GRCh38
rs972198553 Chr.2: 196199894-196199894 on Build GRCh38
rs915981479 Chr.2: 196199779-196199779 on Build GRCh38
rs941132800 Chr.2: 196199644-196199644 on Build GRCh38
rs3762475 Chr.2: 196199774-196199774 on Build GRCh38
rs190918273 Chr.2: 196199679-196199679 on Build GRCh38
rs918261839 Chr.2: 196199677-196199677 on Build GRCh38
rs549392314 Chr.2: 196199931-196199931 on Build GRCh38
rs1053043175 Chr.2: 196199959-196199959 on Build GRCh38
rs540194027 Chr.2: 196199785-196199785 on Build GRCh38
rs773812965 Chr.2: 196199913-196199913 on Build GRCh38
rs568863553 Chr.2: 196199936-196199936 on Build GRCh38
rs900926655 Chr.2: 196199598-196199598 on Build GRCh38
rs144728792 Chr.2: 196199857-196199857 on Build GRCh38
rs1011674498 Chr.2: 196199736-196199737 on Build GRCh38
rs893033798 Chr.2: 196200001-196200001 on Build GRCh38
rs1001448792 Chr.2: 196199715-196199715 on Build GRCh38
rs1045372092 Chr.2: 196199685-196199686 on Build GRCh38
rs1026932167 Chr.2: 196199725-196199725 on Build GRCh38
rs550610930 Chr.2: 196199849-196199849 on Build GRCh38
rs180809062 Chr.2: 196199898-196199898 on Build GRCh38
rs909375055 Chr.2: 196199836-196199836 on Build GRCh38
rs953771896 Chr.2: 196199812-196199812 on Build GRCh38
rs963481254 Chr.2: 196199875-196199875 on Build GRCh38
rs978707926 Chr.2: 196199826-196199826 on Build GRCh38
rs138764990 Chr.2: 196199978-196199978 on Build GRCh38
rs1028059712 Chr.2: 196199643-196199643 on Build GRCh38
rs116640233 Chr.2: 196199708-196199708 on Build GRCh38
rs1038046463 Chr.2: 196199650-196199650 on Build GRCh38
rs998384939 Chr.2: 196199627-196199627 on Build GRCh38
rs948733802 Chr.2: 196199802-196199802 on Build GRCh38
rs761381021 Chr.2: 196199716-196199716 on Build GRCh38
rs1005195565 Chr.2: 196199661-196199661 on Build GRCh38
rs928294356 Chr.2: 196199956-196199956 on Build GRCh38
rs765810996 Chr.2: 196199611-196199611 on Build GRCh38
rs556369873 Chr.2: 196199597-196199597 on Build GRCh38
rs960231635 Chr.2: 196199760-196199760 on Build GRCh38
rs531909236 Chr.2: 196199993-196199993 on Build GRCh38
rs753161532 Chr.2: 196199667-196199667 on Build GRCh38
rs1044148 Chr.2: 196199616-196199616 on Build GRCh38

View Full Product Details