Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196200346-196200844 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

SNP Detail

SNP ID Chromosome Location
rs745762233 Chr.2: 196200375-196200375 on Build GRCh38
rs780108574 Chr.2: 196200448-196200448 on Build GRCh38
rs1023052975 Chr.2: 196200462-196200462 on Build GRCh38
rs190123969 Chr.2: 196200373-196200373 on Build GRCh38
rs527427971 Chr.2: 196200721-196200721 on Build GRCh38
rs554168171 Chr.2: 196200426-196200426 on Build GRCh38
rs538062458 Chr.2: 196200664-196200664 on Build GRCh38
rs762581128 Chr.2: 196200775-196200775 on Build GRCh38
rs918313014 Chr.2: 196200692-196200692 on Build GRCh38
rs993834453 Chr.2: 196200410-196200410 on Build GRCh38
rs541001090 Chr.2: 196200570-196200570 on Build GRCh38
rs955241390 Chr.2: 196200657-196200657 on Build GRCh38
rs540326982 Chr.2: 196200417-196200417 on Build GRCh38
rs545574803 Chr.2: 196200685-196200685 on Build GRCh38
rs1000262367 Chr.2: 196200498-196200498 on Build GRCh38
rs933202730 Chr.2: 196200753-196200753 on Build GRCh38
rs911097608 Chr.2: 196200682-196200682 on Build GRCh38
rs1051823652 Chr.2: 196200435-196200435 on Build GRCh38
rs924447641 Chr.2: 196200722-196200722 on Build GRCh38
rs3748876 Chr.2: 196200691-196200691 on Build GRCh38
rs1026462286 Chr.2: 196200480-196200480 on Build GRCh38
rs529522362 Chr.2: 196200652-196200652 on Build GRCh38
rs993684987 Chr.2: 196200473-196200473 on Build GRCh38
rs896077897 Chr.2: 196200783-196200783 on Build GRCh38
rs577551041 Chr.2: 196200597-196200597 on Build GRCh38
rs137886626 Chr.2: 196200796-196200796 on Build GRCh38
rs768617136 Chr.2: 196200647-196200647 on Build GRCh38
rs758257816 Chr.2: 196200405-196200405 on Build GRCh38
rs188446847 Chr.2: 196200765-196200765 on Build GRCh38
rs968641206 Chr.2: 196200666-196200666 on Build GRCh38
rs4850680 Chr.2: 196200497-196200497 on Build GRCh38
rs199529726 Chr.2: 196200429-196200432 on Build GRCh38
rs899455225 Chr.2: 196200471-196200471 on Build GRCh38
rs182398203 Chr.2: 196200738-196200738 on Build GRCh38
rs896044499 Chr.2: 196200445-196200445 on Build GRCh38
rs374518980 Chr.2: 196200431-196200434 on Build GRCh38
rs3748875 Chr.2: 196200455-196200455 on Build GRCh38
rs763976078 Chr.2: 196200595-196200596 on Build GRCh38
rs991198472 Chr.2: 196200646-196200646 on Build GRCh38
rs941124844 Chr.2: 196200389-196200389 on Build GRCh38
rs982677950 Chr.2: 196200554-196200554 on Build GRCh38
rs574855700 Chr.2: 196200774-196200774 on Build GRCh38
rs1048459844 Chr.2: 196200443-196200443 on Build GRCh38
rs914211267 Chr.2: 196200649-196200649 on Build GRCh38
rs757257374 Chr.2: 196200393-196200393 on Build GRCh38
rs567286690 Chr.2: 196200754-196200754 on Build GRCh38
rs538536673 Chr.2: 196200803-196200803 on Build GRCh38
rs956250025 Chr.2: 196200619-196200619 on Build GRCh38
rs896837872 Chr.2: 196200396-196200396 on Build GRCh38
rs941154555 Chr.2: 196200684-196200684 on Build GRCh38
rs145296439 Chr.2: 196200626-196200626 on Build GRCh38

View Full Product Details