Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196219816-196220349 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4614503 c.4323delT p.F1441fs*16 Chr.2: 196220124-196220124 on Build GRCh38
4764276 c.4388A>G p.N1463S Chr.2: 196220059-196220059 on Build GRCh38
5532257 c.4333G>A p.E1445K Chr.2: 196220114-196220114 on Build GRCh38
7185539 c.4393A>C p.T1465P Chr.2: 196220054-196220054 on Build GRCh38
4912298 c.4375A>G p.S1459G Chr.2: 196220072-196220072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753312009 Chr.2: 196220067-196220067 on Build GRCh38
rs561112004 Chr.2: 196220250-196220250 on Build GRCh38
rs776975365 Chr.2: 196220196-196220196 on Build GRCh38
rs1531114 Chr.2: 196219871-196219871 on Build GRCh38
rs777216163 Chr.2: 196219859-196219859 on Build GRCh38
rs570237987 Chr.2: 196220319-196220319 on Build GRCh38
rs759875255 Chr.2: 196220034-196220034 on Build GRCh38
rs748888621 Chr.2: 196220103-196220103 on Build GRCh38
rs545527605 Chr.2: 196219967-196219967 on Build GRCh38
rs1029264052 Chr.2: 196219888-196219888 on Build GRCh38
rs2305568 Chr.2: 196220258-196220258 on Build GRCh38
rs145284068 Chr.2: 196220310-196220310 on Build GRCh38
rs758948475 Chr.2: 196220071-196220071 on Build GRCh38
rs917226867 Chr.2: 196220280-196220280 on Build GRCh38
rs376196967 Chr.2: 196219964-196219964 on Build GRCh38
rs980406028 Chr.2: 196220300-196220300 on Build GRCh38
rs767052627 Chr.2: 196219984-196219984 on Build GRCh38
rs763162697 Chr.2: 196220052-196220053 on Build GRCh38
rs747056678 Chr.2: 196220188-196220188 on Build GRCh38
rs778658813 Chr.2: 196220109-196220109 on Build GRCh38
rs373242052 Chr.2: 196220189-196220189 on Build GRCh38
rs774578148 Chr.2: 196220303-196220303 on Build GRCh38
rs979074714 Chr.2: 196220257-196220257 on Build GRCh38
rs568799962 Chr.2: 196219945-196219945 on Build GRCh38
rs759661881 Chr.2: 196220203-196220203 on Build GRCh38
rs756975748 Chr.2: 196219969-196219969 on Build GRCh38
rs868330306 Chr.2: 196220143-196220143 on Build GRCh38
rs766579547 Chr.2: 196219991-196219991 on Build GRCh38
rs201066286 Chr.2: 196220029-196220029 on Build GRCh38
rs927153288 Chr.2: 196220301-196220301 on Build GRCh38
rs969973295 Chr.2: 196220171-196220171 on Build GRCh38
rs755743711 Chr.2: 196220096-196220096 on Build GRCh38
rs112491032 Chr.2: 196219948-196219948 on Build GRCh38
rs754860473 Chr.2: 196220108-196220108 on Build GRCh38
rs541467162 Chr.2: 196220157-196220157 on Build GRCh38
rs1531113 Chr.2: 196219863-196219863 on Build GRCh38
rs1027776486 Chr.2: 196220092-196220092 on Build GRCh38
rs779698636 Chr.2: 196220097-196220097 on Build GRCh38
rs1050795172 Chr.2: 196220324-196220324 on Build GRCh38
rs878854424 Chr.2: 196220113-196220113 on Build GRCh38
rs762399108 Chr.2: 196219903-196219903 on Build GRCh38
rs749910927 Chr.2: 196220089-196220089 on Build GRCh38
rs1049256713 Chr.2: 196219941-196219941 on Build GRCh38
rs995425895 Chr.2: 196219917-196219917 on Build GRCh38
rs950012852 Chr.2: 196220287-196220287 on Build GRCh38
rs565418326 Chr.2: 196219978-196219978 on Build GRCh38
rs543315119 Chr.2: 196219884-196219884 on Build GRCh38
rs767174245 Chr.2: 196220082-196220082 on Build GRCh38
rs765511221 Chr.2: 196220046-196220046 on Build GRCh38
rs184161183 Chr.2: 196219877-196219877 on Build GRCh38
rs372766444 Chr.2: 196220292-196220292 on Build GRCh38
rs527782139 Chr.2: 196220017-196220017 on Build GRCh38
rs935868886 Chr.2: 196220302-196220302 on Build GRCh38

View Full Product Details