Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 196222122-196222623 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW2
Entrez Gene ID
57520
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Gene Aliases
NEDL2
UniGene
Hs.654742
Gene Chromosome Location
Chr.2: 196194382-196593692 on Build GRCh38
Transcripts
NM_001304840.1 NM_020760.2 XM_006712646.3 XM_006712648.3 XM_017004563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5953866 c.4129G>C p.E1377Q Chr.2: 196222228-196222228 on Build GRCh38
2905087 c.4128C>T p.N1376N Chr.2: 196222229-196222229 on Build GRCh38
4492896 c.4059C>T p.F1353F Chr.2: 196222298-196222298 on Build GRCh38
4089853 c.4134A>C p.E1378D Chr.2: 196222223-196222223 on Build GRCh38
718697 c.4036C>A p.L1346I Chr.2: 196222321-196222321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369375782 Chr.2: 196222208-196222208 on Build GRCh38
rs751270001 Chr.2: 196222319-196222319 on Build GRCh38
rs149188557 Chr.2: 196222258-196222258 on Build GRCh38
rs375386174 Chr.2: 196222175-196222175 on Build GRCh38
rs3816517 Chr.2: 196222438-196222438 on Build GRCh38
rs776220430 Chr.2: 196222259-196222259 on Build GRCh38
rs546611638 Chr.2: 196222459-196222459 on Build GRCh38
rs909137944 Chr.2: 196222496-196222496 on Build GRCh38
rs758983822 Chr.2: 196222261-196222261 on Build GRCh38
rs762567242 Chr.2: 196222289-196222289 on Build GRCh38
rs535720454 Chr.2: 196222593-196222593 on Build GRCh38
rs368517491 Chr.2: 196222355-196222355 on Build GRCh38
rs769193291 Chr.2: 196222229-196222229 on Build GRCh38
rs780356657 Chr.2: 196222204-196222204 on Build GRCh38
rs749562384 Chr.2: 196222347-196222347 on Build GRCh38
rs372179702 Chr.2: 196222158-196222158 on Build GRCh38
rs756294746 Chr.2: 196222394-196222394 on Build GRCh38
rs775176986 Chr.2: 196222264-196222264 on Build GRCh38
rs965534760 Chr.2: 196222378-196222378 on Build GRCh38
rs1003233776 Chr.2: 196222246-196222246 on Build GRCh38
rs894424081 Chr.2: 196222424-196222424 on Build GRCh38
rs921239492 Chr.2: 196222559-196222559 on Build GRCh38
rs778155321 Chr.2: 196222241-196222241 on Build GRCh38
rs958961764 Chr.2: 196222308-196222308 on Build GRCh38
rs564074442 Chr.2: 196222375-196222375 on Build GRCh38
rs372810440 Chr.2: 196222328-196222328 on Build GRCh38
rs530536903 Chr.2: 196222194-196222194 on Build GRCh38
rs200268472 Chr.2: 196222186-196222186 on Build GRCh38
rs750521410 Chr.2: 196222339-196222339 on Build GRCh38
rs779359684 Chr.2: 196222363-196222363 on Build GRCh38
rs756328127 Chr.2: 196222340-196222340 on Build GRCh38
rs894029333 Chr.2: 196222322-196222322 on Build GRCh38
rs532916300 Chr.2: 196222385-196222385 on Build GRCh38
rs145154786 Chr.2: 196222155-196222155 on Build GRCh38
rs16847486 Chr.2: 196222337-196222337 on Build GRCh38
rs780260651 Chr.2: 196222345-196222345 on Build GRCh38
rs777527425 Chr.2: 196222187-196222187 on Build GRCh38
rs953164927 Chr.2: 196222444-196222444 on Build GRCh38
rs267599146 Chr.2: 196222253-196222253 on Build GRCh38
rs367629745 Chr.2: 196222171-196222171 on Build GRCh38
rs752498435 Chr.2: 196222176-196222176 on Build GRCh38
rs781362800 Chr.2: 196222192-196222192 on Build GRCh38
rs938520399 Chr.2: 196222211-196222211 on Build GRCh38
rs924760660 Chr.2: 196222522-196222522 on Build GRCh38
rs1046918039 Chr.2: 196222592-196222592 on Build GRCh38
rs932556767 Chr.2: 196222563-196222563 on Build GRCh38
rs145667867 Chr.2: 196222265-196222265 on Build GRCh38
rs1046466321 Chr.2: 196222193-196222193 on Build GRCh38
rs144268022 Chr.2: 196222242-196222242 on Build GRCh38
rs16847479 Chr.2: 196222188-196222188 on Build GRCh38
rs368166467 Chr.2: 196222582-196222582 on Build GRCh38
rs751849651 Chr.2: 196222349-196222352 on Build GRCh38
rs763987891 Chr.2: 196222310-196222310 on Build GRCh38
rs944103944 Chr.2: 196222525-196222525 on Build GRCh38
rs772641073 Chr.2: 196222377-196222377 on Build GRCh38
rs375314818 Chr.2: 196222200-196222200 on Build GRCh38
rs566822143 Chr.2: 196222544-196222544 on Build GRCh38
rs113286509 Chr.2: 196222537-196222537 on Build GRCh38

View Full Product Details