Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203957658-203958184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOS
Entrez Gene ID
29851
Gene Name
inducible T-cell costimulator
Gene Aliases
AILIM, CD278, CVID1
UniGene
Hs.56247
Gene Chromosome Location
Chr.2: 203936748-203961577 on Build GRCh38
Transcripts
NM_012092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
390582 c.517G>A p.V173M Chr.2: 203957814-203957814 on Build GRCh38
719701 c.577A>G p.R193G Chr.2: 203957874-203957874 on Build GRCh38
3990995 c.572A>C p.K191T Chr.2: 203957869-203957869 on Build GRCh38
4551655 c.535G>A p.E179K Chr.2: 203957832-203957832 on Build GRCh38
1326509 c.563C>G p.T188R Chr.2: 203957860-203957860 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376095811 Chr.2: 203957919-203957919 on Build GRCh38
rs968135245 Chr.2: 203957771-203957771 on Build GRCh38
rs757933259 Chr.2: 203957774-203957774 on Build GRCh38
rs375035651 Chr.2: 203958010-203958010 on Build GRCh38
rs376555288 Chr.2: 203957819-203957819 on Build GRCh38
rs778519345 Chr.2: 203957735-203957735 on Build GRCh38
rs777163364 Chr.2: 203957983-203957983 on Build GRCh38
rs56366932 Chr.2: 203958107-203958107 on Build GRCh38
rs779182018 Chr.2: 203957923-203957923 on Build GRCh38
rs772210728 Chr.2: 203957935-203957935 on Build GRCh38
rs1037137882 Chr.2: 203958085-203958085 on Build GRCh38
rs778908715 Chr.2: 203957820-203957820 on Build GRCh38
rs758279746 Chr.2: 203957825-203957826 on Build GRCh38
rs771358494 Chr.2: 203957838-203957838 on Build GRCh38
rs889715268 Chr.2: 203958038-203958038 on Build GRCh38
rs369607883 Chr.2: 203957755-203957755 on Build GRCh38
rs1044762811 Chr.2: 203957881-203957881 on Build GRCh38
rs561515783 Chr.2: 203957711-203957711 on Build GRCh38
rs980113377 Chr.2: 203957741-203957741 on Build GRCh38
rs1002155525 Chr.2: 203958156-203958156 on Build GRCh38
rs571259383 Chr.2: 203957980-203957980 on Build GRCh38
rs987007460 Chr.2: 203958030-203958030 on Build GRCh38
rs921546072 Chr.2: 203957752-203957752 on Build GRCh38
rs763476251 Chr.2: 203957776-203957776 on Build GRCh38
rs940859370 Chr.2: 203957960-203957960 on Build GRCh38
rs761295712 Chr.2: 203957898-203957898 on Build GRCh38
rs780068159 Chr.2: 203957913-203957922 on Build GRCh38
rs1006745740 Chr.2: 203957727-203957727 on Build GRCh38
rs181079134 Chr.2: 203958034-203958034 on Build GRCh38
rs750304925 Chr.2: 203957903-203957903 on Build GRCh38
rs371397052 Chr.2: 203957918-203957918 on Build GRCh38
rs370645286 Chr.2: 203957828-203957828 on Build GRCh38
rs755927035 Chr.2: 203957913-203957913 on Build GRCh38
rs767453549 Chr.2: 203957899-203957899 on Build GRCh38
rs761137420 Chr.2: 203957829-203957829 on Build GRCh38
rs371917442 Chr.2: 203957948-203957948 on Build GRCh38
rs552272209 Chr.2: 203957947-203957948 on Build GRCh38
rs373368532 Chr.2: 203957864-203957864 on Build GRCh38
rs780326986 Chr.2: 203957809-203957809 on Build GRCh38
rs1031102664 Chr.2: 203958157-203958157 on Build GRCh38
rs779674689 Chr.2: 203957916-203957916 on Build GRCh38
rs1049237131 Chr.2: 203957912-203957912 on Build GRCh38
rs5837889 Chr.2: 203957948-203957949 on Build GRCh38
rs185783798 Chr.2: 203958057-203958057 on Build GRCh38
rs757222290 Chr.2: 203957784-203957784 on Build GRCh38
rs547175222 Chr.2: 203957877-203957877 on Build GRCh38
rs150573153 Chr.2: 203957813-203957813 on Build GRCh38
rs772773689 Chr.2: 203957827-203957827 on Build GRCh38
rs765042951 Chr.2: 203957761-203957761 on Build GRCh38
rs1022206628 Chr.2: 203958099-203958099 on Build GRCh38
rs781100164 Chr.2: 203957801-203957801 on Build GRCh38
rs148435889 Chr.2: 203957807-203957807 on Build GRCh38
rs550322448 Chr.2: 203958014-203958014 on Build GRCh38
rs200922304 Chr.2: 203957760-203957760 on Build GRCh38
rs916854910 Chr.2: 203957873-203957873 on Build GRCh38
rs909404522 Chr.2: 203957947-203957947 on Build GRCh38
rs931356771 Chr.2: 203958115-203958115 on Build GRCh38
rs373512827 Chr.2: 203957772-203957772 on Build GRCh38
rs751482760 Chr.2: 203957935-203957935 on Build GRCh38
rs748208335 Chr.2: 203957822-203957822 on Build GRCh38
rs948265313 Chr.2: 203957875-203957875 on Build GRCh38
rs960724664 Chr.2: 203957831-203957831 on Build GRCh38
rs397712353 Chr.2: 203957960-203957961 on Build GRCh38
rs751541705 Chr.2: 203957782-203957782 on Build GRCh38
rs999344599 Chr.2: 203958155-203958155 on Build GRCh38
rs755844099 Chr.2: 203957808-203957808 on Build GRCh38
rs1022511401 Chr.2: 203957743-203957743 on Build GRCh38
rs1007187837 Chr.2: 203958043-203958043 on Build GRCh38
rs144520938 Chr.2: 203957872-203957872 on Build GRCh38
rs55649217 Chr.2: 203957686-203957686 on Build GRCh38
rs933671707 Chr.2: 203958013-203958013 on Build GRCh38
rs55663036 Chr.2: 203958000-203958000 on Build GRCh38
rs1051209093 Chr.2: 203957756-203957756 on Build GRCh38
rs987886621 Chr.2: 203957685-203957685 on Build GRCh38
rs377370202 Chr.2: 203957886-203957886 on Build GRCh38
rs62177477 Chr.2: 203958148-203958148 on Build GRCh38

View Full Product Details