Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203959401-203959915 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOS
Entrez Gene ID
29851
Gene Name
inducible T-cell costimulator
Gene Aliases
AILIM, CD278, CVID1
UniGene
Hs.56247
Gene Chromosome Location
Chr.2: 203936748-203961577 on Build GRCh38
Transcripts
NM_012092.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6341025 c.599A>C p.*200S Chr.2: 203959598-203959598 on Build GRCh38
5009591 c.591G>A p.V197V Chr.2: 203959590-203959590 on Build GRCh38
404817 c.593delC p.L199fs? Chr.2: 203959592-203959592 on Build GRCh38
1404693 c.587-1G>T p.? Chr.2: 203959585-203959585 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs78281070 Chr.2: 203959477-203959477 on Build GRCh38
rs750224061 Chr.2: 203959455-203959455 on Build GRCh38
rs146016344 Chr.2: 203959456-203959457 on Build GRCh38
rs575431589 Chr.2: 203959812-203959812 on Build GRCh38
rs769887283 Chr.2: 203959676-203959676 on Build GRCh38
rs987596848 Chr.2: 203959761-203959761 on Build GRCh38
rs1052271321 Chr.2: 203959530-203959530 on Build GRCh38
rs970446044 Chr.2: 203959669-203959669 on Build GRCh38
rs1053358229 Chr.2: 203959879-203959879 on Build GRCh38
rs879011502 Chr.2: 203959456-203959459 on Build GRCh38
rs764972461 Chr.2: 203959507-203959507 on Build GRCh38
rs879447763 Chr.2: 203959620-203959620 on Build GRCh38
rs112172260 Chr.2: 203959478-203959479 on Build GRCh38
rs55655222 Chr.2: 203959590-203959590 on Build GRCh38
rs111644920 Chr.2: 203959588-203959588 on Build GRCh38
rs202010316 Chr.2: 203959457-203959457 on Build GRCh38
rs187152949 Chr.2: 203959865-203959865 on Build GRCh38
rs777371577 Chr.2: 203959529-203959529 on Build GRCh38
rs943395931 Chr.2: 203959791-203959791 on Build GRCh38
rs963224456 Chr.2: 203959685-203959685 on Build GRCh38
rs1033077768 Chr.2: 203959706-203959706 on Build GRCh38
rs568247160 Chr.2: 203959799-203959799 on Build GRCh38
rs903270889 Chr.2: 203959466-203959466 on Build GRCh38
rs111245028 Chr.2: 203959467-203959468 on Build GRCh38
rs759936583 Chr.2: 203959572-203959572 on Build GRCh38
rs919122996 Chr.2: 203959724-203959724 on Build GRCh38
rs867899642 Chr.2: 203959459-203959459 on Build GRCh38
rs1018865594 Chr.2: 203959783-203959783 on Build GRCh38
rs145698501 Chr.2: 203959596-203959596 on Build GRCh38
rs531478782 Chr.2: 203959732-203959732 on Build GRCh38
rs1002223817 Chr.2: 203959423-203959423 on Build GRCh38
rs548691205 Chr.2: 203959867-203959867 on Build GRCh38
rs985365404 Chr.2: 203959500-203959503 on Build GRCh38
rs10172036 Chr.2: 203959560-203959560 on Build GRCh38
rs200367053 Chr.2: 203959615-203959615 on Build GRCh38
rs753634371 Chr.2: 203959606-203959606 on Build GRCh38
rs879019926 Chr.2: 203959456-203959461 on Build GRCh38
rs200852713 Chr.2: 203959453-203959453 on Build GRCh38
rs778288997 Chr.2: 203959684-203959684 on Build GRCh38
rs879167520 Chr.2: 203959458-203959459 on Build GRCh38
rs973217181 Chr.2: 203959720-203959720 on Build GRCh38
rs10183087 Chr.2: 203959601-203959601 on Build GRCh38
rs951228303 Chr.2: 203959809-203959809 on Build GRCh38
rs148800809 Chr.2: 203959481-203959481 on Build GRCh38
rs889174903 Chr.2: 203959728-203959728 on Build GRCh38
rs549965036 Chr.2: 203959796-203959796 on Build GRCh38
rs199913820 Chr.2: 203959452-203959455 on Build GRCh38
rs769306356 Chr.2: 203959457-203959457 on Build GRCh38
rs559764386 Chr.2: 203959630-203959630 on Build GRCh38
rs202183147 Chr.2: 203959639-203959639 on Build GRCh38
rs551768116 Chr.2: 203959657-203959657 on Build GRCh38
rs926517482 Chr.2: 203959450-203959450 on Build GRCh38
rs770140076 Chr.2: 203959650-203959650 on Build GRCh38
rs779006646 Chr.2: 203959616-203959616 on Build GRCh38
rs1029663917 Chr.2: 203959854-203959854 on Build GRCh38
rs758571201 Chr.2: 203959624-203959624 on Build GRCh38
rs546873602 Chr.2: 203959504-203959504 on Build GRCh38
rs1007526165 Chr.2: 203959747-203959747 on Build GRCh38
rs370758705 Chr.2: 203959602-203959602 on Build GRCh38
rs766967127 Chr.2: 203959592-203959592 on Build GRCh38
rs754797113 Chr.2: 203959614-203959614 on Build GRCh38
rs749628535 Chr.2: 203959877-203959877 on Build GRCh38
rs1033463174 Chr.2: 203959477-203959478 on Build GRCh38
rs756570847 Chr.2: 203959672-203959672 on Build GRCh38
rs553609635 Chr.2: 203959622-203959623 on Build GRCh38
rs112626862 Chr.2: 203959477-203959478 on Build GRCh38
rs1039421720 Chr.2: 203959793-203959793 on Build GRCh38
rs778649864 Chr.2: 203959457-203959458 on Build GRCh38
rs73058105 Chr.2: 203959632-203959632 on Build GRCh38
rs777956310 Chr.2: 203959628-203959628 on Build GRCh38
rs911950415 Chr.2: 203959771-203959771 on Build GRCh38
rs936204187 Chr.2: 203959875-203959875 on Build GRCh38
rs752907874 Chr.2: 203959622-203959622 on Build GRCh38
rs796486148 Chr.2: 203959475-203959477 on Build GRCh38
rs34098976 Chr.2: 203959455-203959456 on Build GRCh38
rs142038697 Chr.2: 203959521-203959521 on Build GRCh38
rs747466421 Chr.2: 203959552-203959552 on Build GRCh38

View Full Product Details