Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203961269-203961763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ICOS
Entrez Gene ID
29851
Gene Name
inducible T-cell costimulator
Gene Aliases
AILIM, CD278, CVID1
UniGene
Hs.56247
Gene Chromosome Location
Chr.2: 203936748-203961577 on Build GRCh38
Transcripts
NM_012092.3

SNP Detail

SNP ID Chromosome Location
rs907260903 Chr.2: 203961580-203961580 on Build GRCh38
rs531619607 Chr.2: 203961386-203961386 on Build GRCh38
rs535715577 Chr.2: 203961598-203961598 on Build GRCh38
rs892880226 Chr.2: 203961428-203961428 on Build GRCh38
rs971831035 Chr.2: 203961331-203961331 on Build GRCh38
rs931854904 Chr.2: 203961468-203961468 on Build GRCh38
rs111731634 Chr.2: 203961704-203961704 on Build GRCh38
rs1011278025 Chr.2: 203961437-203961437 on Build GRCh38
rs768122512 Chr.2: 203961691-203961692 on Build GRCh38
rs185636431 Chr.2: 203961719-203961719 on Build GRCh38
rs35586690 Chr.2: 203961706-203961707 on Build GRCh38
rs140979765 Chr.2: 203961453-203961453 on Build GRCh38
rs982084788 Chr.2: 203961631-203961631 on Build GRCh38
rs55727864 Chr.2: 203961691-203961691 on Build GRCh38
rs1002846644 Chr.2: 203961346-203961346 on Build GRCh38
rs1052927667 Chr.2: 203961424-203961424 on Build GRCh38
rs916579063 Chr.2: 203961373-203961373 on Build GRCh38
rs62177480 Chr.2: 203961706-203961706 on Build GRCh38
rs963493872 Chr.2: 203961577-203961577 on Build GRCh38
rs568436539 Chr.2: 203961465-203961465 on Build GRCh38
rs907604352 Chr.2: 203961404-203961404 on Build GRCh38
rs75877336 Chr.2: 203961704-203961704 on Build GRCh38
rs907637122 Chr.2: 203961695-203961695 on Build GRCh38
rs944385951 Chr.2: 203961439-203961439 on Build GRCh38
rs1042205971 Chr.2: 203961557-203961557 on Build GRCh38
rs1054369578 Chr.2: 203961519-203961519 on Build GRCh38
rs777338223 Chr.2: 203961547-203961547 on Build GRCh38
rs570864442 Chr.2: 203961382-203961382 on Build GRCh38
rs934506930 Chr.2: 203961419-203961419 on Build GRCh38
rs747075768 Chr.2: 203961734-203961734 on Build GRCh38
rs927672859 Chr.2: 203961587-203961587 on Build GRCh38
rs958908763 Chr.2: 203961365-203961365 on Build GRCh38
rs951578598 Chr.2: 203961686-203961686 on Build GRCh38
rs576039498 Chr.2: 203961717-203961717 on Build GRCh38
rs1010058438 Chr.2: 203961549-203961549 on Build GRCh38
rs1035031748 Chr.2: 203961352-203961352 on Build GRCh38
rs934557188 Chr.2: 203961689-203961691 on Build GRCh38
rs928509868 Chr.2: 203961418-203961418 on Build GRCh38
rs539568665 Chr.2: 203961703-203961703 on Build GRCh38
rs958691348 Chr.2: 203961606-203961606 on Build GRCh38
rs180928054 Chr.2: 203961682-203961682 on Build GRCh38
rs552385946 Chr.2: 203961375-203961375 on Build GRCh38
rs983631114 Chr.2: 203961399-203961399 on Build GRCh38
rs995754947 Chr.2: 203961665-203961665 on Build GRCh38
rs916612657 Chr.2: 203961596-203961597 on Build GRCh38
rs997748353 Chr.2: 203961462-203961462 on Build GRCh38
rs965785848 Chr.2: 203961697-203961697 on Build GRCh38
rs76784709 Chr.2: 203961692-203961692 on Build GRCh38
rs4675379 Chr.2: 203961372-203961372 on Build GRCh38
rs1038711053 Chr.2: 203961445-203961445 on Build GRCh38
rs1005467629 Chr.2: 203961491-203961491 on Build GRCh38
rs1020346863 Chr.2: 203961308-203961308 on Build GRCh38
rs1040017898 Chr.2: 203961451-203961451 on Build GRCh38

View Full Product Details