Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 206651543-206652047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYTN
Entrez Gene ID
391475
Gene Name
dystrotelin
Gene Aliases
-
UniGene
Hs.640667
Gene Chromosome Location
Chr.2: 206651621-206718396 on Build GRCh38
Transcripts
NM_001093730.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6156917 c.1660C>T p.L554L Chr.2: 206651895-206651895 on Build GRCh38
1631766 c.1642T>C p.S548P Chr.2: 206651913-206651913 on Build GRCh38
6650874 c.1637C>T p.P546L Chr.2: 206651918-206651918 on Build GRCh38
1326494 c.1672G>C p.A558P Chr.2: 206651883-206651883 on Build GRCh38
376748 c.1637C>A p.P546Q Chr.2: 206651918-206651918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1055285821 Chr.2: 206651843-206651843 on Build GRCh38
rs377529149 Chr.2: 206651963-206651963 on Build GRCh38
rs932768255 Chr.2: 206651570-206651570 on Build GRCh38
rs553664414 Chr.2: 206651759-206651759 on Build GRCh38
rs748364166 Chr.2: 206651918-206651918 on Build GRCh38
rs1005706328 Chr.2: 206651898-206651898 on Build GRCh38
rs756737570 Chr.2: 206651856-206651856 on Build GRCh38
rs371213987 Chr.2: 206651957-206651957 on Build GRCh38
rs1029641385 Chr.2: 206651757-206651757 on Build GRCh38
rs200081961 Chr.2: 206651804-206651804 on Build GRCh38
rs35581991 Chr.2: 206651997-206651997 on Build GRCh38
rs945297194 Chr.2: 206651806-206651806 on Build GRCh38
rs779877571 Chr.2: 206651903-206651903 on Build GRCh38
rs769537755 Chr.2: 206651618-206651618 on Build GRCh38
rs745768264 Chr.2: 206651780-206651780 on Build GRCh38
rs1045570812 Chr.2: 206651974-206651985 on Build GRCh38
rs779029478 Chr.2: 206651916-206651916 on Build GRCh38
rs1033427198 Chr.2: 206651960-206651960 on Build GRCh38
rs761233921 Chr.2: 206651821-206651821 on Build GRCh38
rs773627329 Chr.2: 206651810-206651810 on Build GRCh38
rs1002379189 Chr.2: 206651640-206651640 on Build GRCh38
rs749075736 Chr.2: 206651909-206651909 on Build GRCh38
rs1015712535 Chr.2: 206651712-206651712 on Build GRCh38
rs750363921 Chr.2: 206651876-206651876 on Build GRCh38
rs778762981 Chr.2: 206651592-206651595 on Build GRCh38
rs532408366 Chr.2: 206651740-206651740 on Build GRCh38
rs888619923 Chr.2: 206651677-206651677 on Build GRCh38
rs374419183 Chr.2: 206651794-206651794 on Build GRCh38
rs761325413 Chr.2: 206651931-206651931 on Build GRCh38
rs982032185 Chr.2: 206652016-206652016 on Build GRCh38
rs554831382 Chr.2: 206651917-206651917 on Build GRCh38
rs936505114 Chr.2: 206651797-206651797 on Build GRCh38
rs201639314 Chr.2: 206651865-206651865 on Build GRCh38
rs945421448 Chr.2: 206651588-206651588 on Build GRCh38
rs369834015 Chr.2: 206651669-206651669 on Build GRCh38
rs567732933 Chr.2: 206651569-206651569 on Build GRCh38
rs754242640 Chr.2: 206651828-206651828 on Build GRCh38
rs772087336 Chr.2: 206651922-206651922 on Build GRCh38
rs954193799 Chr.2: 206651951-206651951 on Build GRCh38
rs766902018 Chr.2: 206651823-206651823 on Build GRCh38
rs897311429 Chr.2: 206651913-206651913 on Build GRCh38
rs769193114 Chr.2: 206651951-206651951 on Build GRCh38
rs746348595 Chr.2: 206651739-206651739 on Build GRCh38
rs1055782823 Chr.2: 206651628-206651628 on Build GRCh38
rs201045992 Chr.2: 206651900-206651900 on Build GRCh38
rs751556654 Chr.2: 206651673-206651673 on Build GRCh38
rs749556876 Chr.2: 206651796-206651796 on Build GRCh38
rs774570195 Chr.2: 206651798-206651798 on Build GRCh38
rs750269188 Chr.2: 206651924-206651924 on Build GRCh38
rs774849011 Chr.2: 206651626-206651626 on Build GRCh38
rs771560918 Chr.2: 206651950-206651950 on Build GRCh38
rs373373737 Chr.2: 206651832-206651832 on Build GRCh38
rs535249631 Chr.2: 206651635-206651635 on Build GRCh38
rs966503845 Chr.2: 206652007-206652007 on Build GRCh38
rs888240848 Chr.2: 206651874-206651874 on Build GRCh38
rs761817544 Chr.2: 206651802-206651802 on Build GRCh38
rs1005678388 Chr.2: 206651695-206651695 on Build GRCh38
rs926027849 Chr.2: 206651783-206651783 on Build GRCh38
rs368435160 Chr.2: 206651797-206651797 on Build GRCh38
rs764055878 Chr.2: 206651639-206651639 on Build GRCh38
rs192124634 Chr.2: 206651617-206651617 on Build GRCh38
rs769605724 Chr.2: 206651793-206651793 on Build GRCh38
rs895423124 Chr.2: 206651629-206651629 on Build GRCh38
rs985963176 Chr.2: 206651766-206651766 on Build GRCh38
rs777212248 Chr.2: 206651962-206651962 on Build GRCh38
rs998411618 Chr.2: 206651914-206651914 on Build GRCh38
rs546174896 Chr.2: 206651634-206651634 on Build GRCh38
rs560648729 Chr.2: 206651614-206651614 on Build GRCh38
rs768386139 Chr.2: 206651912-206651912 on Build GRCh38
rs566147069 Chr.2: 206651723-206651723 on Build GRCh38
rs373954592 Chr.2: 206651837-206651837 on Build GRCh38
rs751165476 Chr.2: 206651844-206651844 on Build GRCh38

View Full Product Details