Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207529808-207530315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_017003399.1 XM_017003400.1

SNP Detail

SNP ID Chromosome Location
rs1043474743 Chr.2: 207529899-207529899 on Build GRCh38
rs987182550 Chr.2: 207529989-207529989 on Build GRCh38
rs1044984298 Chr.2: 207530177-207530177 on Build GRCh38
rs945995325 Chr.2: 207530095-207530095 on Build GRCh38
rs1027258139 Chr.2: 207529977-207529977 on Build GRCh38
rs922123134 Chr.2: 207530064-207530064 on Build GRCh38
rs886123015 Chr.2: 207530225-207530225 on Build GRCh38
rs975317509 Chr.2: 207529862-207529862 on Build GRCh38
rs1045847057 Chr.2: 207529942-207529942 on Build GRCh38
rs984228210 Chr.2: 207529983-207529983 on Build GRCh38
rs1049002143 Chr.2: 207530229-207530229 on Build GRCh38
rs1042298238 Chr.2: 207530081-207530081 on Build GRCh38
rs911616168 Chr.2: 207529990-207529990 on Build GRCh38
rs963995886 Chr.2: 207529963-207529963 on Build GRCh38
rs368785631 Chr.2: 207530194-207530194 on Build GRCh38
rs146062007 Chr.2: 207529847-207529847 on Build GRCh38
rs950427446 Chr.2: 207529984-207529984 on Build GRCh38
rs966205110 Chr.2: 207530020-207530020 on Build GRCh38
rs946109890 Chr.2: 207529904-207529904 on Build GRCh38
rs1008487638 Chr.2: 207530249-207530249 on Build GRCh38
rs535235961 Chr.2: 207530066-207530066 on Build GRCh38
rs937437966 Chr.2: 207530188-207530188 on Build GRCh38
rs1013067639 Chr.2: 207529952-207529952 on Build GRCh38
rs1046150246 Chr.2: 207530148-207530148 on Build GRCh38
rs1003340145 Chr.2: 207529944-207529944 on Build GRCh38
rs936250308 Chr.2: 207529870-207529870 on Build GRCh38
rs537776419 Chr.2: 207530245-207530245 on Build GRCh38
rs868182226 Chr.2: 207530197-207530197 on Build GRCh38
rs1018549305 Chr.2: 207530275-207530275 on Build GRCh38
rs915060800 Chr.2: 207530056-207530056 on Build GRCh38
rs903181535 Chr.2: 207529923-207529923 on Build GRCh38
rs922184315 Chr.2: 207529864-207529864 on Build GRCh38
rs990438091 Chr.2: 207530067-207530067 on Build GRCh38
rs950406927 Chr.2: 207529982-207529982 on Build GRCh38
rs1038944701 Chr.2: 207529856-207529856 on Build GRCh38
rs994740995 Chr.2: 207529973-207529973 on Build GRCh38
rs1054444922 Chr.2: 207529893-207529893 on Build GRCh38
rs549078229 Chr.2: 207529968-207529968 on Build GRCh38
rs974574139 Chr.2: 207529995-207529995 on Build GRCh38
rs557061644 Chr.2: 207530262-207530262 on Build GRCh38
rs994710635 Chr.2: 207530205-207530205 on Build GRCh38
rs964525774 Chr.2: 207529991-207529991 on Build GRCh38
rs934609415 Chr.2: 207530132-207530132 on Build GRCh38
rs895007273 Chr.2: 207530196-207530196 on Build GRCh38
rs768711416 Chr.2: 207530065-207530065 on Build GRCh38
rs558429035 Chr.2: 207530092-207530092 on Build GRCh38
rs763089965 Chr.2: 207529932-207529932 on Build GRCh38
rs944858305 Chr.2: 207529852-207529852 on Build GRCh38
rs895035691 Chr.2: 207529965-207529965 on Build GRCh38
rs1026535255 Chr.2: 207530195-207530195 on Build GRCh38
rs1050641 Chr.2: 207530072-207530072 on Build GRCh38
rs998973584 Chr.2: 207529924-207529924 on Build GRCh38
rs1026440734 Chr.2: 207529966-207529966 on Build GRCh38
rs907290236 Chr.2: 207529943-207529943 on Build GRCh38
rs928714697 Chr.2: 207530180-207530180 on Build GRCh38
rs976493117 Chr.2: 207530169-207530178 on Build GRCh38
rs950458064 Chr.2: 207529832-207529832 on Build GRCh38
rs913227375 Chr.2: 207530069-207530069 on Build GRCh38
rs1055389740 Chr.2: 207529953-207529953 on Build GRCh38
rs566250318 Chr.2: 207530014-207530014 on Build GRCh38
rs974909985 Chr.2: 207530036-207530036 on Build GRCh38
rs912046138 Chr.2: 207529849-207529849 on Build GRCh38
rs891823692 Chr.2: 207529894-207529894 on Build GRCh38
rs1004582388 Chr.2: 207529985-207529985 on Build GRCh38
rs1057256851 Chr.2: 207530141-207530141 on Build GRCh38
rs1018656687 Chr.2: 207530012-207530012 on Build GRCh38

View Full Product Details