Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207560938-207561348 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_011510650.2 XM_011510651.2 XM_017003399.1 XM_017003400.1 XM_017003401.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5816854 c.284G>T p.R95I Chr.2: 207561141-207561141 on Build GRCh38
4485588 c.294C>T p.S98S Chr.2: 207561151-207561151 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771774509 Chr.2: 207561084-207561084 on Build GRCh38
rs912964610 Chr.2: 207560996-207560996 on Build GRCh38
rs768052771 Chr.2: 207561194-207561194 on Build GRCh38
rs776337772 Chr.2: 207561201-207561202 on Build GRCh38
rs772665320 Chr.2: 207561011-207561011 on Build GRCh38
rs771338448 Chr.2: 207561199-207561202 on Build GRCh38
rs1041525886 Chr.2: 207561043-207561043 on Build GRCh38
rs932674443 Chr.2: 207561006-207561006 on Build GRCh38
rs1005960188 Chr.2: 207561277-207561277 on Build GRCh38
rs569813327 Chr.2: 207561307-207561307 on Build GRCh38
rs775307643 Chr.2: 207561173-207561173 on Build GRCh38
rs1015709813 Chr.2: 207561280-207561280 on Build GRCh38
rs750460311 Chr.2: 207561205-207561205 on Build GRCh38
rs1052912680 Chr.2: 207560973-207560973 on Build GRCh38
rs746912012 Chr.2: 207561111-207561111 on Build GRCh38
rs1012544461 Chr.2: 207561215-207561215 on Build GRCh38
rs776262703 Chr.2: 207561126-207561126 on Build GRCh38
rs372132141 Chr.2: 207561063-207561063 on Build GRCh38
rs961743091 Chr.2: 207561300-207561300 on Build GRCh38
rs150504199 Chr.2: 207561193-207561193 on Build GRCh38
rs778304410 Chr.2: 207561033-207561033 on Build GRCh38
rs35728983 Chr.2: 207561046-207561046 on Build GRCh38
rs763320680 Chr.2: 207561188-207561188 on Build GRCh38
rs773132811 Chr.2: 207561091-207561091 on Build GRCh38
rs756206958 Chr.2: 207561208-207561208 on Build GRCh38
rs768464970 Chr.2: 207561196-207561196 on Build GRCh38
rs770955546 Chr.2: 207561121-207561121 on Build GRCh38
rs760505522 Chr.2: 207561195-207561196 on Build GRCh38
rs747887995 Chr.2: 207561078-207561078 on Build GRCh38
rs982957057 Chr.2: 207561245-207561245 on Build GRCh38
rs759161276 Chr.2: 207561151-207561151 on Build GRCh38
rs752029815 Chr.2: 207561192-207561192 on Build GRCh38
rs764371525 Chr.2: 207561190-207561190 on Build GRCh38
rs200475533 Chr.2: 207561202-207561202 on Build GRCh38
rs747319154 Chr.2: 207561275-207561275 on Build GRCh38
rs952466687 Chr.2: 207561226-207561226 on Build GRCh38
rs531568761 Chr.2: 207561222-207561222 on Build GRCh38
rs893852545 Chr.2: 207561026-207561026 on Build GRCh38
rs1045955633 Chr.2: 207561045-207561046 on Build GRCh38
rs764945260 Chr.2: 207561156-207561156 on Build GRCh38
rs780189583 Chr.2: 207561209-207561209 on Build GRCh38

View Full Product Details