Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222298068-222298563 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC140
Entrez Gene ID
151278
Gene Name
coiled-coil domain containing 140
Gene Aliases
-
UniGene
Hs.350729
Gene Chromosome Location
Chr.2: 222298147-222305217 on Build GRCh38
Transcripts
NM_153038.1 NM_000438.5 NM_001127366.2 NM_013942.4 NM_181457.3 NM_181458.3 NM_181459.3 NM_181460.3 NM_181461.3

Gene Symbol
PAX3
Entrez Gene ID
5077
Gene Name
paired box 3
Gene Aliases
CDHS, HUP2, WS1, WS3
UniGene
Hs.42146
Gene Chromosome Location
Chr.2: 222199887-222298996 on Build GRCh38
Transcripts
NM_153038.1 NM_000438.5 NM_001127366.2 NM_013942.4 NM_181457.3 NM_181458.3 NM_181459.3 NM_181460.3 NM_181461.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5396325 c.76_77CC>TT p.P26L Chr.2: 222298539-222298540 on Build GRCh38
5675703 c.75C>T p.F25F Chr.2: 222298541-222298541 on Build GRCh38
6738206 c.65G>A p.R22H Chr.2: 222298551-222298551 on Build GRCh38
5665107 c.77C>T p.P26L Chr.2: 222298539-222298539 on Build GRCh38
6738207 c.65G>A p.R22H Chr.2: 222298551-222298551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001473111 Chr.2: 222298169-222298169 on Build GRCh38
rs777068833 Chr.2: 222298525-222298525 on Build GRCh38
rs964047184 Chr.2: 222298506-222298506 on Build GRCh38
rs200928747 Chr.2: 222298545-222298545 on Build GRCh38
rs983061716 Chr.2: 222298279-222298279 on Build GRCh38
rs1012468201 Chr.2: 222298139-222298139 on Build GRCh38
rs931238813 Chr.2: 222298277-222298277 on Build GRCh38
rs904034976 Chr.2: 222298162-222298162 on Build GRCh38
rs919807231 Chr.2: 222298241-222298241 on Build GRCh38
rs778034419 Chr.2: 222298510-222298510 on Build GRCh38
rs533746911 Chr.2: 222298188-222298188 on Build GRCh38
rs866429341 Chr.2: 222298546-222298546 on Build GRCh38
rs750227123 Chr.2: 222298487-222298487 on Build GRCh38
rs397987957 Chr.2: 222298119-222298120 on Build GRCh38
rs767371625 Chr.2: 222298484-222298484 on Build GRCh38
rs762987723 Chr.2: 222298106-222298107 on Build GRCh38
rs775063571 Chr.2: 222298105-222298106 on Build GRCh38
rs370833226 Chr.2: 222298538-222298538 on Build GRCh38
rs3835977 Chr.2: 222298107-222298107 on Build GRCh38
rs894135276 Chr.2: 222298120-222298120 on Build GRCh38
rs1057225895 Chr.2: 222298114-222298114 on Build GRCh38
rs1031614976 Chr.2: 222298464-222298464 on Build GRCh38
rs1018963468 Chr.2: 222298438-222298438 on Build GRCh38
rs966078504 Chr.2: 222298470-222298470 on Build GRCh38
rs45614137 Chr.2: 222298184-222298184 on Build GRCh38
rs904151681 Chr.2: 222298361-222298361 on Build GRCh38
rs771417793 Chr.2: 222298519-222298519 on Build GRCh38
rs761586353 Chr.2: 222298353-222298353 on Build GRCh38
rs748220969 Chr.2: 222298496-222298496 on Build GRCh38
rs747001893 Chr.2: 222298180-222298180 on Build GRCh38
rs908354742 Chr.2: 222298282-222298282 on Build GRCh38
rs1001105278 Chr.2: 222298377-222298377 on Build GRCh38
rs964015921 Chr.2: 222298211-222298211 on Build GRCh38
rs938461532 Chr.2: 222298284-222298284 on Build GRCh38
rs770976789 Chr.2: 222298106-222298106 on Build GRCh38
rs894146694 Chr.2: 222298325-222298325 on Build GRCh38
rs781542980 Chr.2: 222298240-222298240 on Build GRCh38
rs756404428 Chr.2: 222298523-222298524 on Build GRCh38
rs758448447 Chr.2: 222298497-222298497 on Build GRCh38
rs954040458 Chr.2: 222298507-222298507 on Build GRCh38
rs1022288577 Chr.2: 222298136-222298136 on Build GRCh38
rs906259712 Chr.2: 222298312-222298312 on Build GRCh38
rs779881016 Chr.2: 222298492-222298492 on Build GRCh38
rs746246231 Chr.2: 222298526-222298526 on Build GRCh38
rs1011145628 Chr.2: 222298390-222298390 on Build GRCh38
rs948354713 Chr.2: 222298343-222298343 on Build GRCh38
rs957029251 Chr.2: 222298181-222298181 on Build GRCh38
rs11397969 Chr.2: 222298107-222298108 on Build GRCh38
rs888104580 Chr.2: 222298239-222298239 on Build GRCh38
rs1022993475 Chr.2: 222298402-222298402 on Build GRCh38
rs867167244 Chr.2: 222298231-222298231 on Build GRCh38
rs553874321 Chr.2: 222298479-222298479 on Build GRCh38
rs978153069 Chr.2: 222298150-222298150 on Build GRCh38
rs775695511 Chr.2: 222298155-222298155 on Build GRCh38
rs984035860 Chr.2: 222298534-222298534 on Build GRCh38
rs78381262 Chr.2: 222298295-222298295 on Build GRCh38
rs999498158 Chr.2: 222298414-222298414 on Build GRCh38
rs570753637 Chr.2: 222298389-222298389 on Build GRCh38
rs756023727 Chr.2: 222298489-222298489 on Build GRCh38
rs747292645 Chr.2: 222298512-222298512 on Build GRCh38
rs892607230 Chr.2: 222298483-222298483 on Build GRCh38
rs895339437 Chr.2: 222298378-222298378 on Build GRCh38
rs967004504 Chr.2: 222298404-222298404 on Build GRCh38
rs932580347 Chr.2: 222298228-222298228 on Build GRCh38
rs768783861 Chr.2: 222298293-222298293 on Build GRCh38
rs45618941 Chr.2: 222298173-222298173 on Build GRCh38

View Full Product Details