Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 224471882-224472385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL3
Entrez Gene ID
8452
Gene Name
cullin 3
Gene Aliases
CUL-3, PHA2E
UniGene
Hs.372286
Gene Chromosome Location
Chr.2: 224470150-224585397 on Build GRCh38
Transcripts
NM_001257197.1 NM_001257198.1 NM_003590.4

SNP Detail

SNP ID Chromosome Location
rs1007952561 Chr.2: 224472057-224472057 on Build GRCh38
rs543811208 Chr.2: 224472312-224472312 on Build GRCh38
rs995971909 Chr.2: 224472078-224472078 on Build GRCh38
rs146425747 Chr.2: 224472150-224472150 on Build GRCh38
rs886055689 Chr.2: 224472152-224472152 on Build GRCh38
rs559063682 Chr.2: 224472106-224472106 on Build GRCh38
rs1027029303 Chr.2: 224472079-224472079 on Build GRCh38
rs1055802710 Chr.2: 224472238-224472239 on Build GRCh38
rs377157866 Chr.2: 224472353-224472353 on Build GRCh38
rs538867247 Chr.2: 224472082-224472082 on Build GRCh38
rs890153682 Chr.2: 224472302-224472302 on Build GRCh38
rs533374759 Chr.2: 224472308-224472311 on Build GRCh38
rs555995940 Chr.2: 224472115-224472115 on Build GRCh38
rs899633662 Chr.2: 224472312-224472314 on Build GRCh38
rs963255245 Chr.2: 224472063-224472063 on Build GRCh38
rs528431659 Chr.2: 224471979-224471979 on Build GRCh38
rs913424704 Chr.2: 224471970-224471970 on Build GRCh38
rs928733060 Chr.2: 224472176-224472176 on Build GRCh38
rs10498161 Chr.2: 224472172-224472172 on Build GRCh38
rs1008046079 Chr.2: 224472301-224472301 on Build GRCh38
rs751347016 Chr.2: 224472258-224472259 on Build GRCh38
rs780174465 Chr.2: 224472330-224472330 on Build GRCh38
rs552315813 Chr.2: 224471928-224471928 on Build GRCh38
rs556914502 Chr.2: 224471973-224471973 on Build GRCh38
rs996510322 Chr.2: 224472334-224472334 on Build GRCh38
rs952330414 Chr.2: 224472097-224472097 on Build GRCh38
rs572505388 Chr.2: 224472127-224472128 on Build GRCh38
rs1005178146 Chr.2: 224472303-224472303 on Build GRCh38
rs886055688 Chr.2: 224472050-224472050 on Build GRCh38
rs770315174 Chr.2: 224472208-224472208 on Build GRCh38
rs1011869070 Chr.2: 224472052-224472055 on Build GRCh38
rs144017210 Chr.2: 224472071-224472071 on Build GRCh38
rs1019400494 Chr.2: 224472302-224472303 on Build GRCh38
rs574916134 Chr.2: 224472251-224472251 on Build GRCh38
rs907220370 Chr.2: 224471975-224471975 on Build GRCh38
rs890373329 Chr.2: 224472050-224472050 on Build GRCh38
rs930885446 Chr.2: 224471918-224471918 on Build GRCh38
rs750546745 Chr.2: 224472250-224472250 on Build GRCh38
rs903083473 Chr.2: 224472286-224472286 on Build GRCh38
rs564393478 Chr.2: 224472182-224472182 on Build GRCh38
rs1042791188 Chr.2: 224472214-224472214 on Build GRCh38
rs1026656474 Chr.2: 224472350-224472350 on Build GRCh38
rs1051459959 Chr.2: 224472027-224472027 on Build GRCh38
rs77211302 Chr.2: 224472064-224472064 on Build GRCh38
rs975085747 Chr.2: 224472065-224472065 on Build GRCh38
rs185186023 Chr.2: 224472237-224472237 on Build GRCh38
rs759079023 Chr.2: 224472031-224472031 on Build GRCh38
rs995847804 Chr.2: 224472018-224472018 on Build GRCh38
rs907994985 Chr.2: 224471933-224471933 on Build GRCh38
rs752490582 Chr.2: 224472039-224472041 on Build GRCh38
rs190478338 Chr.2: 224472362-224472362 on Build GRCh38
rs952451189 Chr.2: 224472358-224472358 on Build GRCh38
rs928679389 Chr.2: 224472116-224472116 on Build GRCh38
rs1055790529 Chr.2: 224472025-224472025 on Build GRCh38
rs1042003568 Chr.2: 224472284-224472284 on Build GRCh38
rs3768898 Chr.2: 224472053-224472053 on Build GRCh38
rs922239893 Chr.2: 224471913-224471913 on Build GRCh38
rs73993899 Chr.2: 224472052-224472052 on Build GRCh38
rs947600979 Chr.2: 224472208-224472208 on Build GRCh38
rs893092876 Chr.2: 224472242-224472242 on Build GRCh38
rs913363674 Chr.2: 224472179-224472179 on Build GRCh38
rs548070553 Chr.2: 224472043-224472043 on Build GRCh38
rs900147779 Chr.2: 224472015-224472015 on Build GRCh38
rs898954615 Chr.2: 224472340-224472340 on Build GRCh38

View Full Product Details