Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229922409-229922905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO36
Entrez Gene ID
130888
Gene Name
F-box protein 36
Gene Aliases
Fbx36
UniGene
Hs.140666
Gene Chromosome Location
Chr.2: 229922491-230013109 on Build GRCh38
Transcripts
NM_174899.4 XM_011512181.2 XM_011512183.2 XM_017005274.1

Gene Symbol
TRIP12
Entrez Gene ID
9320
Gene Name
thyroid hormone receptor interactor 12
Gene Aliases
TRIP-12, ULF
UniGene
Hs.572642
Gene Chromosome Location
Chr.2: 229763837-229923234 on Build GRCh38
Transcripts
NM_174899.4 XM_011512181.2 XM_011512183.2 XM_017005274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6544025 c.87C>T p.T29T Chr.2: 229922600-229922600 on Build GRCh38
397713 c.30T>C p.F10F Chr.2: 229922543-229922543 on Build GRCh38
5736451 c.88C>T p.R30W Chr.2: 229922601-229922601 on Build GRCh38
4928150 c.55C>G p.P19A Chr.2: 229922568-229922568 on Build GRCh38
3798725 c.94C>G p.Q32E Chr.2: 229922607-229922607 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751578903 Chr.2: 229922656-229922656 on Build GRCh38
rs925350974 Chr.2: 229922651-229922651 on Build GRCh38
rs769932493 Chr.2: 229922522-229922522 on Build GRCh38
rs541179990 Chr.2: 229922459-229922459 on Build GRCh38
rs1027447595 Chr.2: 229922699-229922699 on Build GRCh38
rs914130312 Chr.2: 229922458-229922458 on Build GRCh38
rs1020993043 Chr.2: 229922775-229922775 on Build GRCh38
rs568233783 Chr.2: 229922678-229922678 on Build GRCh38
rs762423299 Chr.2: 229922532-229922532 on Build GRCh38
rs775311980 Chr.2: 229922763-229922763 on Build GRCh38
rs921972194 Chr.2: 229922599-229922599 on Build GRCh38
rs759130535 Chr.2: 229922542-229922542 on Build GRCh38
rs748648208 Chr.2: 229922605-229922605 on Build GRCh38
rs762571859 Chr.2: 229922719-229922719 on Build GRCh38
rs928983905 Chr.2: 229922717-229922717 on Build GRCh38
rs547549631 Chr.2: 229922670-229922670 on Build GRCh38
rs200692739 Chr.2: 229922635-229922635 on Build GRCh38
rs1004605473 Chr.2: 229922870-229922870 on Build GRCh38
rs374860333 Chr.2: 229922731-229922731 on Build GRCh38
rs373396478 Chr.2: 229922856-229922856 on Build GRCh38
rs567237799 Chr.2: 229922841-229922841 on Build GRCh38
rs759193158 Chr.2: 229922646-229922646 on Build GRCh38
rs74794441 Chr.2: 229922533-229922533 on Build GRCh38
rs979837423 Chr.2: 229922765-229922765 on Build GRCh38
rs377607027 Chr.2: 229922493-229922493 on Build GRCh38
rs369209491 Chr.2: 229922676-229922676 on Build GRCh38
rs775037132 Chr.2: 229922482-229922482 on Build GRCh38
rs1014139848 Chr.2: 229922462-229922462 on Build GRCh38
rs749576716 Chr.2: 229922588-229922588 on Build GRCh38
rs1014607036 Chr.2: 229922871-229922871 on Build GRCh38
rs763909393 Chr.2: 229922655-229922655 on Build GRCh38
rs866074670 Chr.2: 229922770-229922770 on Build GRCh38
rs993101380 Chr.2: 229922543-229922543 on Build GRCh38
rs1002682409 Chr.2: 229922754-229922754 on Build GRCh38
rs952250104 Chr.2: 229922518-229922518 on Build GRCh38
rs935409722 Chr.2: 229922777-229922777 on Build GRCh38
rs949852917 Chr.2: 229922808-229922808 on Build GRCh38
rs951802902 Chr.2: 229922704-229922704 on Build GRCh38
rs968362316 Chr.2: 229922665-229922665 on Build GRCh38
rs569773423 Chr.2: 229922869-229922869 on Build GRCh38
rs190804056 Chr.2: 229922882-229922882 on Build GRCh38
rs759036863 Chr.2: 229922553-229922553 on Build GRCh38
rs753365906 Chr.2: 229922509-229922509 on Build GRCh38
rs1035072124 Chr.2: 229922636-229922636 on Build GRCh38
rs1046043315 Chr.2: 229922662-229922662 on Build GRCh38
rs772894444 Chr.2: 229922616-229922616 on Build GRCh38
rs111793003 Chr.2: 229922877-229922877 on Build GRCh38
rs773476398 Chr.2: 229922534-229922534 on Build GRCh38
rs935858764 Chr.2: 229922654-229922654 on Build GRCh38
rs769504180 Chr.2: 229922581-229922581 on Build GRCh38
rs976187855 Chr.2: 229922839-229922839 on Build GRCh38
rs946877122 Chr.2: 229922486-229922486 on Build GRCh38
rs975822845 Chr.2: 229922680-229922680 on Build GRCh38
rs376553274 Chr.2: 229922620-229922620 on Build GRCh38
rs910750924 Chr.2: 229922675-229922675 on Build GRCh38
rs754042543 Chr.2: 229922661-229922661 on Build GRCh38
rs1052496104 Chr.2: 229922785-229922785 on Build GRCh38
rs778820320 Chr.2: 229922514-229922514 on Build GRCh38
rs758158081 Chr.2: 229922519-229922519 on Build GRCh38
rs922162690 Chr.2: 229922760-229922760 on Build GRCh38
rs779134417 Chr.2: 229922601-229922601 on Build GRCh38
rs376734071 Chr.2: 229922660-229922660 on Build GRCh38
rs749345823 Chr.2: 229922524-229922524 on Build GRCh38
rs754525648 Chr.2: 229922510-229922510 on Build GRCh38
rs1000017238 Chr.2: 229922449-229922449 on Build GRCh38
rs370957305 Chr.2: 229922523-229922523 on Build GRCh38
rs1006544293 Chr.2: 229922559-229922559 on Build GRCh38
rs904378190 Chr.2: 229922439-229922439 on Build GRCh38
rs1049214628 Chr.2: 229922861-229922861 on Build GRCh38
rs773707211 Chr.2: 229922536-229922537 on Build GRCh38
rs1015056856 Chr.2: 229922744-229922744 on Build GRCh38
rs976384369 Chr.2: 229922750-229922750 on Build GRCh38
rs1044690210 Chr.2: 229922749-229922749 on Build GRCh38
rs200357690 Chr.2: 229922516-229922516 on Build GRCh38
rs763506243 Chr.2: 229922463-229922463 on Build GRCh38
rs188152765 Chr.2: 229922854-229922854 on Build GRCh38
rs760244630 Chr.2: 229922508-229922508 on Build GRCh38
rs1045684481 Chr.2: 229922831-229922831 on Build GRCh38
rs913093452 Chr.2: 229922794-229922794 on Build GRCh38
rs966156385 Chr.2: 229922746-229922746 on Build GRCh38
rs775007256 Chr.2: 229922650-229922650 on Build GRCh38
rs554596651 Chr.2: 229922443-229922443 on Build GRCh38
rs762786478 Chr.2: 229922790-229922790 on Build GRCh38
rs750034122 Chr.2: 229922507-229922507 on Build GRCh38
rs964911465 Chr.2: 229922837-229922837 on Build GRCh38
rs533599464 Chr.2: 229922769-229922769 on Build GRCh38
rs767828636 Chr.2: 229922504-229922504 on Build GRCh38
rs755046970 Chr.2: 229922590-229922590 on Build GRCh38
rs998083947 Chr.2: 229922879-229922879 on Build GRCh38
rs925234294 Chr.2: 229922771-229922771 on Build GRCh38
rs765923907 Chr.2: 229922541-229922541 on Build GRCh38
rs776210339 Chr.2: 229922640-229922640 on Build GRCh38
rs577947245 Chr.2: 229922521-229922521 on Build GRCh38
rs770663773 Chr.2: 229922723-229922723 on Build GRCh38
rs753556519 Chr.2: 229922547-229922547 on Build GRCh38
rs1031627035 Chr.2: 229922453-229922453 on Build GRCh38
rs960681938 Chr.2: 229922443-229922443 on Build GRCh38
rs941984579 Chr.2: 229922745-229922745 on Build GRCh38
rs756986680 Chr.2: 229922659-229922659 on Build GRCh38
rs887670235 Chr.2: 229922556-229922556 on Build GRCh38
rs769314957 Chr.2: 229922529-229922529 on Build GRCh38
rs370257979 Chr.2: 229922649-229922649 on Build GRCh38
rs183069679 Chr.2: 229922578-229922578 on Build GRCh38
rs537072453 Chr.2: 229922564-229922564 on Build GRCh38
rs1018250144 Chr.2: 229922820-229922820 on Build GRCh38
rs771344759 Chr.2: 229922637-229922637 on Build GRCh38
rs201264237 Chr.2: 229922618-229922618 on Build GRCh38
rs759116467 Chr.2: 229922584-229922585 on Build GRCh38
rs751292712 Chr.2: 229922580-229922580 on Build GRCh38
rs202219810 Chr.2: 229922538-229922538 on Build GRCh38
rs970105282 Chr.2: 229922489-229922489 on Build GRCh38
rs769265279 Chr.2: 229922476-229922476 on Build GRCh38
rs764027415 Chr.2: 229922865-229922865 on Build GRCh38
rs1039327201 Chr.2: 229922754-229922755 on Build GRCh38

View Full Product Details