Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229996658-229997205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXO36
Entrez Gene ID
130888
Gene Name
F-box protein 36
Gene Aliases
Fbx36
UniGene
Hs.140666
Gene Chromosome Location
Chr.2: 229922491-230013109 on Build GRCh38
Transcripts
NM_174899.4 XM_005246317.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1017560 c.286C>T p.R96W Chr.2: 229996831-229996831 on Build GRCh38
4092222 c.301T>C p.L101L Chr.2: 229996846-229996846 on Build GRCh38
1017557 c.245A>G p.Y82C Chr.2: 229996790-229996790 on Build GRCh38
6662217 c.332T>G p.L111R Chr.2: 229996877-229996877 on Build GRCh38
5904717 c.258G>T p.L86F Chr.2: 229996803-229996803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143439395 Chr.2: 229996790-229996790 on Build GRCh38
rs980188920 Chr.2: 229997062-229997062 on Build GRCh38
rs369585800 Chr.2: 229996749-229996749 on Build GRCh38
rs1041124967 Chr.2: 229996690-229996690 on Build GRCh38
rs145930016 Chr.2: 229996782-229996782 on Build GRCh38
rs111536378 Chr.2: 229996984-229996984 on Build GRCh38
rs963774665 Chr.2: 229997050-229997050 on Build GRCh38
rs908688794 Chr.2: 229996887-229996887 on Build GRCh38
rs1057372884 Chr.2: 229996799-229996799 on Build GRCh38
rs761870007 Chr.2: 229996761-229996761 on Build GRCh38
rs776477646 Chr.2: 229996858-229996858 on Build GRCh38
rs567324905 Chr.2: 229997133-229997133 on Build GRCh38
rs747946927 Chr.2: 229997014-229997014 on Build GRCh38
rs373387898 Chr.2: 229996842-229996842 on Build GRCh38
rs530001418 Chr.2: 229997170-229997171 on Build GRCh38
rs574202659 Chr.2: 229996821-229996821 on Build GRCh38
rs543528603 Chr.2: 229997145-229997145 on Build GRCh38
rs564682833 Chr.2: 229997075-229997075 on Build GRCh38
rs1046264245 Chr.2: 229997175-229997175 on Build GRCh38
rs767954505 Chr.2: 229996980-229996981 on Build GRCh38
rs776504024 Chr.2: 229996783-229996783 on Build GRCh38
rs756788593 Chr.2: 229996963-229996963 on Build GRCh38
rs766599623 Chr.2: 229996805-229996805 on Build GRCh38
rs375109352 Chr.2: 229996746-229996746 on Build GRCh38
rs1035834 Chr.2: 229996803-229996803 on Build GRCh38
rs376803621 Chr.2: 229996962-229996962 on Build GRCh38
rs963447944 Chr.2: 229996708-229996708 on Build GRCh38
rs1050159893 Chr.2: 229997061-229997061 on Build GRCh38
rs755336934 Chr.2: 229996822-229996822 on Build GRCh38
rs1049793522 Chr.2: 229996775-229996775 on Build GRCh38
rs774625090 Chr.2: 229996750-229996750 on Build GRCh38
rs922675844 Chr.2: 229997042-229997042 on Build GRCh38
rs761633689 Chr.2: 229996812-229996812 on Build GRCh38
rs1025479268 Chr.2: 229996982-229996982 on Build GRCh38
rs191623828 Chr.2: 229997134-229997134 on Build GRCh38
rs530953457 Chr.2: 229996729-229996729 on Build GRCh38
rs570572146 Chr.2: 229996736-229996736 on Build GRCh38
rs144211805 Chr.2: 229997010-229997010 on Build GRCh38
rs757917554 Chr.2: 229996836-229996836 on Build GRCh38
rs760713596 Chr.2: 229996915-229996915 on Build GRCh38
rs13382327 Chr.2: 229997149-229997149 on Build GRCh38
rs777618847 Chr.2: 229996955-229996955 on Build GRCh38
rs984339155 Chr.2: 229996885-229996885 on Build GRCh38
rs773267242 Chr.2: 229996702-229996702 on Build GRCh38
rs770592982 Chr.2: 229996716-229996716 on Build GRCh38
rs746894162 Chr.2: 229996843-229996843 on Build GRCh38
rs140479647 Chr.2: 229996831-229996831 on Build GRCh38
rs1005794811 Chr.2: 229997027-229997027 on Build GRCh38
rs959996053 Chr.2: 229997126-229997126 on Build GRCh38
rs773298727 Chr.2: 229996904-229996904 on Build GRCh38
rs983561490 Chr.2: 229996697-229996697 on Build GRCh38
rs201098656 Chr.2: 229996762-229996762 on Build GRCh38
rs909247457 Chr.2: 229996699-229996699 on Build GRCh38
rs781178212 Chr.2: 229996971-229996971 on Build GRCh38
rs1009383268 Chr.2: 229997165-229997165 on Build GRCh38
rs1039324823 Chr.2: 229996893-229996893 on Build GRCh38
rs769538225 Chr.2: 229996886-229996886 on Build GRCh38
rs745676136 Chr.2: 229996879-229996879 on Build GRCh38
rs747276723 Chr.2: 229996870-229996870 on Build GRCh38
rs140140630 Chr.2: 229996898-229996898 on Build GRCh38
rs752358341 Chr.2: 229996942-229996942 on Build GRCh38
rs776507892 Chr.2: 229996723-229996723 on Build GRCh38
rs1033384470 Chr.2: 229996848-229996848 on Build GRCh38
rs936551007 Chr.2: 229997127-229997127 on Build GRCh38
rs770879889 Chr.2: 229996852-229996852 on Build GRCh38
rs970524734 Chr.2: 229996834-229996834 on Build GRCh38
rs758148675 Chr.2: 229996948-229996948 on Build GRCh38
rs562328679 Chr.2: 229996715-229996715 on Build GRCh38
rs1006587213 Chr.2: 229996884-229996884 on Build GRCh38
rs774502195 Chr.2: 229996896-229996896 on Build GRCh38
rs765708957 Chr.2: 229996931-229996931 on Build GRCh38
rs1043605658 Chr.2: 229997174-229997174 on Build GRCh38
rs754629116 Chr.2: 229996882-229996882 on Build GRCh38
rs747866198 Chr.2: 229996832-229996832 on Build GRCh38
rs766877980 Chr.2: 229996918-229996918 on Build GRCh38
rs187205263 Chr.2: 229996928-229996928 on Build GRCh38
rs758196179 Chr.2: 229997049-229997049 on Build GRCh38
rs754369086 Chr.2: 229996925-229996925 on Build GRCh38
rs112462272 Chr.2: 229997181-229997181 on Build GRCh38
rs925980510 Chr.2: 229997072-229997072 on Build GRCh38

View Full Product Details