Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230907632-230908125 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR55
Entrez Gene ID
9290
Gene Name
G protein-coupled receptor 55
Gene Aliases
LPIR1
UniGene
Hs.114545
Gene Chromosome Location
Chr.2: 230907318-230961269 on Build GRCh38
Transcripts
NM_005683.3 XM_005246952.3 XM_011512175.2 XM_011512176.2

SNP Detail

SNP ID Chromosome Location
rs937532601 Chr.2: 230908066-230908066 on Build GRCh38
rs762704732 Chr.2: 230908055-230908056 on Build GRCh38
rs765897643 Chr.2: 230908075-230908075 on Build GRCh38
rs925962541 Chr.2: 230907692-230907692 on Build GRCh38
rs1045894416 Chr.2: 230907723-230907723 on Build GRCh38
rs1992188 Chr.2: 230907721-230907721 on Build GRCh38
rs1030809515 Chr.2: 230908091-230908091 on Build GRCh38
rs866256210 Chr.2: 230907834-230907834 on Build GRCh38
rs913520102 Chr.2: 230907712-230907712 on Build GRCh38
rs1028753703 Chr.2: 230907895-230907895 on Build GRCh38
rs760038173 Chr.2: 230907963-230907963 on Build GRCh38
rs953241895 Chr.2: 230907898-230907898 on Build GRCh38
rs192702221 Chr.2: 230907873-230907873 on Build GRCh38
rs547254648 Chr.2: 230907762-230907762 on Build GRCh38
rs906049610 Chr.2: 230907737-230907737 on Build GRCh38
rs570646845 Chr.2: 230908000-230908000 on Build GRCh38
rs1049314948 Chr.2: 230907741-230907741 on Build GRCh38
rs184053115 Chr.2: 230907955-230907955 on Build GRCh38
rs1004987241 Chr.2: 230907789-230907789 on Build GRCh38
rs989074152 Chr.2: 230907701-230907701 on Build GRCh38
rs566666602 Chr.2: 230907982-230907982 on Build GRCh38
rs772830866 Chr.2: 230907884-230907884 on Build GRCh38
rs957385762 Chr.2: 230907974-230907974 on Build GRCh38
rs985544356 Chr.2: 230907931-230907931 on Build GRCh38
rs940793856 Chr.2: 230907918-230907918 on Build GRCh38
rs537714669 Chr.2: 230908052-230908052 on Build GRCh38
rs916540245 Chr.2: 230907780-230907780 on Build GRCh38
rs56241531 Chr.2: 230907907-230907907 on Build GRCh38
rs569144644 Chr.2: 230907890-230907890 on Build GRCh38
rs34670968 Chr.2: 230908058-230908059 on Build GRCh38
rs72539509 Chr.2: 230908056-230908057 on Build GRCh38
rs889407347 Chr.2: 230908016-230908016 on Build GRCh38
rs186815970 Chr.2: 230907764-230907764 on Build GRCh38
rs753310353 Chr.2: 230908056-230908056 on Build GRCh38
rs999693712 Chr.2: 230908063-230908063 on Build GRCh38
rs901727271 Chr.2: 230907851-230907851 on Build GRCh38
rs1036503589 Chr.2: 230907935-230907935 on Build GRCh38
rs937079509 Chr.2: 230907738-230907738 on Build GRCh38
rs1022047417 Chr.2: 230907660-230907660 on Build GRCh38
rs200477178 Chr.2: 230908056-230908056 on Build GRCh38
rs767231190 Chr.2: 230907801-230907801 on Build GRCh38
rs1041672551 Chr.2: 230907717-230907726 on Build GRCh38
rs560315167 Chr.2: 230907700-230907700 on Build GRCh38
rs969368370 Chr.2: 230907833-230907833 on Build GRCh38
rs534038759 Chr.2: 230907993-230907993 on Build GRCh38
rs985961535 Chr.2: 230907690-230907690 on Build GRCh38
rs558283223 Chr.2: 230907996-230907996 on Build GRCh38
rs11414001 Chr.2: 230908057-230908058 on Build GRCh38
rs984790541 Chr.2: 230907894-230907894 on Build GRCh38
rs1006561650 Chr.2: 230908046-230908046 on Build GRCh38
rs913239034 Chr.2: 230907995-230907995 on Build GRCh38
rs532668838 Chr.2: 230907790-230907790 on Build GRCh38
rs1033550097 Chr.2: 230907932-230907932 on Build GRCh38
rs35856899 Chr.2: 230907793-230907794 on Build GRCh38
rs536679596 Chr.2: 230907917-230907917 on Build GRCh38

View Full Product Details