Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230909407-230909916 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR55
Entrez Gene ID
9290
Gene Name
G protein-coupled receptor 55
Gene Aliases
LPIR1
UniGene
Hs.114545
Gene Chromosome Location
Chr.2: 230907318-230961269 on Build GRCh38
Transcripts
NM_005683.3 XM_005246952.3 XM_011512175.2 XM_011512176.2

SNP Detail

SNP ID Chromosome Location
rs759195061 Chr.2: 230909886-230909886 on Build GRCh38
rs1020751774 Chr.2: 230909479-230909479 on Build GRCh38
rs987396757 Chr.2: 230909654-230909654 on Build GRCh38
rs376644032 Chr.2: 230909724-230909724 on Build GRCh38
rs962890362 Chr.2: 230909600-230909600 on Build GRCh38
rs1025438303 Chr.2: 230909846-230909846 on Build GRCh38
rs972499809 Chr.2: 230909840-230909840 on Build GRCh38
rs950772111 Chr.2: 230909848-230909848 on Build GRCh38
rs762509299 Chr.2: 230909604-230909604 on Build GRCh38
rs372746444 Chr.2: 230909441-230909441 on Build GRCh38
rs111573399 Chr.2: 230909524-230909524 on Build GRCh38
rs1008230920 Chr.2: 230909430-230909430 on Build GRCh38
rs943618174 Chr.2: 230909677-230909677 on Build GRCh38
rs919379878 Chr.2: 230909608-230909608 on Build GRCh38
rs62195307 Chr.2: 230909565-230909565 on Build GRCh38
rs772849350 Chr.2: 230909561-230909561 on Build GRCh38
rs1045854356 Chr.2: 230909663-230909663 on Build GRCh38
rs770274564 Chr.2: 230909553-230909554 on Build GRCh38
rs184410278 Chr.2: 230909793-230909793 on Build GRCh38
rs192956440 Chr.2: 230909738-230909738 on Build GRCh38
rs1012151681 Chr.2: 230909792-230909792 on Build GRCh38
rs1039172005 Chr.2: 230909431-230909431 on Build GRCh38
rs894827112 Chr.2: 230909777-230909777 on Build GRCh38
rs758975790 Chr.2: 230909771-230909771 on Build GRCh38
rs991163129 Chr.2: 230909541-230909541 on Build GRCh38
rs1010644127 Chr.2: 230909455-230909455 on Build GRCh38
rs911887128 Chr.2: 230909882-230909882 on Build GRCh38
rs987532511 Chr.2: 230909878-230909878 on Build GRCh38
rs370782609 Chr.2: 230909761-230909761 on Build GRCh38
rs1016748463 Chr.2: 230909555-230909555 on Build GRCh38
rs1042583100 Chr.2: 230909749-230909749 on Build GRCh38
rs1001709688 Chr.2: 230909736-230909736 on Build GRCh38
rs959290626 Chr.2: 230909536-230909536 on Build GRCh38
rs113799481 Chr.2: 230909692-230909692 on Build GRCh38
rs139821600 Chr.2: 230909713-230909714 on Build GRCh38
rs962570294 Chr.2: 230909806-230909806 on Build GRCh38
rs201637205 Chr.2: 230909716-230909716 on Build GRCh38
rs1003171920 Chr.2: 230909517-230909517 on Build GRCh38
rs902179310 Chr.2: 230909490-230909490 on Build GRCh38
rs769286187 Chr.2: 230909774-230909774 on Build GRCh38
rs902231622 Chr.2: 230909760-230909760 on Build GRCh38
rs143899095 Chr.2: 230909602-230909602 on Build GRCh38
rs929393372 Chr.2: 230909631-230909631 on Build GRCh38
rs560441276 Chr.2: 230909651-230909651 on Build GRCh38
rs994493131 Chr.2: 230909787-230909787 on Build GRCh38
rs891866656 Chr.2: 230909628-230909629 on Build GRCh38
rs10201627 Chr.2: 230909662-230909662 on Build GRCh38
rs890666443 Chr.2: 230909429-230909429 on Build GRCh38
rs893992121 Chr.2: 230909453-230909453 on Build GRCh38
rs1039723971 Chr.2: 230909690-230909690 on Build GRCh38
rs946916272 Chr.2: 230909747-230909747 on Build GRCh38
rs199972477 Chr.2: 230909718-230909718 on Build GRCh38
rs987288305 Chr.2: 230909481-230909481 on Build GRCh38

View Full Product Details