Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 230910994-230911490 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR55
Entrez Gene ID
9290
Gene Name
G protein-coupled receptor 55
Gene Aliases
LPIR1
UniGene
Hs.114545
Gene Chromosome Location
Chr.2: 230907318-230961269 on Build GRCh38
Transcripts
NM_005683.3 XM_005246952.3 XM_011512175.2 XM_011512176.2

SNP Detail

SNP ID Chromosome Location
rs899656030 Chr.2: 230911096-230911096 on Build GRCh38
rs146480493 Chr.2: 230911182-230911182 on Build GRCh38
rs766737684 Chr.2: 230911238-230911238 on Build GRCh38
rs760946910 Chr.2: 230911069-230911069 on Build GRCh38
rs62195308 Chr.2: 230911410-230911410 on Build GRCh38
rs945811556 Chr.2: 230911361-230911361 on Build GRCh38
rs907047356 Chr.2: 230911426-230911426 on Build GRCh38
rs756124310 Chr.2: 230911325-230911326 on Build GRCh38
rs753137210 Chr.2: 230911444-230911444 on Build GRCh38
rs1027124328 Chr.2: 230911309-230911309 on Build GRCh38
rs144115429 Chr.2: 230911103-230911103 on Build GRCh38
rs947280729 Chr.2: 230911111-230911111 on Build GRCh38
rs999622584 Chr.2: 230911166-230911166 on Build GRCh38
rs111311729 Chr.2: 230911313-230911313 on Build GRCh38
rs1034273350 Chr.2: 230911118-230911118 on Build GRCh38
rs943752921 Chr.2: 230911373-230911373 on Build GRCh38
rs982919966 Chr.2: 230911194-230911194 on Build GRCh38
rs191719070 Chr.2: 230911057-230911057 on Build GRCh38
rs975671606 Chr.2: 230911302-230911302 on Build GRCh38
rs758998694 Chr.2: 230911458-230911458 on Build GRCh38
rs912754979 Chr.2: 230911232-230911232 on Build GRCh38
rs1039653263 Chr.2: 230911082-230911082 on Build GRCh38
rs755026150 Chr.2: 230911316-230911316 on Build GRCh38
rs1023931220 Chr.2: 230911244-230911244 on Build GRCh38
rs548525977 Chr.2: 230911100-230911100 on Build GRCh38
rs1042912082 Chr.2: 230911124-230911124 on Build GRCh38
rs563480834 Chr.2: 230911046-230911046 on Build GRCh38
rs114869253 Chr.2: 230911129-230911129 on Build GRCh38
rs912217111 Chr.2: 230911333-230911333 on Build GRCh38
rs781427551 Chr.2: 230911324-230911324 on Build GRCh38
rs896506854 Chr.2: 230911203-230911203 on Build GRCh38
rs753918942 Chr.2: 230911286-230911286 on Build GRCh38
rs974135241 Chr.2: 230911144-230911144 on Build GRCh38
rs974235354 Chr.2: 230911294-230911294 on Build GRCh38

View Full Product Details