Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233161641-233162137 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5D
Entrez Gene ID
3635
Gene Name
inositol polyphosphate-5-phosphatase D
Gene Aliases
SHIP, SHIP-1, SHIP1, SIP-145, hp51CN, p150Ship
UniGene
Hs.262886
Gene Chromosome Location
Chr.2: 233059967-233207903 on Build GRCh38
Transcripts
NM_001017915.2 NM_005541.4 XM_017004004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5588055 c.1224C>T p.I408I Chr.2: 233161810-233161810 on Build GRCh38
1285544 c.1192C>A p.Q398K Chr.2: 233161778-233161778 on Build GRCh38
3722169 c.1196C>T p.P399L Chr.2: 233161782-233161782 on Build GRCh38
1406339 c.1173_1175delGAA p.K392delK Chr.2: 233161759-233161761 on Build GRCh38
4692598 c.1197G>A p.P399P Chr.2: 233161783-233161783 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046151467 Chr.2: 233161902-233161902 on Build GRCh38
rs553129476 Chr.2: 233161691-233161691 on Build GRCh38
rs990465416 Chr.2: 233161917-233161917 on Build GRCh38
rs755603032 Chr.2: 233162028-233162028 on Build GRCh38
rs952940596 Chr.2: 233161921-233161921 on Build GRCh38
rs902709692 Chr.2: 233162044-233162044 on Build GRCh38
rs146190387 Chr.2: 233162034-233162034 on Build GRCh38
rs756793738 Chr.2: 233161858-233161858 on Build GRCh38
rs1026629175 Chr.2: 233161920-233161920 on Build GRCh38
rs894912526 Chr.2: 233162084-233162084 on Build GRCh38
rs751116963 Chr.2: 233161857-233161857 on Build GRCh38
rs752372116 Chr.2: 233161753-233161753 on Build GRCh38
rs976870344 Chr.2: 233161808-233161808 on Build GRCh38
rs751418147 Chr.2: 233161911-233161911 on Build GRCh38
rs1004433181 Chr.2: 233161923-233161923 on Build GRCh38
rs777854512 Chr.2: 233161978-233161978 on Build GRCh38
rs1001925730 Chr.2: 233162051-233162051 on Build GRCh38
rs779559227 Chr.2: 233162029-233162029 on Build GRCh38
rs766304946 Chr.2: 233161840-233161840 on Build GRCh38
rs766438873 Chr.2: 233161683-233161683 on Build GRCh38
rs758115314 Chr.2: 233161847-233161847 on Build GRCh38
rs1013720298 Chr.2: 233161894-233161894 on Build GRCh38
rs753029961 Chr.2: 233161846-233161846 on Build GRCh38
rs375564765 Chr.2: 233161934-233161934 on Build GRCh38
rs780526930 Chr.2: 233161868-233161868 on Build GRCh38
rs1052360799 Chr.2: 233162019-233162019 on Build GRCh38
rs770834125 Chr.2: 233161787-233161787 on Build GRCh38
rs376435860 Chr.2: 233161837-233161837 on Build GRCh38
rs763790732 Chr.2: 233161851-233161851 on Build GRCh38
rs28751960 Chr.2: 233161941-233161941 on Build GRCh38
rs769741057 Chr.2: 233161822-233161822 on Build GRCh38
rs755166057 Chr.2: 233161789-233161789 on Build GRCh38
rs904982767 Chr.2: 233161912-233161912 on Build GRCh38
rs754211352 Chr.2: 233161692-233161692 on Build GRCh38
rs530746394 Chr.2: 233161839-233161839 on Build GRCh38
rs550713961 Chr.2: 233162065-233162065 on Build GRCh38
rs188454720 Chr.2: 233161971-233161971 on Build GRCh38
rs534405290 Chr.2: 233161810-233161810 on Build GRCh38
rs752976341 Chr.2: 233161901-233161901 on Build GRCh38
rs922698045 Chr.2: 233161998-233161998 on Build GRCh38
rs367936476 Chr.2: 233161948-233161948 on Build GRCh38
rs777189163 Chr.2: 233161782-233161782 on Build GRCh38
rs894074406 Chr.2: 233161682-233161682 on Build GRCh38
rs893772428 Chr.2: 233162023-233162023 on Build GRCh38
rs765643124 Chr.2: 233161843-233161843 on Build GRCh38
rs563567520 Chr.2: 233161909-233161909 on Build GRCh38
rs1015679986 Chr.2: 233161926-233161926 on Build GRCh38
rs982892853 Chr.2: 233161714-233161714 on Build GRCh38
rs373235759 Chr.2: 233161936-233161936 on Build GRCh38
rs1048019957 Chr.2: 233162078-233162078 on Build GRCh38
rs376324528 Chr.2: 233161783-233161783 on Build GRCh38
rs28476748 Chr.2: 233161838-233161838 on Build GRCh38
rs755342582 Chr.2: 233161696-233161696 on Build GRCh38
rs565379792 Chr.2: 233162015-233162015 on Build GRCh38
rs983350252 Chr.2: 233161931-233161931 on Build GRCh38
rs1001861653 Chr.2: 233161913-233161913 on Build GRCh38
rs573135762 Chr.2: 233161706-233161706 on Build GRCh38
rs1003696435 Chr.2: 233162047-233162047 on Build GRCh38
rs977364378 Chr.2: 233161970-233161970 on Build GRCh38
rs28437902 Chr.2: 233162003-233162003 on Build GRCh38
rs1011918847 Chr.2: 233162092-233162092 on Build GRCh38
rs748898463 Chr.2: 233162045-233162045 on Build GRCh38
rs1046696452 Chr.2: 233162031-233162031 on Build GRCh38
rs866285497 Chr.2: 233162027-233162027 on Build GRCh38
rs867948305 Chr.2: 233161778-233161778 on Build GRCh38
rs774715461 Chr.2: 233161834-233161835 on Build GRCh38
rs543063428 Chr.2: 233161885-233161885 on Build GRCh38
rs758023759 Chr.2: 233161774-233161774 on Build GRCh38
rs759991291 Chr.2: 233161842-233161842 on Build GRCh38
rs551900778 Chr.2: 233162013-233162013 on Build GRCh38
rs557479821 Chr.2: 233161836-233161836 on Build GRCh38
rs775375341 Chr.2: 233161834-233161834 on Build GRCh38

View Full Product Details