Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 233273639-233274144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG16L1
Entrez Gene ID
55054
Gene Name
autophagy related 16 like 1
Gene Aliases
APG16L, ATG16A, ATG16L, IBD10, WDR30
UniGene
Hs.529322
Gene Chromosome Location
Chr.2: 233251571-233295674 on Build GRCh38
Transcripts
NM_001190266.1 NM_001190267.1 NM_017974.3 NM_030803.6 NM_198890.2 XM_005246082.2 XM_005246084.2 XM_005246086.2 XM_006712608.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6010717 c.846C>A p.I282I Chr.2: 233273772-233273772 on Build GRCh38
3379253 c.833C>T p.S278F Chr.2: 233273759-233273759 on Build GRCh38
1017965 c.833C>A p.S278Y Chr.2: 233273759-233273759 on Build GRCh38
6328331 c.815C>T p.A272V Chr.2: 233273741-233273741 on Build GRCh38
5354874 c.842A>C p.N281T Chr.2: 233273768-233273768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1046456818 Chr.2: 233274060-233274060 on Build GRCh38
rs781316125 Chr.2: 233273672-233273672 on Build GRCh38
rs990777632 Chr.2: 233273789-233273789 on Build GRCh38
rs769075316 Chr.2: 233273764-233273764 on Build GRCh38
rs890204720 Chr.2: 233273901-233273901 on Build GRCh38
rs777534666 Chr.2: 233274070-233274070 on Build GRCh38
rs1046949505 Chr.2: 233273835-233273835 on Build GRCh38
rs960759541 Chr.2: 233274017-233274017 on Build GRCh38
rs553042730 Chr.2: 233273960-233273960 on Build GRCh38
rs530145512 Chr.2: 233274097-233274097 on Build GRCh38
rs150010037 Chr.2: 233274107-233274107 on Build GRCh38
rs768985323 Chr.2: 233273694-233273694 on Build GRCh38
rs1048030441 Chr.2: 233273887-233273887 on Build GRCh38
rs372958037 Chr.2: 233273710-233273710 on Build GRCh38
rs748850556 Chr.2: 233273977-233273978 on Build GRCh38
rs758460489 Chr.2: 233273671-233273671 on Build GRCh38
rs140476598 Chr.2: 233274083-233274083 on Build GRCh38
rs771541752 Chr.2: 233273708-233273708 on Build GRCh38
rs780004910 Chr.2: 233274019-233274019 on Build GRCh38
rs960809787 Chr.2: 233273769-233273769 on Build GRCh38
rs1054905100 Chr.2: 233274096-233274096 on Build GRCh38
rs1023688049 Chr.2: 233274024-233274024 on Build GRCh38
rs944481132 Chr.2: 233273903-233273903 on Build GRCh38
rs750544059 Chr.2: 233273736-233273736 on Build GRCh38
rs538445839 Chr.2: 233273811-233273811 on Build GRCh38
rs1002014188 Chr.2: 233273868-233273868 on Build GRCh38
rs780587327 Chr.2: 233273755-233273755 on Build GRCh38
rs770342140 Chr.2: 233273810-233273811 on Build GRCh38
rs753782891 Chr.2: 233273718-233273718 on Build GRCh38
rs929115586 Chr.2: 233273870-233273870 on Build GRCh38
rs759544513 Chr.2: 233273818-233273818 on Build GRCh38
rs757511801 Chr.2: 233273732-233273732 on Build GRCh38
rs928034559 Chr.2: 233273748-233273748 on Build GRCh38
rs565076810 Chr.2: 233274033-233274033 on Build GRCh38
rs755491454 Chr.2: 233273999-233273999 on Build GRCh38
rs1038703385 Chr.2: 233273906-233273906 on Build GRCh38
rs780101797 Chr.2: 233273979-233273979 on Build GRCh38
rs983304309 Chr.2: 233274067-233274067 on Build GRCh38
rs974484114 Chr.2: 233274044-233274044 on Build GRCh38
rs558116998 Chr.2: 233273781-233273781 on Build GRCh38
rs569382385 Chr.2: 233273692-233273692 on Build GRCh38
rs920527326 Chr.2: 233273858-233273858 on Build GRCh38
rs761524765 Chr.2: 233273707-233273707 on Build GRCh38
rs772701483 Chr.2: 233273810-233273810 on Build GRCh38
rs779324948 Chr.2: 233273768-233273768 on Build GRCh38
rs201424939 Chr.2: 233273827-233273827 on Build GRCh38
rs950486407 Chr.2: 233273959-233273959 on Build GRCh38
rs771760776 Chr.2: 233273786-233273786 on Build GRCh38
rs776751904 Chr.2: 233273817-233273817 on Build GRCh38
rs1054955344 Chr.2: 233273900-233273900 on Build GRCh38
rs767802062 Chr.2: 233273963-233273963 on Build GRCh38
rs756137498 Chr.2: 233273742-233273742 on Build GRCh38
rs528429008 Chr.2: 233273964-233273964 on Build GRCh38
rs548086211 Chr.2: 233274065-233274065 on Build GRCh38
rs892505480 Chr.2: 233273776-233273776 on Build GRCh38
rs773913752 Chr.2: 233273705-233273705 on Build GRCh38
rs770635082 Chr.2: 233274089-233274089 on Build GRCh38
rs530757301 Chr.2: 233274108-233274108 on Build GRCh38
rs1048142364 Chr.2: 233274108-233274109 on Build GRCh38
rs528334979 Chr.2: 233273851-233273851 on Build GRCh38
rs748578560 Chr.2: 233274020-233274020 on Build GRCh38
rs777956010 Chr.2: 233273733-233273733 on Build GRCh38
rs545003258 Chr.2: 233273997-233273997 on Build GRCh38
rs921491114 Chr.2: 233273988-233273988 on Build GRCh38
rs962663597 Chr.2: 233274034-233274034 on Build GRCh38
rs994441763 Chr.2: 233273904-233273904 on Build GRCh38
rs770286194 Chr.2: 233273683-233273683 on Build GRCh38
rs759456183 Chr.2: 233273724-233273724 on Build GRCh38
rs770547236 Chr.2: 233273816-233273816 on Build GRCh38
rs551897739 Chr.2: 233274021-233274023 on Build GRCh38
rs765096319 Chr.2: 233273726-233273726 on Build GRCh38
rs1010593533 Chr.2: 233273934-233273934 on Build GRCh38
rs201415775 Chr.2: 233273730-233273730 on Build GRCh38
rs530585966 Chr.2: 233274048-233274048 on Build GRCh38
rs965556747 Chr.2: 233273968-233273970 on Build GRCh38
rs763416099 Chr.2: 233273761-233273761 on Build GRCh38
rs989446071 Chr.2: 233274014-233274014 on Build GRCh38
rs202032029 Chr.2: 233273818-233273819 on Build GRCh38
rs199558257 Chr.2: 233273689-233273689 on Build GRCh38
rs748405935 Chr.2: 233273772-233273772 on Build GRCh38
rs542604072 Chr.2: 233273958-233273958 on Build GRCh38
rs973315558 Chr.2: 233273843-233273843 on Build GRCh38
rs114509633 Chr.2: 233273972-233273972 on Build GRCh38
rs958279583 Chr.2: 233273994-233273994 on Build GRCh38
rs573781731 Chr.2: 233273921-233273921 on Build GRCh38
rs745873956 Chr.2: 233273676-233273676 on Build GRCh38

View Full Product Details