Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 241558464-241558962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BOK
Entrez Gene ID
666
Gene Name
BOK, BCL2 family apoptosis regulator
Gene Aliases
BCL2L9, BOKL
UniGene
Hs.293753
Gene Chromosome Location
Chr.2: 241558731-241574138 on Build GRCh38
Transcripts
NM_032515.4 XM_011511697.2 XM_017004775.1 NR_033346.1

Gene Symbol
BOK-AS1
Entrez Gene ID
100379249
Gene Name
BOK antisense RNA 1
Gene Aliases
BOK-AS, BOKAS, NAToB, NCRNA00151
UniGene
Hs.621128
Gene Chromosome Location
Chr.2: 241544384-241559143 on Build GRCh38
Transcripts
NM_032515.4 XM_011511697.2 XM_017004775.1 NR_033346.1

SNP Detail

SNP ID Chromosome Location
rs931117336 Chr.2: 241558930-241558930 on Build GRCh38
rs143686179 Chr.2: 241558519-241558519 on Build GRCh38
rs909064975 Chr.2: 241558637-241558637 on Build GRCh38
rs999083861 Chr.2: 241558529-241558529 on Build GRCh38
rs940476222 Chr.2: 241558653-241558653 on Build GRCh38
rs533582873 Chr.2: 241558920-241558920 on Build GRCh38
rs965359870 Chr.2: 241558600-241558600 on Build GRCh38
rs1011347125 Chr.2: 241558724-241558724 on Build GRCh38
rs971929748 Chr.2: 241558818-241558818 on Build GRCh38
rs986329938 Chr.2: 241558494-241558494 on Build GRCh38
rs1009357948 Chr.2: 241558875-241558875 on Build GRCh38
rs370846660 Chr.2: 241558603-241558603 on Build GRCh38
rs894118289 Chr.2: 241558705-241558705 on Build GRCh38
rs939908990 Chr.2: 241558878-241558878 on Build GRCh38
rs947014750 Chr.2: 241558893-241558893 on Build GRCh38
rs935762577 Chr.2: 241558546-241558546 on Build GRCh38
rs921261285 Chr.2: 241558612-241558612 on Build GRCh38
rs112259601 Chr.2: 241558850-241558850 on Build GRCh38
rs961875254 Chr.2: 241558816-241558816 on Build GRCh38
rs1042821656 Chr.2: 241558900-241558900 on Build GRCh38
rs551879321 Chr.2: 241558514-241558516 on Build GRCh38
rs902904596 Chr.2: 241558905-241558905 on Build GRCh38
rs1045437201 Chr.2: 241558610-241558610 on Build GRCh38
rs948364426 Chr.2: 241558587-241558587 on Build GRCh38
rs998948063 Chr.2: 241558743-241558743 on Build GRCh38
rs867149388 Chr.2: 241558676-241558676 on Build GRCh38
rs1036981808 Chr.2: 241558888-241558888 on Build GRCh38
rs567946465 Chr.2: 241558810-241558811 on Build GRCh38
rs569130568 Chr.2: 241558751-241558751 on Build GRCh38
rs1054641440 Chr.2: 241558568-241558568 on Build GRCh38
rs1028354893 Chr.2: 241558785-241558785 on Build GRCh38
rs928322731 Chr.2: 241558873-241558873 on Build GRCh38
rs1021821066 Chr.2: 241558507-241558507 on Build GRCh38
rs991795549 Chr.2: 241558863-241558863 on Build GRCh38
rs984599815 Chr.2: 241558633-241558633 on Build GRCh38
rs997500149 Chr.2: 241558759-241558759 on Build GRCh38
rs898312597 Chr.2: 241558891-241558891 on Build GRCh38
rs1018909973 Chr.2: 241558783-241558783 on Build GRCh38
rs994920510 Chr.2: 241558720-241558720 on Build GRCh38
rs1019829916 Chr.2: 241558747-241558747 on Build GRCh38
rs981063178 Chr.2: 241558868-241558868 on Build GRCh38
rs56390343 Chr.2: 241558484-241558484 on Build GRCh38
rs201711208 Chr.2: 241558826-241558836 on Build GRCh38
rs1007487016 Chr.2: 241558779-241558779 on Build GRCh38
rs1016059523 Chr.2: 241558815-241558815 on Build GRCh38
rs539603349 Chr.2: 241558757-241558757 on Build GRCh38
rs186908585 Chr.2: 241558805-241558805 on Build GRCh38
rs529372360 Chr.2: 241558662-241558662 on Build GRCh38
rs915595774 Chr.2: 241558880-241558880 on Build GRCh38
rs915561248 Chr.2: 241558576-241558576 on Build GRCh38
rs966227993 Chr.2: 241558800-241558800 on Build GRCh38
rs938606482 Chr.2: 241558861-241558861 on Build GRCh38
rs546967405 Chr.2: 241558570-241558570 on Build GRCh38
rs1019797970 Chr.2: 241558538-241558538 on Build GRCh38
rs550626322 Chr.2: 241558680-241558680 on Build GRCh38
rs1036860644 Chr.2: 241558669-241558669 on Build GRCh38
rs375404902 Chr.2: 241558860-241558860 on Build GRCh38
rs952860228 Chr.2: 241558630-241558630 on Build GRCh38
rs573525303 Chr.2: 241558933-241558933 on Build GRCh38
rs903376051 Chr.2: 241558740-241558740 on Build GRCh38
rs1049597047 Chr.2: 241558931-241558931 on Build GRCh38
rs1043177399 Chr.2: 241558738-241558738 on Build GRCh38
rs1011733002 Chr.2: 241558500-241558500 on Build GRCh38
rs1042512222 Chr.2: 241558883-241558883 on Build GRCh38
rs544348055 Chr.2: 241558938-241558938 on Build GRCh38
rs898359364 Chr.2: 241558708-241558708 on Build GRCh38
rs952804913 Chr.2: 241558795-241558795 on Build GRCh38
rs966280599 Chr.2: 241558527-241558527 on Build GRCh38
rs760099315 Chr.2: 241558496-241558496 on Build GRCh38
rs555328312 Chr.2: 241558932-241558932 on Build GRCh38
rs568487520 Chr.2: 241558661-241558661 on Build GRCh38
rs901473533 Chr.2: 241558755-241558755 on Build GRCh38
rs975347379 Chr.2: 241558611-241558611 on Build GRCh38
rs1055496984 Chr.2: 241558701-241558701 on Build GRCh38

View Full Product Details