Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 289483-289988 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf96
Entrez Gene ID
140680
Gene Name
chromosome 20 open reading frame 96
Gene Aliases
dJ1103G7.2
UniGene
Hs.348112
Gene Chromosome Location
Chr.20: 270863-290778 on Build GRCh38
Transcripts
NM_080571.1 NM_153269.2 XM_006723543.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7400356 c.183A>G p.Q61Q Chr.20: 289563-289563 on Build GRCh38
1025449 c.96G>A p.K32K Chr.20: 289650-289650 on Build GRCh38
6190123 c.113C>T p.S38F Chr.20: 289633-289633 on Build GRCh38
2887930 c.143G>A p.S48N Chr.20: 289603-289603 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144519838 Chr.20: 289508-289508 on Build GRCh38
rs6035784 Chr.20: 289787-289787 on Build GRCh38
rs778657963 Chr.20: 289514-289514 on Build GRCh38
rs753605083 Chr.20: 289687-289687 on Build GRCh38
rs189947916 Chr.20: 289710-289710 on Build GRCh38
rs1018969384 Chr.20: 289677-289677 on Build GRCh38
rs753735089 Chr.20: 289599-289599 on Build GRCh38
rs879111239 Chr.20: 289530-289530 on Build GRCh38
rs1011898879 Chr.20: 289685-289685 on Build GRCh38
rs77300649 Chr.20: 289809-289809 on Build GRCh38
rs924529492 Chr.20: 289740-289740 on Build GRCh38
rs774711308 Chr.20: 289575-289575 on Build GRCh38
rs796326308 Chr.20: 289775-289775 on Build GRCh38
rs1045792316 Chr.20: 289800-289800 on Build GRCh38
rs746321638 Chr.20: 289675-289675 on Build GRCh38
rs372566973 Chr.20: 289634-289634 on Build GRCh38
rs375746621 Chr.20: 289576-289576 on Build GRCh38
rs774508454 Chr.20: 289718-289718 on Build GRCh38
rs752522624 Chr.20: 289616-289616 on Build GRCh38
rs183288558 Chr.20: 289839-289839 on Build GRCh38
rs114292813 Chr.20: 289563-289563 on Build GRCh38
rs1004241442 Chr.20: 289601-289601 on Build GRCh38
rs866977719 Chr.20: 289902-289902 on Build GRCh38
rs11478105 Chr.20: 289774-289774 on Build GRCh38
rs999376793 Chr.20: 289795-289795 on Build GRCh38
rs948527662 Chr.20: 289794-289794 on Build GRCh38
rs751785155 Chr.20: 289665-289665 on Build GRCh38
rs985404432 Chr.20: 289963-289963 on Build GRCh38
rs777448744 Chr.20: 289631-289631 on Build GRCh38
rs369026329 Chr.20: 289617-289617 on Build GRCh38
rs775785622 Chr.20: 289691-289691 on Build GRCh38
rs565341096 Chr.20: 289512-289512 on Build GRCh38
rs563370355 Chr.20: 289851-289851 on Build GRCh38
rs531176170 Chr.20: 289552-289552 on Build GRCh38
rs566824606 Chr.20: 289771-289771 on Build GRCh38
rs369238697 Chr.20: 289538-289538 on Build GRCh38
rs761911002 Chr.20: 289725-289725 on Build GRCh38
rs747686248 Chr.20: 289522-289522 on Build GRCh38
rs760014698 Chr.20: 289555-289555 on Build GRCh38
rs530397827 Chr.20: 289581-289581 on Build GRCh38
rs529039501 Chr.20: 289930-289930 on Build GRCh38
rs889179961 Chr.20: 289854-289854 on Build GRCh38
rs1036541428 Chr.20: 289806-289806 on Build GRCh38
rs770528771 Chr.20: 289652-289652 on Build GRCh38
rs754772928 Chr.20: 289610-289610 on Build GRCh38
rs766326969 Chr.20: 289595-289595 on Build GRCh38
rs957152794 Chr.20: 289766-289766 on Build GRCh38
rs763347240 Chr.20: 289704-289704 on Build GRCh38
rs372739591 Chr.20: 289553-289553 on Build GRCh38
rs201857645 Chr.20: 289562-289562 on Build GRCh38
rs772652544 Chr.20: 289549-289549 on Build GRCh38
rs765635385 Chr.20: 289558-289558 on Build GRCh38
rs761859654 Chr.20: 289589-289589 on Build GRCh38
rs954069821 Chr.20: 289961-289961 on Build GRCh38
rs778512311 Chr.20: 289614-289614 on Build GRCh38
rs928361117 Chr.20: 289805-289805 on Build GRCh38
rs375633351 Chr.20: 289824-289824 on Build GRCh38

View Full Product Details