Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2814895-2815390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C20orf141
Entrez Gene ID
128653
Gene Name
chromosome 20 open reading frame 141
Gene Aliases
dJ860F19.4
UniGene
Hs.352187
Gene Chromosome Location
Chr.20: 2814987-2815830 on Build GRCh38
Transcripts
NM_001256538.1 NM_080739.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5623227 c.200C>A p.T67N Chr.20: 2815384-2815384 on Build GRCh38
7227218 c.27C>A p.P9P Chr.20: 2815211-2815211 on Build GRCh38
5527689 c.9G>A p.R3R Chr.20: 2815193-2815193 on Build GRCh38
4135248 c.71G>A p.G24D Chr.20: 2815255-2815255 on Build GRCh38
5527691 c.78G>A p.G26G Chr.20: 2815262-2815262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542384944 Chr.20: 2815309-2815309 on Build GRCh38
rs781064368 Chr.20: 2815340-2815340 on Build GRCh38
rs201808334 Chr.20: 2815156-2815156 on Build GRCh38
rs749954548 Chr.20: 2815303-2815303 on Build GRCh38
rs943921883 Chr.20: 2815127-2815127 on Build GRCh38
rs140149148 Chr.20: 2815218-2815218 on Build GRCh38
rs754740354 Chr.20: 2815318-2815318 on Build GRCh38
rs1025345489 Chr.20: 2814959-2814959 on Build GRCh38
rs370762036 Chr.20: 2815165-2815165 on Build GRCh38
rs916736670 Chr.20: 2815064-2815064 on Build GRCh38
rs377416880 Chr.20: 2815159-2815159 on Build GRCh38
rs891017434 Chr.20: 2814969-2814969 on Build GRCh38
rs981443009 Chr.20: 2815032-2815032 on Build GRCh38
rs942599418 Chr.20: 2815087-2815087 on Build GRCh38
rs201353059 Chr.20: 2815166-2815166 on Build GRCh38
rs749560600 Chr.20: 2815350-2815350 on Build GRCh38
rs748066707 Chr.20: 2815229-2815229 on Build GRCh38
rs532252353 Chr.20: 2815074-2815074 on Build GRCh38
rs566548168 Chr.20: 2815049-2815049 on Build GRCh38
rs374163831 Chr.20: 2815285-2815285 on Build GRCh38
rs12625619 Chr.20: 2815361-2815361 on Build GRCh38
rs530919799 Chr.20: 2815048-2815048 on Build GRCh38
rs780300645 Chr.20: 2815349-2815349 on Build GRCh38
rs187897075 Chr.20: 2814947-2814947 on Build GRCh38
rs763432652 Chr.20: 2815284-2815284 on Build GRCh38
rs903513663 Chr.20: 2814937-2814937 on Build GRCh38
rs148113970 Chr.20: 2815052-2815052 on Build GRCh38
rs992311552 Chr.20: 2815063-2815063 on Build GRCh38
rs762468233 Chr.20: 2815289-2815289 on Build GRCh38
rs774869772 Chr.20: 2815355-2815355 on Build GRCh38
rs886842754 Chr.20: 2815122-2815122 on Build GRCh38
rs745990496 Chr.20: 2815346-2815346 on Build GRCh38
rs371152917 Chr.20: 2815246-2815246 on Build GRCh38
rs141922354 Chr.20: 2815150-2815150 on Build GRCh38
rs553305824 Chr.20: 2815077-2815077 on Build GRCh38
rs756804548 Chr.20: 2815221-2815221 on Build GRCh38
rs193060559 Chr.20: 2815106-2815106 on Build GRCh38
rs754057137 Chr.20: 2814955-2814955 on Build GRCh38
rs149402527 Chr.20: 2815192-2815192 on Build GRCh38
rs769858186 Chr.20: 2815348-2815348 on Build GRCh38
rs752613186 Chr.20: 2815333-2815333 on Build GRCh38
rs1046855874 Chr.20: 2815290-2815290 on Build GRCh38
rs773239420 Chr.20: 2815183-2815183 on Build GRCh38
rs749124398 Chr.20: 2815176-2815176 on Build GRCh38
rs781387739 Chr.20: 2815271-2815271 on Build GRCh38
rs116718424 Chr.20: 2815083-2815083 on Build GRCh38
rs143838675 Chr.20: 2815219-2815219 on Build GRCh38
rs951168564 Chr.20: 2814972-2814972 on Build GRCh38
rs754863190 Chr.20: 2815250-2815250 on Build GRCh38
rs760207763 Chr.20: 2815305-2815305 on Build GRCh38
rs765507987 Chr.20: 2815357-2815360 on Build GRCh38
rs147547175 Chr.20: 2815211-2815211 on Build GRCh38
rs779648219 Chr.20: 2815242-2815242 on Build GRCh38
rs969642993 Chr.20: 2815018-2815018 on Build GRCh38
rs898454649 Chr.20: 2815104-2815104 on Build GRCh38
rs991773920 Chr.20: 2814978-2814978 on Build GRCh38
rs778804020 Chr.20: 2815264-2815264 on Build GRCh38
rs776935421 Chr.20: 2815136-2815136 on Build GRCh38
rs755806399 Chr.20: 2815234-2815234 on Build GRCh38
rs952082315 Chr.20: 2815039-2815039 on Build GRCh38
rs753486981 Chr.20: 2815313-2815313 on Build GRCh38
rs147238709 Chr.20: 2815326-2815326 on Build GRCh38
rs765246023 Chr.20: 2815016-2815016 on Build GRCh38
rs763301245 Chr.20: 2815215-2815215 on Build GRCh38
rs1018100336 Chr.20: 2814983-2814983 on Build GRCh38
rs779960065 Chr.20: 2815172-2815172 on Build GRCh38
rs78096730 Chr.20: 2815133-2815133 on Build GRCh38
rs1013252803 Chr.20: 2815314-2815314 on Build GRCh38
rs919827097 Chr.20: 2815268-2815268 on Build GRCh38
rs768588928 Chr.20: 2815177-2815177 on Build GRCh38
rs548656776 Chr.20: 2814956-2814956 on Build GRCh38
rs1015143340 Chr.20: 2815050-2815050 on Build GRCh38
rs200127783 Chr.20: 2815205-2815205 on Build GRCh38
rs1051744777 Chr.20: 2815079-2815079 on Build GRCh38
rs369961888 Chr.20: 2815153-2815153 on Build GRCh38
rs1046910775 Chr.20: 2815110-2815110 on Build GRCh38
rs770058169 Chr.20: 2815280-2815280 on Build GRCh38
rs775675120 Chr.20: 2815282-2815282 on Build GRCh38
rs529897750 Chr.20: 2815265-2815265 on Build GRCh38
rs775934073 Chr.20: 2815214-2815214 on Build GRCh38
rs747445102 Chr.20: 2815092-2815092 on Build GRCh38
rs116201210 Chr.20: 2814948-2814948 on Build GRCh38
rs867291866 Chr.20: 2815160-2815160 on Build GRCh38
rs542997052 Chr.20: 2815146-2815146 on Build GRCh38
rs770991057 Chr.20: 2815266-2815266 on Build GRCh38
rs771052342 Chr.20: 2815196-2815196 on Build GRCh38
rs758342612 Chr.20: 2815335-2815335 on Build GRCh38
rs377743222 Chr.20: 2815204-2815204 on Build GRCh38
rs925439876 Chr.20: 2815036-2815036 on Build GRCh38
rs200722113 Chr.20: 2815124-2815124 on Build GRCh38
rs535967780 Chr.20: 2815099-2815099 on Build GRCh38
rs887830397 Chr.20: 2815161-2815161 on Build GRCh38
rs778499041 Chr.20: 2815228-2815228 on Build GRCh38
rs746016551 Chr.20: 2815274-2815274 on Build GRCh38
rs995741587 Chr.20: 2815107-2815107 on Build GRCh38
rs769130162 Chr.20: 2815354-2815354 on Build GRCh38
rs765504886 Chr.20: 2815212-2815212 on Build GRCh38
rs996266474 Chr.20: 2815034-2815034 on Build GRCh38
rs913979271 Chr.20: 2815068-2815068 on Build GRCh38
rs377607985 Chr.20: 2815292-2815292 on Build GRCh38
rs552625562 Chr.20: 2815369-2815369 on Build GRCh38
rs765670560 Chr.20: 2815148-2815148 on Build GRCh38
rs1018080071 Chr.20: 2814933-2814933 on Build GRCh38
rs914094334 Chr.20: 2815193-2815193 on Build GRCh38
rs757686904 Chr.20: 2815269-2815269 on Build GRCh38
rs215560 Chr.20: 2814979-2814979 on Build GRCh38

View Full Product Details