Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2835956-2836457 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCED1A
Entrez Gene ID
64773
Gene Name
PC-esterase domain containing 1A
Gene Aliases
C20orf81, FAM113A, bA12M19.1
UniGene
Hs.29341
Gene Chromosome Location
Chr.20: 2835314-2841243 on Build GRCh38
Transcripts
NM_001271168.1 NM_022760.4 XM_005260804.2 XM_005260805.2 XM_005260806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1751368 c.848G>C p.W283S Chr.20: 2836308-2836308 on Build GRCh38
3840643 c.936A>C p.P312P Chr.20: 2836220-2836220 on Build GRCh38
5511140 c.846G>A p.P282P Chr.20: 2836310-2836310 on Build GRCh38
1251705 c.1051C>T p.P351S Chr.20: 2836105-2836105 on Build GRCh38
3544972 c.940C>T p.P314S Chr.20: 2836216-2836216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144737230 Chr.20: 2836116-2836116 on Build GRCh38
rs766884222 Chr.20: 2836235-2836235 on Build GRCh38
rs763782803 Chr.20: 2836352-2836352 on Build GRCh38
rs760131200 Chr.20: 2836330-2836330 on Build GRCh38
rs775280488 Chr.20: 2836247-2836253 on Build GRCh38
rs769680922 Chr.20: 2836204-2836206 on Build GRCh38
rs762134179 Chr.20: 2836097-2836097 on Build GRCh38
rs966432609 Chr.20: 2836397-2836397 on Build GRCh38
rs772354602 Chr.20: 2836227-2836227 on Build GRCh38
rs555113168 Chr.20: 2836189-2836189 on Build GRCh38
rs761248958 Chr.20: 2836140-2836140 on Build GRCh38
rs777179912 Chr.20: 2836311-2836311 on Build GRCh38
rs990293435 Chr.20: 2836122-2836122 on Build GRCh38
rs1001639528 Chr.20: 2836289-2836289 on Build GRCh38
rs1013965900 Chr.20: 2836259-2836259 on Build GRCh38
rs761038739 Chr.20: 2836234-2836234 on Build GRCh38
rs765765075 Chr.20: 2836141-2836141 on Build GRCh38
rs892957142 Chr.20: 2836409-2836409 on Build GRCh38
rs779952575 Chr.20: 2836210-2836210 on Build GRCh38
rs369459163 Chr.20: 2836215-2836215 on Build GRCh38
rs527740863 Chr.20: 2836039-2836039 on Build GRCh38
rs754580140 Chr.20: 2836146-2836146 on Build GRCh38
rs769708727 Chr.20: 2836042-2836042 on Build GRCh38
rs971295319 Chr.20: 2836045-2836045 on Build GRCh38
rs887669382 Chr.20: 2836182-2836182 on Build GRCh38
rs755876387 Chr.20: 2836263-2836263 on Build GRCh38
rs2274669 Chr.20: 2836175-2836175 on Build GRCh38
rs151220721 Chr.20: 2836257-2836257 on Build GRCh38
rs773654732 Chr.20: 2836230-2836230 on Build GRCh38
rs773355386 Chr.20: 2836296-2836296 on Build GRCh38
rs371113087 Chr.20: 2836007-2836007 on Build GRCh38
rs913008721 Chr.20: 2836133-2836133 on Build GRCh38
rs557505834 Chr.20: 2836218-2836218 on Build GRCh38
rs753409794 Chr.20: 2836144-2836144 on Build GRCh38
rs747029732 Chr.20: 2836041-2836041 on Build GRCh38
rs759110127 Chr.20: 2836240-2836240 on Build GRCh38
rs139908823 Chr.20: 2836219-2836219 on Build GRCh38
rs140133504 Chr.20: 2836310-2836310 on Build GRCh38
rs781153226 Chr.20: 2836264-2836264 on Build GRCh38
rs751122175 Chr.20: 2836035-2836035 on Build GRCh38
rs200413942 Chr.20: 2836206-2836206 on Build GRCh38
rs762649688 Chr.20: 2836348-2836348 on Build GRCh38
rs1001403895 Chr.20: 2836380-2836380 on Build GRCh38
rs931499954 Chr.20: 2836223-2836223 on Build GRCh38
rs772303555 Chr.20: 2836291-2836291 on Build GRCh38
rs745875389 Chr.20: 2836269-2836269 on Build GRCh38
rs538199123 Chr.20: 2836069-2836069 on Build GRCh38
rs764777241 Chr.20: 2836342-2836342 on Build GRCh38
rs761065287 Chr.20: 2836305-2836305 on Build GRCh38
rs752343671 Chr.20: 2836244-2836244 on Build GRCh38
rs763018980 Chr.20: 2836364-2836365 on Build GRCh38
rs1039906489 Chr.20: 2835988-2835988 on Build GRCh38
rs1005219058 Chr.20: 2836231-2836231 on Build GRCh38
rs768711454 Chr.20: 2836270-2836270 on Build GRCh38
rs6051426 Chr.20: 2836385-2836385 on Build GRCh38
rs76189925 Chr.20: 2836139-2836139 on Build GRCh38
rs1001284692 Chr.20: 2836047-2836047 on Build GRCh38
rs201461695 Chr.20: 2836164-2836164 on Build GRCh38
rs775174130 Chr.20: 2836345-2836345 on Build GRCh38
rs200739809 Chr.20: 2836056-2836056 on Build GRCh38
rs148521607 Chr.20: 2836161-2836161 on Build GRCh38
rs142408621 Chr.20: 2836245-2836245 on Build GRCh38
rs1034252357 Chr.20: 2836396-2836396 on Build GRCh38
rs376885720 Chr.20: 2836005-2836005 on Build GRCh38
rs777395249 Chr.20: 2836254-2836254 on Build GRCh38
rs763048760 Chr.20: 2836051-2836051 on Build GRCh38
rs944767949 Chr.20: 2836160-2836160 on Build GRCh38
rs754338314 Chr.20: 2836238-2836238 on Build GRCh38
rs868726822 Chr.20: 2836098-2836098 on Build GRCh38
rs748327901 Chr.20: 2836282-2836282 on Build GRCh38
rs79089526 Chr.20: 2836408-2836408 on Build GRCh38
rs898702235 Chr.20: 2835989-2835989 on Build GRCh38
rs201605909 Chr.20: 2836046-2836046 on Build GRCh38
rs887088256 Chr.20: 2836025-2836025 on Build GRCh38
rs778967448 Chr.20: 2836278-2836278 on Build GRCh38
rs75415902 Chr.20: 2836176-2836176 on Build GRCh38
rs764895840 Chr.20: 2836243-2836243 on Build GRCh38
rs957642950 Chr.20: 2836060-2836060 on Build GRCh38
rs199605064 Chr.20: 2836214-2836214 on Build GRCh38

View Full Product Details