Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 2838754-2839250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PCED1A
Entrez Gene ID
64773
Gene Name
PC-esterase domain containing 1A
Gene Aliases
C20orf81, FAM113A, bA12M19.1
UniGene
Hs.29341
Gene Chromosome Location
Chr.20: 2835314-2841243 on Build GRCh38
Transcripts
NM_001271168.1 NM_022760.4 XM_005260804.2 XM_005260805.2 XM_005260806.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1251704 c.205-8delG p.? Chr.20: 2839090-2839090 on Build GRCh38
3963443 c.305G>T p.G102V Chr.20: 2838982-2838982 on Build GRCh38
5421205 c.419A>G p.N140S Chr.20: 2838868-2838868 on Build GRCh38
6525587 c.343C>T p.R115C Chr.20: 2838944-2838944 on Build GRCh38
5484367 c.280C>T p.R94C Chr.20: 2839007-2839007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764900179 Chr.20: 2839204-2839204 on Build GRCh38
rs200026517 Chr.20: 2838797-2838797 on Build GRCh38
rs201973881 Chr.20: 2839117-2839117 on Build GRCh38
rs745927668 Chr.20: 2838908-2838908 on Build GRCh38
rs776303385 Chr.20: 2839092-2839092 on Build GRCh38
rs915236115 Chr.20: 2839030-2839030 on Build GRCh38
rs771542382 Chr.20: 2839167-2839167 on Build GRCh38
rs762518607 Chr.20: 2839216-2839216 on Build GRCh38
rs775680046 Chr.20: 2838926-2838926 on Build GRCh38
rs147093102 Chr.20: 2838919-2838919 on Build GRCh38
rs750955680 Chr.20: 2838962-2838962 on Build GRCh38
rs962454525 Chr.20: 2838824-2838824 on Build GRCh38
rs982273544 Chr.20: 2838911-2838911 on Build GRCh38
rs916407862 Chr.20: 2839089-2839089 on Build GRCh38
rs1006537023 Chr.20: 2838814-2838814 on Build GRCh38
rs768891725 Chr.20: 2839077-2839077 on Build GRCh38
rs778979321 Chr.20: 2838776-2838776 on Build GRCh38
rs759111796 Chr.20: 2839191-2839191 on Build GRCh38
rs774729351 Chr.20: 2838947-2838947 on Build GRCh38
rs776799370 Chr.20: 2838799-2838799 on Build GRCh38
rs553600846 Chr.20: 2839050-2839050 on Build GRCh38
rs751447559 Chr.20: 2839104-2839104 on Build GRCh38
rs569893220 Chr.20: 2839208-2839208 on Build GRCh38
rs758514863 Chr.20: 2839084-2839085 on Build GRCh38
rs765996867 Chr.20: 2838985-2838985 on Build GRCh38
rs200352855 Chr.20: 2839112-2839112 on Build GRCh38
rs181003440 Chr.20: 2838811-2838811 on Build GRCh38
rs777950791 Chr.20: 2839084-2839089 on Build GRCh38
rs762387006 Chr.20: 2838816-2838816 on Build GRCh38
rs764504882 Chr.20: 2838813-2838813 on Build GRCh38
rs764951009 Chr.20: 2839094-2839094 on Build GRCh38
rs548170569 Chr.20: 2839158-2839158 on Build GRCh38
rs368046555 Chr.20: 2839166-2839166 on Build GRCh38
rs372922557 Chr.20: 2838834-2838834 on Build GRCh38
rs762179453 Chr.20: 2838951-2838951 on Build GRCh38
rs202246106 Chr.20: 2838800-2838800 on Build GRCh38
rs777316526 Chr.20: 2839182-2839182 on Build GRCh38
rs760216654 Chr.20: 2838983-2838983 on Build GRCh38
rs768942602 Chr.20: 2838941-2838941 on Build GRCh38
rs770033439 Chr.20: 2838803-2838803 on Build GRCh38
rs138501389 Chr.20: 2838984-2838984 on Build GRCh38
rs778914220 Chr.20: 2839034-2839051 on Build GRCh38
rs530988995 Chr.20: 2838836-2838836 on Build GRCh38
rs748109959 Chr.20: 2838783-2838783 on Build GRCh38
rs774608518 Chr.20: 2839084-2839084 on Build GRCh38
rs768047955 Chr.20: 2838961-2838961 on Build GRCh38
rs933800407 Chr.20: 2839221-2839221 on Build GRCh38
rs752512694 Chr.20: 2839020-2839020 on Build GRCh38
rs200852591 Chr.20: 2838885-2838885 on Build GRCh38
rs754708835 Chr.20: 2838998-2838998 on Build GRCh38
rs773707666 Chr.20: 2839090-2839090 on Build GRCh38
rs763180365 Chr.20: 2838933-2838933 on Build GRCh38
rs772410826 Chr.20: 2839164-2839164 on Build GRCh38
rs765075651 Chr.20: 2839005-2839005 on Build GRCh38
rs375558975 Chr.20: 2839140-2839140 on Build GRCh38
rs866469982 Chr.20: 2839040-2839040 on Build GRCh38
rs763910162 Chr.20: 2839228-2839228 on Build GRCh38
rs758201018 Chr.20: 2839096-2839096 on Build GRCh38
rs369121124 Chr.20: 2838872-2838872 on Build GRCh38
rs763809754 Chr.20: 2839098-2839098 on Build GRCh38
rs916346502 Chr.20: 2839160-2839160 on Build GRCh38
rs761087856 Chr.20: 2839091-2839091 on Build GRCh38
rs747317971 Chr.20: 2839085-2839086 on Build GRCh38
rs377396516 Chr.20: 2839032-2839032 on Build GRCh38
rs766898930 Chr.20: 2838817-2838817 on Build GRCh38
rs571531085 Chr.20: 2839062-2839062 on Build GRCh38
rs752356696 Chr.20: 2839095-2839095 on Build GRCh38
rs748260316 Chr.20: 2839078-2839079 on Build GRCh38
rs749476896 Chr.20: 2839076-2839076 on Build GRCh38
rs758165691 Chr.20: 2839022-2839022 on Build GRCh38
rs990517993 Chr.20: 2839146-2839146 on Build GRCh38
rs374547907 Chr.20: 2839031-2839031 on Build GRCh38
rs902045240 Chr.20: 2838870-2838870 on Build GRCh38
rs775361979 Chr.20: 2839089-2839090 on Build GRCh38
rs748517314 Chr.20: 2839085-2839085 on Build GRCh38
rs768667049 Chr.20: 2839090-2839090 on Build GRCh38
rs1042865653 Chr.20: 2839084-2839084 on Build GRCh38
rs565534307 Chr.20: 2839116-2839116 on Build GRCh38
rs34194713 Chr.20: 2838867-2838867 on Build GRCh38
rs779180097 Chr.20: 2839151-2839151 on Build GRCh38
rs753469763 Chr.20: 2838997-2838997 on Build GRCh38
rs369507132 Chr.20: 2838815-2838815 on Build GRCh38
rs370508727 Chr.20: 2839175-2839175 on Build GRCh38
rs979122759 Chr.20: 2839203-2839203 on Build GRCh38
rs759165162 Chr.20: 2839093-2839093 on Build GRCh38
rs371631991 Chr.20: 2839007-2839007 on Build GRCh38
rs373091650 Chr.20: 2838901-2838901 on Build GRCh38
rs373653313 Chr.20: 2839052-2839052 on Build GRCh38

View Full Product Details