Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3547196-3547702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRN
Entrez Gene ID
8455
Gene Name
attractin
Gene Aliases
DPPT-L, MGCA
UniGene
Hs.276252
Gene Chromosome Location
Chr.20: 3471018-3651122 on Build GRCh38
Transcripts
NM_001207047.2 NM_001323332.1 NM_139321.2 NM_139322.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6438807 c.755A>G p.N252S Chr.20: 3547301-3547301 on Build GRCh38
3927769 c.943+2T>G p.? Chr.20: 3547491-3547491 on Build GRCh38
6438806 c.755A>G p.N252S Chr.20: 3547301-3547301 on Build GRCh38
6093253 c.792T>C p.N264N Chr.20: 3547338-3547338 on Build GRCh38
7221708 c.886C>T p.R296* Chr.20: 3547432-3547432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749481178 Chr.20: 3547325-3547325 on Build GRCh38
rs759262534 Chr.20: 3547257-3547257 on Build GRCh38
rs760194714 Chr.20: 3547251-3547251 on Build GRCh38
rs536310570 Chr.20: 3547418-3547418 on Build GRCh38
rs199516815 Chr.20: 3547488-3547488 on Build GRCh38
rs150589475 Chr.20: 3547438-3547438 on Build GRCh38
rs1016675848 Chr.20: 3547543-3547543 on Build GRCh38
rs239187 Chr.20: 3547237-3547237 on Build GRCh38
rs239188 Chr.20: 3547248-3547248 on Build GRCh38
rs774183730 Chr.20: 3547456-3547456 on Build GRCh38
rs563446138 Chr.20: 3547243-3547243 on Build GRCh38
rs6107308 Chr.20: 3547454-3547454 on Build GRCh38
rs776808161 Chr.20: 3547401-3547401 on Build GRCh38
rs189706633 Chr.20: 3547666-3547666 on Build GRCh38
rs540107990 Chr.20: 3547574-3547574 on Build GRCh38
rs947231523 Chr.20: 3547493-3547493 on Build GRCh38
rs752025078 Chr.20: 3547511-3547511 on Build GRCh38
rs762754702 Chr.20: 3547389-3547389 on Build GRCh38
rs758595124 Chr.20: 3547461-3547468 on Build GRCh38
rs375560346 Chr.20: 3547331-3547331 on Build GRCh38
rs148724830 Chr.20: 3547608-3547608 on Build GRCh38
rs981702313 Chr.20: 3547657-3547657 on Build GRCh38
rs750330928 Chr.20: 3547470-3547470 on Build GRCh38
rs1040807100 Chr.20: 3547580-3547580 on Build GRCh38
rs752583734 Chr.20: 3547285-3547285 on Build GRCh38
rs758730661 Chr.20: 3547503-3547503 on Build GRCh38
rs758267933 Chr.20: 3547297-3547297 on Build GRCh38
rs570081534 Chr.20: 3547328-3547328 on Build GRCh38
rs972834920 Chr.20: 3547582-3547582 on Build GRCh38
rs201440872 Chr.20: 3547345-3547345 on Build GRCh38
rs76980512 Chr.20: 3547514-3547514 on Build GRCh38
rs765010172 Chr.20: 3547363-3547363 on Build GRCh38
rs755148899 Chr.20: 3547411-3547411 on Build GRCh38
rs763846365 Chr.20: 3547455-3547455 on Build GRCh38
rs753977218 Chr.20: 3547405-3547405 on Build GRCh38
rs574040216 Chr.20: 3547664-3547664 on Build GRCh38
rs751514447 Chr.20: 3547316-3547316 on Build GRCh38
rs747482817 Chr.20: 3547420-3547420 on Build GRCh38
rs780140690 Chr.20: 3547323-3547323 on Build GRCh38
rs751459326 Chr.20: 3547395-3547395 on Build GRCh38
rs1029082702 Chr.20: 3547604-3547604 on Build GRCh38
rs767508779 Chr.20: 3547469-3547469 on Build GRCh38
rs906713102 Chr.20: 3547388-3547388 on Build GRCh38
rs774017880 Chr.20: 3547596-3547596 on Build GRCh38
rs112144103 Chr.20: 3547627-3547627 on Build GRCh38
rs748332214 Chr.20: 3547416-3547416 on Build GRCh38
rs763901740 Chr.20: 3547393-3547393 on Build GRCh38
rs962312640 Chr.20: 3547643-3547643 on Build GRCh38
rs781742757 Chr.20: 3547538-3547538 on Build GRCh38
rs771612123 Chr.20: 3547427-3547427 on Build GRCh38
rs141713516 Chr.20: 3547349-3547349 on Build GRCh38
rs748613292 Chr.20: 3547333-3547333 on Build GRCh38
rs558196805 Chr.20: 3547578-3547578 on Build GRCh38
rs752921148 Chr.20: 3547489-3547489 on Build GRCh38
rs770522690 Chr.20: 3547347-3547347 on Build GRCh38
rs772326371 Chr.20: 3547341-3547341 on Build GRCh38
rs764906552 Chr.20: 3547259-3547259 on Build GRCh38
rs749536344 Chr.20: 3547236-3547236 on Build GRCh38
rs775014369 Chr.20: 3547382-3547382 on Build GRCh38
rs756139477 Chr.20: 3547318-3547318 on Build GRCh38
rs373231195 Chr.20: 3547360-3547360 on Build GRCh38
rs1006427538 Chr.20: 3547609-3547609 on Build GRCh38
rs996558495 Chr.20: 3547602-3547602 on Build GRCh38
rs766426779 Chr.20: 3547402-3547402 on Build GRCh38
rs989588303 Chr.20: 3547491-3547491 on Build GRCh38
rs772692421 Chr.20: 3547247-3547247 on Build GRCh38
rs138628890 Chr.20: 3547445-3547445 on Build GRCh38
rs777702069 Chr.20: 3547300-3547300 on Build GRCh38
rs373042007 Chr.20: 3547253-3547253 on Build GRCh38
rs370607029 Chr.20: 3547504-3547504 on Build GRCh38
rs767309450 Chr.20: 3547488-3547489 on Build GRCh38
rs941029010 Chr.20: 3547315-3547315 on Build GRCh38
rs369774374 Chr.20: 3547344-3547344 on Build GRCh38
rs903150116 Chr.20: 3547555-3547555 on Build GRCh38
rs779121969 Chr.20: 3547414-3547414 on Build GRCh38
rs915309684 Chr.20: 3547633-3547633 on Build GRCh38
rs761726916 Chr.20: 3547464-3547464 on Build GRCh38
rs919543996 Chr.20: 3547329-3547329 on Build GRCh38
rs181721247 Chr.20: 3547462-3547462 on Build GRCh38
rs375979101 Chr.20: 3547242-3547242 on Build GRCh38
rs573306149 Chr.20: 3547585-3547585 on Build GRCh38
rs376268737 Chr.20: 3547451-3547451 on Build GRCh38

View Full Product Details