Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3549130-3549668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATRN
Entrez Gene ID
8455
Gene Name
attractin
Gene Aliases
DPPT-L, MGCA
UniGene
Hs.276252
Gene Chromosome Location
Chr.20: 3471018-3651122 on Build GRCh38
Transcripts
NM_001207047.2 NM_001323332.1 NM_139321.2 NM_139322.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3933857 c.992G>T p.R331L Chr.20: 3549218-3549218 on Build GRCh38
6278258 c.991C>T p.R331* Chr.20: 3549217-3549217 on Build GRCh38
1713110 c.980C>T p.S327L Chr.20: 3549206-3549206 on Build GRCh38
5883550 c.1036G>T p.A346S Chr.20: 3549262-3549262 on Build GRCh38
1411508 c.1013A>C p.K338T Chr.20: 3549239-3549239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028043917 Chr.20: 3549175-3549175 on Build GRCh38
rs191709090 Chr.20: 3549548-3549548 on Build GRCh38
rs757275611 Chr.20: 3549520-3549520 on Build GRCh38
rs760560964 Chr.20: 3549182-3549182 on Build GRCh38
rs772840717 Chr.20: 3549205-3549205 on Build GRCh38
rs549829925 Chr.20: 3549283-3549283 on Build GRCh38
rs763072583 Chr.20: 3549321-3549321 on Build GRCh38
rs763251726 Chr.20: 3549231-3549231 on Build GRCh38
rs201622481 Chr.20: 3549346-3549346 on Build GRCh38
rs766246897 Chr.20: 3549561-3549561 on Build GRCh38
rs765324915 Chr.20: 3549188-3549188 on Build GRCh38
rs938670425 Chr.20: 3549278-3549278 on Build GRCh38
rs747561144 Chr.20: 3549437-3549437 on Build GRCh38
rs748817073 Chr.20: 3549284-3549284 on Build GRCh38
rs567686443 Chr.20: 3549393-3549393 on Build GRCh38
rs374246567 Chr.20: 3549250-3549250 on Build GRCh38
rs371587385 Chr.20: 3549530-3549530 on Build GRCh38
rs751847658 Chr.20: 3549233-3549233 on Build GRCh38
rs764474867 Chr.20: 3549232-3549232 on Build GRCh38
rs768157365 Chr.20: 3549285-3549285 on Build GRCh38
rs771865961 Chr.20: 3549156-3549156 on Build GRCh38
rs756641962 Chr.20: 3549351-3549351 on Build GRCh38
rs1055632585 Chr.20: 3549605-3549605 on Build GRCh38
rs954030033 Chr.20: 3549178-3549178 on Build GRCh38
rs1043200196 Chr.20: 3549345-3549345 on Build GRCh38
rs775631692 Chr.20: 3549309-3549309 on Build GRCh38
rs754355538 Chr.20: 3549386-3549386 on Build GRCh38
rs757649933 Chr.20: 3549243-3549243 on Build GRCh38
rs919240372 Chr.20: 3549244-3549244 on Build GRCh38
rs556664653 Chr.20: 3549465-3549465 on Build GRCh38
rs750201064 Chr.20: 3549156-3549157 on Build GRCh38
rs762169974 Chr.20: 3549338-3549338 on Build GRCh38
rs781276657 Chr.20: 3549560-3549560 on Build GRCh38
rs145181041 Chr.20: 3549304-3549304 on Build GRCh38
rs1042785586 Chr.20: 3549621-3549621 on Build GRCh38
rs537830495 Chr.20: 3549397-3549397 on Build GRCh38
rs778477025 Chr.20: 3549292-3549292 on Build GRCh38
rs961276222 Chr.20: 3549230-3549230 on Build GRCh38
rs751796320 Chr.20: 3549334-3549334 on Build GRCh38
rs377163365 Chr.20: 3549245-3549245 on Build GRCh38
rs983913453 Chr.20: 3549184-3549184 on Build GRCh38
rs549126574 Chr.20: 3549372-3549372 on Build GRCh38
rs773153051 Chr.20: 3549160-3549160 on Build GRCh38
rs535059564 Chr.20: 3549498-3549498 on Build GRCh38
rs866728425 Chr.20: 3549570-3549570 on Build GRCh38
rs750911567 Chr.20: 3549260-3549260 on Build GRCh38
rs139431664 Chr.20: 3549300-3549300 on Build GRCh38
rs1052748280 Chr.20: 3549380-3549380 on Build GRCh38
rs909806675 Chr.20: 3549217-3549217 on Build GRCh38
rs928500697 Chr.20: 3549542-3549542 on Build GRCh38
rs750811817 Chr.20: 3549350-3549350 on Build GRCh38
rs539257459 Chr.20: 3549617-3549617 on Build GRCh38
rs1018347650 Chr.20: 3549406-3549406 on Build GRCh38
rs867467663 Chr.20: 3549289-3549289 on Build GRCh38
rs947114275 Chr.20: 3549610-3549610 on Build GRCh38
rs779604268 Chr.20: 3549273-3549273 on Build GRCh38
rs764294840 Chr.20: 3549331-3549331 on Build GRCh38
rs150469117 Chr.20: 3549423-3549426 on Build GRCh38
rs867624838 Chr.20: 3549611-3549611 on Build GRCh38
rs147729396 Chr.20: 3549305-3549305 on Build GRCh38
rs147361324 Chr.20: 3549271-3549271 on Build GRCh38
rs756629370 Chr.20: 3549270-3549270 on Build GRCh38
rs915684990 Chr.20: 3549607-3549607 on Build GRCh38
rs752765173 Chr.20: 3549218-3549218 on Build GRCh38

View Full Product Details