Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 68476685-68477232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEPDC1
Entrez Gene ID
55635
Gene Name
DEP domain containing 1
Gene Aliases
DEP.8, DEPDC1-V2, DEPDC1A, SDP35
UniGene
Hs.445098
Gene Chromosome Location
Chr.1: 68474152-68497221 on Build GRCh38
Transcripts
NM_001114120.2 NM_017779.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
911897 c.1502A>G p.E501G Chr.1: 68477014-68477014 on Build GRCh38
911896 c.2354A>G p.E785G Chr.1: 68477014-68477014 on Build GRCh38
5381467 c.1462C>T p.P488S Chr.1: 68477054-68477054 on Build GRCh38
7423111 c.2422C>T p.R808C Chr.1: 68476946-68476946 on Build GRCh38
6485999 c.2344A>T p.T782S Chr.1: 68477024-68477024 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750680947 Chr.1: 68476886-68476886 on Build GRCh38
rs758224175 Chr.1: 68476893-68476893 on Build GRCh38
rs779770378 Chr.1: 68477027-68477027 on Build GRCh38
rs35067316 Chr.1: 68476857-68476858 on Build GRCh38
rs1018491060 Chr.1: 68476740-68476740 on Build GRCh38
rs937502244 Chr.1: 68476845-68476845 on Build GRCh38
rs370916107 Chr.1: 68476951-68476951 on Build GRCh38
rs377757010 Chr.1: 68476885-68476885 on Build GRCh38
rs766315169 Chr.1: 68477065-68477065 on Build GRCh38
rs944697980 Chr.1: 68477114-68477114 on Build GRCh38
rs767901490 Chr.1: 68477079-68477079 on Build GRCh38
rs745563506 Chr.1: 68476938-68476938 on Build GRCh38
rs61739526 Chr.1: 68477056-68477056 on Build GRCh38
rs761996516 Chr.1: 68477006-68477006 on Build GRCh38
rs764039714 Chr.1: 68477086-68477086 on Build GRCh38
rs547743206 Chr.1: 68476787-68476787 on Build GRCh38
rs981635272 Chr.1: 68476821-68476821 on Build GRCh38
rs774688113 Chr.1: 68477053-68477053 on Build GRCh38
rs760835070 Chr.1: 68477060-68477060 on Build GRCh38
rs753955255 Chr.1: 68477002-68477002 on Build GRCh38
rs141814200 Chr.1: 68477169-68477169 on Build GRCh38
rs148054870 Chr.1: 68477160-68477160 on Build GRCh38
rs538609171 Chr.1: 68476736-68476736 on Build GRCh38
rs370272621 Chr.1: 68476900-68476900 on Build GRCh38
rs377577562 Chr.1: 68476945-68476945 on Build GRCh38
rs769077645 Chr.1: 68477049-68477049 on Build GRCh38
rs1008076511 Chr.1: 68476711-68476712 on Build GRCh38
rs757852645 Chr.1: 68476887-68476887 on Build GRCh38
rs34349147 Chr.1: 68476750-68476750 on Build GRCh38
rs561161572 Chr.1: 68477139-68477139 on Build GRCh38
rs200717783 Chr.1: 68477014-68477014 on Build GRCh38
rs144066767 Chr.1: 68476940-68476940 on Build GRCh38
rs777713894 Chr.1: 68476942-68476942 on Build GRCh38
rs142787595 Chr.1: 68476751-68476751 on Build GRCh38
rs574952697 Chr.1: 68476985-68476985 on Build GRCh38
rs764994928 Chr.1: 68476890-68476890 on Build GRCh38
rs768952825 Chr.1: 68476946-68476946 on Build GRCh38
rs182395472 Chr.1: 68477145-68477145 on Build GRCh38
rs1020663612 Chr.1: 68477130-68477130 on Build GRCh38
rs773323414 Chr.1: 68476978-68476978 on Build GRCh38
rs929663140 Chr.1: 68476867-68476867 on Build GRCh38
rs757245539 Chr.1: 68477011-68477011 on Build GRCh38
rs988785476 Chr.1: 68476778-68476778 on Build GRCh38
rs566375684 Chr.1: 68476939-68476939 on Build GRCh38
rs781504455 Chr.1: 68476926-68476929 on Build GRCh38
rs191012228 Chr.1: 68476975-68476975 on Build GRCh38
rs114891085 Chr.1: 68476933-68476933 on Build GRCh38
rs187381505 Chr.1: 68476806-68476806 on Build GRCh38
rs930746398 Chr.1: 68477168-68477168 on Build GRCh38
rs774608716 Chr.1: 68476928-68476928 on Build GRCh38
rs755531273 Chr.1: 68477023-68477023 on Build GRCh38
rs760850614 Chr.1: 68476988-68476988 on Build GRCh38
rs115356775 Chr.1: 68477096-68477096 on Build GRCh38
rs761883461 Chr.1: 68477099-68477099 on Build GRCh38
rs767645513 Chr.1: 68476977-68476977 on Build GRCh38
rs532826584 Chr.1: 68477208-68477208 on Build GRCh38
rs199780991 Chr.1: 68476922-68476922 on Build GRCh38
rs973822384 Chr.1: 68476857-68476857 on Build GRCh38
rs553707743 Chr.1: 68476776-68476776 on Build GRCh38
rs535098066 Chr.1: 68476765-68476765 on Build GRCh38
rs766651307 Chr.1: 68476995-68476995 on Build GRCh38
rs770321605 Chr.1: 68476954-68476954 on Build GRCh38
rs927491456 Chr.1: 68476825-68476825 on Build GRCh38
rs759835285 Chr.1: 68477081-68477081 on Build GRCh38
rs56248801 Chr.1: 68476958-68476958 on Build GRCh38
rs748649501 Chr.1: 68476935-68476936 on Build GRCh38
rs745351903 Chr.1: 68477022-68477022 on Build GRCh38
rs748873861 Chr.1: 68477035-68477035 on Build GRCh38
rs763912769 Chr.1: 68477007-68477007 on Build GRCh38
rs1033088818 Chr.1: 68476757-68476757 on Build GRCh38
rs751612490 Chr.1: 68477009-68477009 on Build GRCh38
rs893194643 Chr.1: 68477075-68477075 on Build GRCh38
rs943915550 Chr.1: 68476917-68476917 on Build GRCh38
rs1000689498 Chr.1: 68476956-68476957 on Build GRCh38
rs748699841 Chr.1: 68477054-68477054 on Build GRCh38
rs375420726 Chr.1: 68476729-68476729 on Build GRCh38
rs569724124 Chr.1: 68476722-68476722 on Build GRCh38
rs958887463 Chr.1: 68477187-68477187 on Build GRCh38
rs374281152 Chr.1: 68476902-68476902 on Build GRCh38
rs375812118 Chr.1: 68477055-68477055 on Build GRCh38
rs752047357 Chr.1: 68476860-68476860 on Build GRCh38

View Full Product Details