Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5126671-5127159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDS2
Entrez Gene ID
8760
Gene Name
CDP-diacylglycerol synthase 2
Gene Aliases
-
UniGene
Hs.126857
Gene Chromosome Location
Chr.20: 5126761-5197887 on Build GRCh38
Transcripts
NM_003818.3 XM_006723660.1 XM_011529390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4137046 c.41C>T p.A14V Chr.20: 5127133-5127133 on Build GRCh38
6341831 c.56_57insG p.E20fs*17 Chr.20: 5127148-5127149 on Build GRCh38
5631502 c.28C>T p.H10Y Chr.20: 5127120-5127120 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs530075130 Chr.20: 5126794-5126794 on Build GRCh38
rs925111694 Chr.20: 5126873-5126873 on Build GRCh38
rs1056798513 Chr.20: 5126823-5126823 on Build GRCh38
rs1001231458 Chr.20: 5126815-5126815 on Build GRCh38
rs1018043474 Chr.20: 5127077-5127077 on Build GRCh38
rs576596370 Chr.20: 5127045-5127045 on Build GRCh38
rs976762129 Chr.20: 5126891-5126891 on Build GRCh38
rs768390346 Chr.20: 5127053-5127053 on Build GRCh38
rs540968610 Chr.20: 5127046-5127046 on Build GRCh38
rs892189235 Chr.20: 5126969-5126969 on Build GRCh38
rs1056851262 Chr.20: 5127040-5127040 on Build GRCh38
rs538977706 Chr.20: 5126704-5126704 on Build GRCh38
rs554259077 Chr.20: 5126909-5126909 on Build GRCh38
rs942720032 Chr.20: 5126729-5126729 on Build GRCh38
rs951382544 Chr.20: 5126921-5126921 on Build GRCh38
rs527312329 Chr.20: 5126734-5126734 on Build GRCh38
rs1052194841 Chr.20: 5126953-5126953 on Build GRCh38
rs968957107 Chr.20: 5126849-5126849 on Build GRCh38
rs748063424 Chr.20: 5127110-5127110 on Build GRCh38
rs370206454 Chr.20: 5126748-5126748 on Build GRCh38
rs947958223 Chr.20: 5126864-5126864 on Build GRCh38
rs922599068 Chr.20: 5126901-5126901 on Build GRCh38
rs993210893 Chr.20: 5126843-5126843 on Build GRCh38
rs879095880 Chr.20: 5127001-5127001 on Build GRCh38
rs982879194 Chr.20: 5126782-5126782 on Build GRCh38
rs926886657 Chr.20: 5126974-5126974 on Build GRCh38
rs909945935 Chr.20: 5126950-5126950 on Build GRCh38
rs574633123 Chr.20: 5127076-5127076 on Build GRCh38
rs1010393241 Chr.20: 5126833-5126833 on Build GRCh38
rs749178904 Chr.20: 5126970-5126970 on Build GRCh38
rs779207252 Chr.20: 5127047-5127048 on Build GRCh38
rs113493863 Chr.20: 5127043-5127043 on Build GRCh38
rs1000464762 Chr.20: 5127071-5127071 on Build GRCh38
rs59606548 Chr.20: 5127000-5127001 on Build GRCh38
rs747045753 Chr.20: 5126717-5126717 on Build GRCh38
rs904240725 Chr.20: 5127065-5127065 on Build GRCh38
rs1029497283 Chr.20: 5127128-5127128 on Build GRCh38
rs749059827 Chr.20: 5127050-5127050 on Build GRCh38
rs796649336 Chr.20: 5126878-5126878 on Build GRCh38
rs895606800 Chr.20: 5126826-5126826 on Build GRCh38
rs1044897083 Chr.20: 5126868-5126868 on Build GRCh38
rs536729756 Chr.20: 5127011-5127011 on Build GRCh38
rs997099607 Chr.20: 5127002-5127002 on Build GRCh38
rs781375210 Chr.20: 5126835-5126835 on Build GRCh38
rs905416488 Chr.20: 5126812-5126812 on Build GRCh38
rs939580161 Chr.20: 5127025-5127025 on Build GRCh38
rs983276808 Chr.20: 5126930-5126930 on Build GRCh38
rs1004748826 Chr.20: 5127036-5127036 on Build GRCh38
rs1027189509 Chr.20: 5127019-5127019 on Build GRCh38
rs1047263825 Chr.20: 5126780-5126780 on Build GRCh38
rs895479378 Chr.20: 5127042-5127042 on Build GRCh38
rs34266571 Chr.20: 5126987-5126988 on Build GRCh38
rs866786243 Chr.20: 5127126-5127126 on Build GRCh38
rs553190177 Chr.20: 5127057-5127057 on Build GRCh38
rs930049866 Chr.20: 5126770-5126770 on Build GRCh38
rs565544428 Chr.20: 5126787-5126787 on Build GRCh38
rs536206315 Chr.20: 5126808-5126808 on Build GRCh38
rs900109946 Chr.20: 5126988-5126988 on Build GRCh38
rs529439663 Chr.20: 5127032-5127033 on Build GRCh38
rs770975430 Chr.20: 5127140-5127140 on Build GRCh38
rs1027138495 Chr.20: 5126754-5126754 on Build GRCh38
rs941539218 Chr.20: 5126966-5126966 on Build GRCh38
rs11466986 Chr.20: 5126986-5126987 on Build GRCh38
rs952810667 Chr.20: 5126755-5126755 on Build GRCh38
rs575780306 Chr.20: 5126922-5126922 on Build GRCh38
rs963965935 Chr.20: 5127098-5127098 on Build GRCh38
rs998068794 Chr.20: 5127118-5127118 on Build GRCh38
rs1000006799 Chr.20: 5126851-5126851 on Build GRCh38
rs1018159213 Chr.20: 5126863-5126863 on Build GRCh38
rs1019998005 Chr.20: 5126845-5126845 on Build GRCh38
rs969314559 Chr.20: 5127108-5127108 on Build GRCh38
rs1008271918 Chr.20: 5126692-5126692 on Build GRCh38
rs992869661 Chr.20: 5127061-5127061 on Build GRCh38

View Full Product Details