Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 18595915-18596425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTD1
Entrez Gene ID
92675
Gene Name
D-tyrosyl-tRNA deacylase 1
Gene Aliases
C20orf88, DUE-B, DUEB, HARS2, pqn-68
UniGene
Hs.659442
Gene Chromosome Location
Chr.20: 18587893-18763917 on Build GRCh38
Transcripts
NM_001318043.1 NM_080820.5 XM_005260872.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4837353 c.231C>A p.V77V Chr.20: 18596102-18596102 on Build GRCh38
7365573 c.137T>C p.V46A Chr.20: 18596008-18596008 on Build GRCh38
4679571 c.245T>A p.L82H Chr.20: 18596116-18596116 on Build GRCh38
7323823 c.366C>G p.I122M Chr.20: 18596237-18596237 on Build GRCh38
3544439 c.279C>T p.F93F Chr.20: 18596150-18596150 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760710961 Chr.20: 18595971-18595971 on Build GRCh38
rs749074053 Chr.20: 18596063-18596063 on Build GRCh38
rs1033660715 Chr.20: 18596289-18596289 on Build GRCh38
rs11545149 Chr.20: 18596080-18596080 on Build GRCh38
rs775288002 Chr.20: 18595959-18595959 on Build GRCh38
rs776717723 Chr.20: 18596082-18596082 on Build GRCh38
rs140341803 Chr.20: 18596108-18596108 on Build GRCh38
rs148004891 Chr.20: 18596219-18596219 on Build GRCh38
rs766955834 Chr.20: 18595974-18595974 on Build GRCh38
rs749127844 Chr.20: 18596165-18596165 on Build GRCh38
rs542977068 Chr.20: 18595941-18595941 on Build GRCh38
rs140517016 Chr.20: 18595958-18595958 on Build GRCh38
rs760835609 Chr.20: 18596072-18596072 on Build GRCh38
rs957073378 Chr.20: 18596011-18596011 on Build GRCh38
rs138223373 Chr.20: 18596176-18596176 on Build GRCh38
rs1003993271 Chr.20: 18596228-18596228 on Build GRCh38
rs993170001 Chr.20: 18596134-18596134 on Build GRCh38
rs11545148 Chr.20: 18596157-18596157 on Build GRCh38
rs749824989 Chr.20: 18596277-18596277 on Build GRCh38
rs146076783 Chr.20: 18596189-18596189 on Build GRCh38
rs370656276 Chr.20: 18596168-18596168 on Build GRCh38
rs865993103 Chr.20: 18596144-18596144 on Build GRCh38
rs778740662 Chr.20: 18596245-18596245 on Build GRCh38
rs373430756 Chr.20: 18595968-18595968 on Build GRCh38
rs745796443 Chr.20: 18596028-18596028 on Build GRCh38
rs142292945 Chr.20: 18596185-18596185 on Build GRCh38
rs763536456 Chr.20: 18595970-18595970 on Build GRCh38
rs565278506 Chr.20: 18596120-18596120 on Build GRCh38
rs747973912 Chr.20: 18596253-18596253 on Build GRCh38
rs753802514 Chr.20: 18596227-18596227 on Build GRCh38
rs200181178 Chr.20: 18596008-18596008 on Build GRCh38
rs151273800 Chr.20: 18596094-18596094 on Build GRCh38
rs533829805 Chr.20: 18596325-18596325 on Build GRCh38
rs762035347 Chr.20: 18595963-18595963 on Build GRCh38
rs200560779 Chr.20: 18596186-18596186 on Build GRCh38
rs386813034 Chr.20: 18596267-18596268 on Build GRCh38
rs759634650 Chr.20: 18596184-18596184 on Build GRCh38
rs1022380880 Chr.20: 18596058-18596058 on Build GRCh38
rs753153008 Chr.20: 18596086-18596086 on Build GRCh38
rs372659945 Chr.20: 18595998-18595998 on Build GRCh38
rs777537211 Chr.20: 18595991-18595991 on Build GRCh38
rs757464252 Chr.20: 18596255-18596255 on Build GRCh38
rs11545154 Chr.20: 18596127-18596127 on Build GRCh38
rs753102102 Chr.20: 18595983-18595983 on Build GRCh38
rs763389025 Chr.20: 18596088-18596088 on Build GRCh38
rs779881694 Chr.20: 18596062-18596062 on Build GRCh38
rs754854758 Chr.20: 18596242-18596242 on Build GRCh38
rs909732209 Chr.20: 18596384-18596384 on Build GRCh38
rs951305309 Chr.20: 18596357-18596357 on Build GRCh38
rs760023541 Chr.20: 18596084-18596084 on Build GRCh38
rs138667905 Chr.20: 18596191-18596191 on Build GRCh38
rs772317718 Chr.20: 18596068-18596068 on Build GRCh38
rs144518792 Chr.20: 18596211-18596211 on Build GRCh38
rs868583438 Chr.20: 18596154-18596154 on Build GRCh38
rs375941640 Chr.20: 18595979-18595979 on Build GRCh38
rs768459646 Chr.20: 18596167-18596167 on Build GRCh38
rs775646927 Chr.20: 18596270-18596270 on Build GRCh38
rs763829337 Chr.20: 18596024-18596024 on Build GRCh38
rs777964321 Chr.20: 18596212-18596212 on Build GRCh38
rs983989744 Chr.20: 18595989-18595989 on Build GRCh38
rs776062652 Chr.20: 18595937-18595937 on Build GRCh38
rs761352624 Chr.20: 18596029-18596029 on Build GRCh38
rs757465377 Chr.20: 18596287-18596287 on Build GRCh38
rs755448313 Chr.20: 18595977-18595977 on Build GRCh38
rs781454790 Chr.20: 18596262-18596262 on Build GRCh38
rs984168066 Chr.20: 18596308-18596308 on Build GRCh38
rs751451792 Chr.20: 18596195-18596195 on Build GRCh38
rs992208104 Chr.20: 18596339-18596339 on Build GRCh38
rs935716502 Chr.20: 18596182-18596182 on Build GRCh38
rs6081237 Chr.20: 18596336-18596336 on Build GRCh38
rs939923604 Chr.20: 18596025-18596025 on Build GRCh38
rs764505729 Chr.20: 18596090-18596090 on Build GRCh38
rs779486817 Chr.20: 18596222-18596222 on Build GRCh38
rs980727134 Chr.20: 18596087-18596087 on Build GRCh38
rs764556543 Chr.20: 18596192-18596192 on Build GRCh38
rs1884875 Chr.20: 18596268-18596268 on Build GRCh38
rs776645097 Chr.20: 18596181-18596181 on Build GRCh38
rs149232229 Chr.20: 18596204-18596204 on Build GRCh38
rs570765329 Chr.20: 18596383-18596383 on Build GRCh38
rs779047169 Chr.20: 18595956-18595956 on Build GRCh38
rs769271245 Chr.20: 18596283-18596283 on Build GRCh38
rs917153909 Chr.20: 18596347-18596347 on Build GRCh38
rs145628436 Chr.20: 18596069-18596069 on Build GRCh38
rs758195760 Chr.20: 18595990-18595990 on Build GRCh38
rs756741664 Chr.20: 18596006-18596006 on Build GRCh38

View Full Product Details